TLC Analysis of Hormones on HPTLC Silica Gel 60 RP-18 wF 254s with Concentrating Xone
TLC Analysis of Hormones on HPTLC Silica Gel 60 RP-18 wF 254s with Concentrating Xone
Conditions
sample 5 µL
plate HPTLC pre-coated plate RP-18 wF 254s with concentrating zone (1.13124.0001)
migration distance 5 cm
chamber normal chamber without chamber saturation
eluent [A] acetonitrile; [B] water; (40:60, A:B)
detector Dipped in MnCl2-sulfuric acid heated up to 110 °C for 10 min in situ evaluation
(with TLC/HPTLC scanner (Camag) UV 366 nm)
Description
Categories Analytical Chromatography, TLC Applications, hormones
Featured Industry Life Science and Biopharma
suitability application for TLC