TLC Analysis of Ingredients in Contraceptic Pills in HPTLC DIOL F254s
TLC Analysis of Ingredients in Contraceptic Pills in HPTLC DIOL F254s
Conditions
sample 0.2 µL
migration distance 7 cm
plate HPTLC pre-coated plate DIOL F 254s (1.112668.0001)
chamber normal chamber without chamber saturation
eluent [A] benzene (40-60 ); [B] acetone; (80:20, A:B)
detector
Dipped in MnCl2-sulfuric acid heated up to 110 °C for 10 min
in situ evaluation (with TLC/HPTLC scanner (Camag) UV 366 nm)
Description
Categories Analytical Chromatography, TLC Applications, hormones
Featured Industry Life Science and Biopharma
suitability application for TLC