TLC Analysis of Mycotoxins on HPTLC pre-coated plate RP-8 F254s
TLC Analysis of Mycotoxins on HPTLC pre-coated plate RP-8 F254s
Conditions
sample 0.3 µL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nmdipped in
dichlorofluorescein (0.2% in ethanol)
eluent [A] acetonitrile; [B] buffer solution pH 2.00; (50:50, A:B)
migration distance 8 cm
chamber normal chamber without chamber saturation
plate TLC pre-coated plate RP-8 F 254s (1.15388.0001, 1.15424.0001)
Description
Categories Analytical Chromatography, TLC Applications, mycotoxins
Featured Industry Food and Beverages
General description TLC Plate Information: 1.15388.0001
suitability application for TLC