TLC Analysis of Nucleosides on HPTLC Silica gel 60 RP-18 WF254s
TLC Analysis of Nucleosides on HPTLC Silica gel 60 RP-18 WF254s
Conditions
sample 5 µL (0.005%)
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 265 nm
eluent [A] water; [B] ethanol; (95:5, A:B)
plate HPTLC pre-coated plate RP-18 WF 254s with concentrating zone (1.13124.0001)
migration distance 7 cm
chamber normal chamber without chamber saturation
Description
Categories Analytical Chromatography, TLC Applications, nucleosides
Featured Industry Life Science and Biopharma
suitability suitable for TLC