TLC Analysis of Plasticizers on HPTLC pre-coated plate RP-18 WF 254s with concentrating zone
TLC Analysis of Plasticizers on HPTLC pre-coated plate RP-18 WF 254s with concentrating zone
Conditions
sample 2 µL (0.05%)
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
plate HPTLC pre-coated plate RP-18 WF 254s with concentrating zone (1.13124.0001)
migration distance 7 cm
chamber normal chamber without chamber saturation
eluent [A] methanol; [B] water; (80:20, A:B) containing 0.1 M lithium chloride
Description
Categories Analytical Chromatography, TLC Applications, phthalate esters
Featured Industry Chemicals and Industrial Polymers
Food and Beverages
suitability suitable for TLC