TLC Analysis of Sex Hormone Derivatives on HPTLC CN F254s
TLC Analysis of Sex Hormone Derivatives on HPTLC CN F254s
Conditions
sample 0.2 µL
detector Spray reagent: 5% methanolic perchoric acid heated up to 110 °C for 5 min UV 366 nm
migration distance: 7 cm
chamber normal chamber without chamber saturation
plate HPTLC pre-coated plate CN F 254s (1.16464.0001)
eluent [A] petroleum benzene (40-600); [B] ethanol; (80:20, A:B)
Description
Categories Analytical Chromatography, TLC Applications, hormones
Featured Industry Life Science and Biopharma
General description TLC Plate Information: 1.16464.0001
suitability application for TLC