TLC Analysis of Steroids on HPTLC Silica gel 60 RP-18 WF254s: Effect of Stabilizer
TLC Analysis of Steroids on HPTLC Silica gel 60 RP-18 WF254s: Effect of Stabilizer
Conditions
sample 0.2 µL
detector spray reagent: MnCl2-sulfuric acid heated up to 110 °C for 5 min, in-situ evaluation
with TLC/HPTLC scanner (Camag) UV 265 nm
plate HPTLC pre-coated plate RP-18 WF 254s (1.13124.0001)
eluent chloroform with stabilizer, 2-methyl-2-butene or ethanol as shown
Description
Categories Analytical Chromatography, TLC Applications, steroids
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
General description TLC Plate Information: 1.134124.0001
suitability application for TLC