TLC Analysis of Steroids on HPTLC Silica gel 60 RP-18 F254s
TLC Analysis of Steroids on HPTLC Silica gel 60 RP-18 F254s
Conditions
sample 0.2 mL
detector in-situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
test parameters eluent: [A] acetone; [B] water; (60:40, A:B)
test parameters migration distance: 5 cm
test parameters chamber: normal chamber without chamber saturation
test parameters plate: HPTLC pre-coated plate RP-18 F 254s (1.13724.0001)
Description
Categories Analytical Chromatography, TLC Applications, steroids
Featured Industry Life Science and Biopharma
Pharmaceutical (small molecule)
General description TLC Plate Information: 1.13724.0001
suitability application for TLC