TLC Analysis of Tripeptides on HPTLC Silica Gel 60 RP-8 F254s
TLC Analysis of Tripeptides on HPTLC Silica Gel 60 RP-8 F254s
Conditions
detector spray reagent: ninhydrin, heated to 110 °C for 20 min
eluent [A] acetonitrile; [B] buffer solution, pH 3.00; (90:10, A:B) containing 0.1 M lithium chloride
migration distance 5 cm
chamber normal chamber without chamber saturation
plate TLC pre-coated plate RP-18 F 254s (1.13725.0001)
Description
Categories Analytical Chromatography, TLC Applications, peptides
Featured Industry Life Science and Biopharma
suitability application for TLC