Clinical Application of SPME: Analysis of VOCs in Exhaled Breath as Cancer Biomarkers
By: Bogusław Buszewski, Tomasz Ligor, Joanna Rudnicka, Reporter US Volume 30.3
The following article was generated with the assistance of an outside source using Sigma-Aldrich® products. Technical content was generated and provided by:
Bogusław Buszewski, Tomasz Ligor, Joanna Rudnicka
Department of Environmental Chemistry and Bioanalytics, Faculty of Chemistry Nicolaus Copernicus University, Gagarina 7 St., PL-87-100 Toruń, Polandk
Introduction
Characteristic odors in breath have long been used as a tool for medical diagnosis. Familiar examples are diabetes with the odor of overripe apples, renal diseases with the fishy smell of amines and ammonia, and dental or liver diseases with the cabbage-like odor of organic sulfides[1]. Linus Pauling applied formal science to the analysis by using gas chromatography (GC) to detect volatile organic compounds (VOC) in breath[2]. More recently, Michael Philips and his Menssana coworkers focused on determination of breath compounds that are attracting attention in clinical and toxicological analysis[3]. Although breath analysis is of great importance in disease detection, toxicology, and the study of metabolic processes, its use by doctors and clinicians as a diagnostic tool is a lost art.
The classes of VOCs that can be present in exhaled breath include hydrocarbons, alcohols, ketones, aldehydes, esters, and organic sulfides[4]. The determination of VOCs in exhaled air requires the detection of very low concentrations. Hence, the analytical methods employed must include a preconcentration step. The common preconcentration methodologies currently utilized for VOCs are sorption onto an adsorbent and cold trapping.
However, solid phase microextraction (SPME) is a viable alternative to these methods, as will be shown here. SPME has been widely used for the determination of volatile organic compounds in various matrices, including exhaled breath[5]. Compared to other preconcentration techniques, SPME is simple, inexpensive, and solvent-free. It is fully automatable, and no thermal desorption unit or modifications to the GC instrument are necessary. Compatible with all GC systems, SPME can be used by practically every laboratory. The objective of this study was to use SPME with GC-MS analysis to identify volatile biomarkers of lung cancer.
Experimental
Breath samples were collected from ten healthy volunteers and twelve patients with lung cancer. Each participant provided via questionnaire their age, sex, other diseases, medications, smoking habits, and composition of recent meals. Breath samples were collected in 1 L Tedlar® bags which were kept at a constant 25 °C. A gas standard containing the compounds of interest was made by vaporizing a liquid mixture of the compounds in a glass bulb. A defined volume of the mixture was transferred into the Tedlar bag prior to sampling. During extraction, the CAR/PDMS SPME fiber was introduced into the bag containing breath sample or gas standards through a septum and exposed for 15 minutes. Ambient air samples were collected for background. External calibration was employed. The fiber desorption and sample analysis (GC-MS) conditions are shown in Figure 1.
Figure 1. GC-MS Analysis of VOC in Breath from Lung Cancer Patient after SPME Using CAR /PDMS Fibers
SPME Conditions: sample: exhaled breath, 1 L in Tedlar bag; fiber: Carboxen®/Polydimethylsiloxane (CAR/PDMS), 75 µm film (57318); holder: manual SPME holder (57330-U); extraction: 15 min at 25 °C; desorption: 1 min at 220 °C
GC -MS Conditions: column conditions: GC PLOT column (Q-type), 25 m × 0.25 mm I.D., 3 µm; inj. temp.: 200 °C; oven: 40 °C (2 min), 10 °C/min to 140 °C, 5 °C/min to 270 °C (3 min); carrier gas: helium, 40 cm/sec, constant; injection: splitless 1 min, then split at 35:1; detector: MS, full scan, m/z 15-220, rate 3.46 scans/sec, EI ion source and transfer line temp: 220 °C
column: Ascentis® Express C18, 15 cm x 4.6 mm, 2.7 µm particles (53829-U); mobile phase: (A) 5:95 acetonitrile:water w/ 0.05% formic acid; (B) 95:5 acetonitrile:water w/ 0.05% formic acid; gradient: 40% to 70% B in 3 min; 70% to 90% B in 2 min; hold at 90% for 6 min, 90% to 40% in 0.1 min, hold at 40% for 3.9 min; flow rate: 1 mL/min; column temp.: 35 °C; detector: AB3200 QTrap MS, ESI(+), SIM; injection: 2 µL
  1. Ethanol
  2. Acetonitrile
  3. Acetone
  4. 2-Propanol
  5. Isoprene
  6. Pentane
  7. Methacrolein
    8. Methyl vinyl ketone
  9. Ethyl acetate
10. 2-Methylpentane
11. 3-Methylpentane
12. Hexane
13. Toluene
14. Hexanal
15. Limonene
Figure 1 shows a typical GC-MS chromatogram of breath from a lung cancer patient. Analysis of exhaled air from healthy volunteers and cancer patients identified seventeen volatile compounds, mainly hydrocarbons, ketones, aldehydes, and alcohols. Similar compounds were found in both healthy and cancer patients, except furan derivatives which are considered to be markers for tobacco smoking. Statistical tests were applied to distinguish cancer patients from the healthy control group. The VOC concentration data were log-transformed and tested for normality using the Shapiro-Wilks W test, (p<0.05). Full details of the statistical analysis is beyond the scope of this brief report; however, they are available upon request.
Summarizing: Although there was variation between the patients and not all patients exhibited the same biomarker pattern, four compounds stood out statistically from the others: methyl vinyl ketone, 1-propanol, 2-propanol, and o-xylene. These compounds showed statistically higher levels in cancer patients compared to the healthy control group.
Results
The linearity, precision, and detection limits for VOCs determination in human breath are presented in Table 1. The relative standard deviation (RSD) was in the range of 3.4% to 9.4%. Linear regression coefficient values (r²) were close to 1. The lowest LOD values obtained for hydrocarbons varied from 0.3 to 0.49 ppb.
Table 1. Validation Parameters for Volatile Organic Compounds (VOCs)*
Compounds Liner Range
(ppb)
%RSD
(n=3)
LOD
(ppb)
LOQ
(ppb)
Acetone 1.6 - 920.7 0.991 8.9 0.54 1.62
Acetonitrile 2.3 - 234.0 0.996 3.4 0.75 2.25
Ethyl acetate 1.3 - 136.7 0.995 4.6 0.43 1.29
Methyl vinyl ketone 9.9 - 135.6 0.997 8.4 3.30 9.90
3-Methylpentane 1.3- 136.5 0.997 3.3 0.45 1.35
Ethanol 1.0 - 99.8 0.992 4.5 0.33 1.00
2-Methylfuran 1.6 - 165.6 0.999 4.9 0.54 1.63
2-Methylpentane 0.9 - 87.8 0.992 4.5 0.32 0.96
Hexanal 9.9 - 133.9 0.996 6.2 3.30 9.90
Hexane 1.5 - 150.0 0.994 3.4 0.49 1.47
Isoprene 2.6 - 380.2 0.998 3.7 0.87 2.62
Pentane 1.5 - 150.0 0.998 5.2 0.49 1.47
Methacrolein 11.7 - 170.4 0.998 7.2 3.91 11.73
1-Propanol 1.6 - 163.5 0.995 5.1 0.53 1.59
2-Propanol 1.6 - 159.6 0.998 9.4 0.52 1.57
Toluene 1.1 - 114.7 0.991 5.9 0.41 1.11
o-Xylene 1.0 - 100.1 0.994 4.8 0.33 0.99
* Conditions same as Figure 1, except sample is a mixture of VOC standards.
Bolded compounds had statistical significance.
Conclusion
This brief report is intended to demonstrate the potential of solid phase microextraction (SPME) as a clinical research tool, in this case toward the extraction from human breath of VOCs associated with lung cancer. By using SPME with GC-MS analysis and applying rigorous statistical methods, we found the VOC profiles between a small set of healthy individuals and those diagnosed with lung cancer were significantly different. These promising findings would necessarily be followed up with studies on larger populations for definitive associations. The SPME-GC-MS method presented here had the requisite linearity and sensitivity, and could be easily adopted by laboratories as an investigational tool into biomarker discovery, among many other applications relevant to the clinical and biochemical fields.
Featured Products  
Description Cat. No.
SPME Fiber & Holder  
SPME fiber assembly Carboxen®/Polydimethylsiloxane (CAR/PDMS), needle size 24 ga, coating 75 µm, for use with manual holder 57318
SPME fiber holder for use with manual sampling 57330-U
Capillary GC Column  
Supel™-Q PLOT, 30 m x 0.32 mm I.D. 24242
Analytical Standards  
For a complete listing of our volatile organic standards, visit sigma-aldrich.com/standards  
Acknowledgement
This research work was supported by Grant N N 204 026238 (2010-2013).
References
  1. Risby T.H. Current status of clinical breath analysis. In Breath Analysis for Clinical Diagnosis and Therapeutic Monitoring; Amann, A., Smith, D., Eds.; World Scientific: Singapore, 2005.
  2. Pauling, L.; Robinson, A.B.; Teranishi R.; Cary P. Quantitative analysis of urine vapour and breath by gas-liquid partition chromatography. Proc. Nat. Acad. Sci. 1971, 68(10), 2374-2376.
  3. Phillips, M.; Gleeson, K.; Hughes, J.M.; Greenberg, J.; Cataneo, R.N.; Baker, L., McVay, W.P. Volatile organic compounds in breath as markers of lung cancer: a cross-sectional study. Lancet, 1999, 353 (9168), 1930-1933.
  4. Buszewski, B.; Kêsy, M.; Ligor, T.; Amann, A. Human exhaled air analytics: biomarkers of diseases. Biomed. Chromatogr. 2007, 21, 553-566.
  5. Grote C.; Pawliszyn J. Solid phase microextraction for the analysis of human breath. Anal. Chem. 1997, 69, 587-596.
Trademarks
Tedlar is a registered trademark of E. I. du Pont de Nemours and Company
Carboxen is a registered trademark of Sigma-Aldrich Co. LLC
Supel is a trademark of Sigma-Aldrich Co. LLC