Certified Standard Solutions (CRM) for Photometric Applications
Diluted, Ready-To-Use and in Varying Concentrations
Standards bottlesPrecise and accurate results are crucial, particularly when used for analysis of water and waste water. Photometric rapid tests for in-process controls can be used to test the quality of water without hassle in accordance with DIN, ISO, EN, APHA, EPA and USP, but for reliable and reproducible results, these methods must be checked regularly.
When it comes to analytical quality control, standard solutions that are traceable to globally accepted primary standards are essential. We have developed certified, diluted standard solutions which are traceable to standard reference materials (SRM) from the National Institute of Standards and Technology (NIST) or to United States Pharmacopeia USP.
With our standard solutions (CRM) for photometric applications, TOC analyzers, ICP-OES and Ion chromatography, you can perform your internal analytical quality control reliably and without extra effort. Time-consuming dilution is no longer necessary, since our standard solutions are ready-to-use, certified and come in a range of appropriate concentrations. Most importantly, they provide users with the certainty that they are traceable directly to primary standard reference materials from NIST or USP. This ensures that quality standards are comparable internationally.
Features and Benefits
  • Diluted standard solutions that are ready-to-use for accurate measurements
  • Traceable to SRM from NIST and USP
  • Full certificates of analysis with comprehensive quality documentation
  • Information on concentration and expanded measurement uncertainty
  • Save time and minimize errors
Materials
Standard Solutions | 100 ml or 250 ml | traceable to SRM from NIST or USP
Product No. Description Concentration Expanded Measurement Uncertainty Composition pH range solution Method used for uncertainty measurement Tracable to
132226 Aluminium Standard Solution 0.0500 mg/l Al ± 0.0020 mg/l Al Al(NO3)3 in H2O 3-5 ICP-OES NIST
132225 Aluminium Standard Solution 0.200 mg/l Al ± 0.006 mg/l Al Al(NO3)3 in H2O 3-5 ICP-OES NIST
132227 Ammonium Standard Solution 0.250 mg/l NH4+ ± 0.011 mg/l NH4+ NH4Cl in H2O 4-6 Photometry NIST
125022 Ammonium Standard Solution 0.400 mg/l NH4-N ± 0.012 mg/l NH4-N NH4Cl in H2O 4-6 Photometry NIST
125023 Ammonium Standard Solution 1.00 mg/l NH4-N ± 0.04 mg/l NH4-N NH4Cl in H2O 4-6 Photometry NIST
125024 Ammonium Standard Solution 2.00 mg/l NH4-N ± 0.07 mg/l NH4-N NH4Cl in H2O 4-6 Photometry NIST
125025 Ammonium Standard Solution 6.00 mg/l NH4-N ± 0.13 mg/l NH4-N NH4Cl in H2O 4-6 Photometry NIST
125026 Ammonium Standard Solution 12.0 mg/l NH4-N ± 0.4 mg/l NH4-N NH4Cl in H2O 4-6 Photometry NIST
125027 Ammonium Standard Solution 50.0 mg/l NH4-N ± 1.2 mg/l NH4-N NH4Cl in H2O 4-6 Photometry NIST
133002 Arsenic Standard Solution 1.00 mg/l As ± 0.05 mg/l As H3AsO4 in HNO3 2-3 ICP-OES NIST
133005 Boron Standard Solution 1.00 mg/l B ± 0.06 mg/l B H3BO3 in H2O 5-7 Photometry NIST
133006 Bromate Standard Solution 0.0100 mg/l BrO3- ± 0.0006 mg/l BrO3- KBrO3 in H2O 6-8 IC NIST
133007 Bromate Standard Solution 0.1000 mg/l BrO3- ± 0.0040 mg/l BrO3- KBrO3 in H2O 6-8 IC NIST
133008 Cadmium Standard Solution 0.00500 mg/l Cd ± 0.00200 mg/l Cd Cd(NO₃)₂ in HNO3 3-4 ICP-OES NIST
132228 Cadmium Standard Solution 0.100 mg/l Cd ± 0.003 mg/l Cd Cd(NO₃)₂ in H2O 3-5 ICP-OES NIST
133009 Chloride Standard Solution 0.100 mg/l Cl- ± 0.006 mg/l Cl- NaCl in H₂O 5-7 IC NIST
133100 Chloride Standard Solution 1.00 mg/l Cl- ± 0.04 mg/l Cl- NaCl in H₂O 5-7 IC NIST
133011 Chloride Standard Solution 2.50 mg/l Cl- ± 0.08 mg/l Cl- NaCl in H₂O 5-7 Photometry NIST
132229 Chloride Standard Solution 10.0 mg/l Cl- ± 0.5 mg/l Cl- NaCl in H₂O 5-7 Photometry NIST
132230 Chloride Standard Solution 50 mg/l Cl- ± 3 mg/l Cl- NaCl in H₂O 5-7 Photometry NIST
132231 Chloride Standard Solution 250 mg/l Cl- ± 8 mg/l Cl- NaCl in H₂O 5-7 Photometry NIST
133012 Chromium Standard Solution 0.050 mg/l Cr(VI) ± 0.002 mg/l Cr(VI) K2CrO4 in H2O 6-8 ICP-OES NIST
133013 Chromium Standard Solution 1.00 mg/l Cr(VI) ± 0.03 mg/l Cr(VI) K2CrO4 in H2O 6-8 Photometry NIST
125028 COD Standard Solution 20.0 mg/l COD ± 0.7 mg/l COD Kaliumhydrogenphthalat in H2O 4-6 Photometry NIST
125029 COD Standard Solution 100 mg/l COD ± 3 mg/l COD Kaliumhydrogenphthalat in H2O 4-6 Photometry NIST
125030 COD Standard Solution 200 mg/l COD ± 4 mg/l COD Kaliumhydrogenphthalat in H2O 3-5 Photometry NIST
125031 COD Standard Solution 400 mg/l COD ± 5 mg/l COD Kaliumhydrogenphthalat in H2O 3-5 Photometry NIST
125032 COD Standard Solution 1,000 mg/l COD ± 11 mg/l COD Kaliumhydrogenphthalat in H2O 3-5 Photometry NIST
125033 COD Standard Solution 2,000 mg/l COD ± 32 mg/l COD Kaliumhydrogenphthalat in H2O 3-5 Photometry NIST
125034 COD Standard Solution 8,000 mg/l COD ± 68 mg/l COD Kaliumhydrogenphthalat in H2O 3-5 Photometry NIST
125035 COD Standard Solution 50,000 mg/l COD ± 894 mg/l COD Kaliumhydrogenphthalat in H2O 3-5 Photometry NIST
132234 Fluoride Standard Solution 0.200 mg/l F- ± 0.012 mg/l F- NaF in H2O 4-6 Photometry NIST
132233 Fluoride Standard Solution 0.50 mg/l F- ± 0.02 mg/l F- NaF in H2O 4-6 Photometry NIST
132235 Fluoride Standard Solution 1.00 mg/l F- ± 0.03 mg/l F- NaF in H2O 4-6 Photometry NIST
132236 Fluoride Standard Solution 1.50 mg/l F- ± 0.04 mg/l F- NaF in H2O 4-6 Photometry NIST
133014 Iron Standard Solution 0.0500 mg/l Fe(III) ± 0.0015 mg/l Fe(III) Fe(NO₃)₃ in HNO₃ 2 ICP-OES NIST
133018 Iron Standard Solution 0.1000 mg/l Fe(III) ± 0.0030 mg/l Fe(III) Fe(NO₃)₃ in HNO₃ 2 ICP-OES NIST
133019 Iron Standard Solution 0.300 mg/l Fe(III) ± 0.009 mg/l Fe(III) Fe(NO₃)₃ in HNO₃ 2 Photometry NIST
133020 Iron Standard Solution 1.00 mg/l Fe(III) ± 0.04 mg/l Fe(III) Fe(NO₃)₃ in HNO₃ 2 Photometry NIST
133003 Lead Standard Solution 0.0500 mg/l Pb ± 0.0040 mg/l Pb Pb(NO₃)₂ in HNO₃ 3,5-4,5 ICP-OES NIST
133004 Lead Standard Solution 0.100 mg/l Pb ± 0.005 mg/l Pb Pb(NO₃)₂ in HNO₃ 3,5-4,5 ICP-OES NIST
132237 Manganese Standard Solution 0.050 mg/l Mn ± 0.004 mg/l Mn Mn(NO3)2 in H2O 3-5 Photometry NIST
132238 Manganese Standard Solution 0.200 mg/l Mn ± 0.005 mg/l Mn Mn(NO3)2 in H2O 3-5 Photometry NIST
132239 Manganese Standard Solution 1.00 mg/l Mn ± 0.03 mg/l Mn Mn(NO3)2 in H2O 3-5 Photometry NIST
132240 Nitrate Standard Solution 1.00 mg/l NO3- ± 0.03 mg/l NO3- NaNO3 in H2O 5-7 IC NIST
132241 Nitrate Standard Solution 10.0 mg/l NO3- ± 0.3 mg/l NO3- NaNO3 in H2O 5-7 Photometry NIST
132242 Nitrate Standard Solution 50.0 mg/l NO3- ± 2.0 mg/l NO3- NaNO3 in H2O 5-7 Photometry NIST
125036 Nitrate Standard Solution 0.50 mg/l NO3-N ± 0.05 mg/l NO3-N NaNO3 in H2O 5-7 Photometry NIST
125037 Nitrate Standard Solution 2.50 mg/l NO3-N ± 0.06 mg/l NO3-N NaNO3 in H2O 5-7 Photometry NIST
125038 Nitrate Standard Solution 15.0 mg/l NO3-N ± 0.4 mg/l NO3-N NaNO3 in H2O 5-7 Photometry NIST
125039 Nitrate Standard Solution 40.0 mg/l NO3-N ± 1.0 mg/l NO3-N NaNO3 in H2O 5-7 Photometry NIST
125040 Nitrate Standard Solution 200 mg/l NO3-N ± 5 mg/l NO3-N NaNO3 in H2O 5-7 Photometry NIST
133021 Nitrite Standard Solution 0.0100 mg/l NO2- ± 0.0012 mg/l NO2- NaNO2 in NaOH 8,5-9,5 IC NIST
125041 Nitrite Standard Solution 0.200 mg/l NO2-N ± 0.009 mg/l NO2-N NaNO2 in H2O 5-7 Photometry NIST
125042 Nitrite Standard Solution 40.0 mg/l NO2-N ± 1.3 mg/l NO2-N NaNO2 in H2O 5-7 Photometry NIST
125043 Nitrogen (total) Standard Solution 2.50 mg/l N ± 0.06 mg/l N Glycin in H2O 5-7 Photometry NIST
125044 Nitrogen (total) Standard Solution 12.0 mg/l N ± 0.3 mg/l N Glycin in H2O 5-7 Photometry NIST
125045 Nitrogen (total) Standard Solution 100 mg/l N ± 3 mg/l N Glycin in H2O 5-7 Photometry NIST
125046 Phosphorus Standard Solution 0.400 mg/l PO4-P ± 0.016 mg/l PO4-P Turpinal SL in H2O 4-6 Photometry NIST
125047 Phosphorus Standard Solution 4.00 mg/l PO4-P ± 0.08 mg/l PO4-P Turpinal SL in H2O 3-5 Photometry NIST
125048 Phosphorus Standard Solution 15.0 mg/l PO4-P ± 0.4 mg/l PO4-P Turpinal SL in H2O 2-4 Photometry NIST
125049 Phosphorus Standard Solution 75.0 mg/l PO4-P ± 1.6 mg/l PO4-P Turpinal SL in H2O 2-4 Photometry NIST
132244 Silicate Standard Solution 0.1000 mg/l SiO2 ± 0.0040 mg/l SiO2 SiO2 in H2O 6-8 Photometry NIST
132243 Silicate Standard Solution 0.500 mg/l SiO2 ± 0.025 mg/l SiO2 SiO2 in H2O 6-8 Photometry NIST
132245 Silicate Standard Solution 1.000 mg/l SiO2 ± 0.030 mg/l SiO2 SiO2 in H2O 6-8 Photometry NIST
125050 Sulfate Standard Solution 40 mg/l SO4- ± 6 mg/l SO4- Na2SO4 in H2O 4-6 Photometry NIST
125051 Sulfate Standard Solution 125 mg/l SO4- ± 6 mg/l SO4- Na2SO4 in H2O 4-6 Photometry NIST
125052 Sulfate Standard Solution 400 mg/l SO4- ± 20 mg/l SO4- Na2SO4 in H2O 4-6 Photometry NIST
125053 Sulfate Standard Solution 800 mg/l SO4- ± 27 mg/l SO4- Na2SO4 in H2O 4-6 Photometry NIST
133022 Surfactants nonionic Standard Solution 1.00 mg/l Triton® X-100 ± 0.16 mg/l Triton® X-100 Triton X-100® in H2O 4-6 Photometry USP
133023 Surfactants nonionic Standard Solution 5.00 mg/l Triton® X-100 ± 0.30 mg/l Triton® X-100 Triton X-100® in H2O 4-6 Photometry USP
133024 Surfactants nonionic Standard Solution 10.00 mg/l Triton® X-100 ± 0.60 mg/l Triton® X-100 Triton X-100® in H2O 4-6 Photometry USP
132246 TOC Standard Solution 5.00 mg/l TOC ± 0.10 mg/l TOC Kaliumhydrogenphthalat in H2O 4-6 TOC-Analyzer NIST
132247 TOC Standard Solution 10.0 mg/l TOC ± 0.2 mg/l TOC Kaliumhydrogenphthalat in H2O 4-6 TOC-Analyzer NIST
132248 TOC Standard Solution 25.0 mg/l TOC ± 0.5 mg/l TOC Kaliumhydrogenphthalat in H2O 4-6 TOC-Analyzer NIST
132249 TOC Standard Solution 50.0 mg/l TOC ± 1.0 mg/l TOC Kaliumhydrogenphthalat in H2O 3-5 TOC-Analyzer NIST
132251 TOC Standard Solution 100 mg/l TOC ± 2 mg/l TOC Kaliumhydrogenphthalat in H2O 3-5 TOC-Analyzer NIST
132252 TOC Standard Solution 200 mg/l TOC ± 4 mg/l TOC Kaliumhydrogenphthalat in H2O 3-5 TOC-Analyzer NIST
132253 TOC Standard Solution 500 mg/l TOC ± 10 mg/l TOC Kaliumhydrogenphthalat in H2O 3-5 TOC-Analyzer NIST