Diluted, Certified Standard Solutions (CRM) for Water and Wastewater Analysis
No Dilution. No Doubts. No Delays. Just Precision.
Our new certified standard solutions (CRMs) are perfect for quality control for photometric methods, standards and our Spectroquant® Test Kits. We also offer ready-to-use standards for TOC analyzers, ICP-OES or Ion chromatography. To ensure accurate results, our CRMs can also be used as standards for test kits from other suppliers.
Our CRMs are directly traceable to NIST primary reference materials, and offer an expanded measurement uncertainty for every concentration. Whether you are analyzing drinking water, process water or wastewater, you’ll have no doubts, no dilution, and no delays. Just open a bottle of our certified reference materials and experience the highest standard in precision.
Learn more about Certified Standard Solutions (CRM) for Photometric Applications.
Feature and Benefits
Extended Portfolio
Complete parameters for quality control in photometric analysis of wastewater, drinking water and process water.
Compatible with all Photometric Rapid Tests
Spectroquant® test kits or those from other suppliers.
Spectroquant® Test Kits.
Certificate of Analysis (COA)
Includes batch-specific parameters, exact concentration, expanded measurement uncertainty, and more.
Download your COA
Perfect Combination with Spectroquant® Prove
New class of spectrophotometers for simple, secure and robust water analysis.
Spectroquant® Prove Spectrophotometers
Guideline to Calculate Your Working Tolerance
According to ISO 17025 every lab needs to calculate their working tolerance. Here is a hint for this procedure.
Calculating your working tolerance
Conversion Table
Conversion tables for different parameters.
Download here
Ideal for Validating International Norm Methods
ISO, EN, APHA, and EPA.
Ready to use
Diluted certified reference materials with exact concentrations save time and prevent dilution errors.
Diluted, Certified Standard Solutions Brochure
Our ready-to-use certified reference materials (CRM) are now available for photometry methods and standards for TOC analyzers, ICP-OES or Ion chromatography for example. Save time, and avoid dilution errors for accurate results.
Download the brochure to learn more about our new certified standard solutions.
Materials
Standard Solutions | 100 ml or 250 ml | traceable to SRM from NIST or USP
Product No. Description Concentration Expanded Measurement Uncertainty Composition pH range solution Method used for uncertainty measurement Tracable to
132226 Aluminium Standard Solution 0.0500 mg/l Al ± 0.0020 mg/l Al Al(NO3)3 in H2O 3-5 ICP-OES NIST
132225 Aluminium Standard Solution 0.200 mg/l Al ± 0.006 mg/l Al Al(NO3)3 in H2O 3-5 ICP-OES NIST
132227 Ammonium Standard Solution 0.250 mg/l NH4+ ± 0.011 mg/l NH4+ NH4Cl in H2O 4-6 Photometry NIST
125022 Ammonium Standard Solution 0.400 mg/l NH4-N ± 0.012 mg/l NH4-N NH4Cl in H2O 4-6 Photometry NIST
125023 Ammonium Standard Solution 1.00 mg/l NH4-N ± 0.04 mg/l NH4-N NH4Cl in H2O 4-6 Photometry NIST
125024 Ammonium Standard Solution 2.00 mg/l NH4-N ± 0.07 mg/l NH4-N NH4Cl in H2O 4-6 Photometry NIST
125025 Ammonium Standard Solution 6.00 mg/l NH4-N ± 0.13 mg/l NH4-N NH4Cl in H2O 4-6 Photometry NIST
125026 Ammonium Standard Solution 12.0 mg/l NH4-N ± 0.4 mg/l NH4-N NH4Cl in H2O 4-6 Photometry NIST
125027 Ammonium Standard Solution 50.0 mg/l NH4-N ± 1.2 mg/l NH4-N NH4Cl in H2O 4-6 Photometry NIST
133002 Arsenic Standard Solution 1.00 mg/l As ± 0.05 mg/l As H3AsO4 in HNO3 2-3 ICP-OES NIST
133005 Boron Standard Solution 1.00 mg/l B ± 0.06 mg/l B H3BO3 in H2O 5-7 Photometry NIST
133006 Bromate Standard Solution 0.0100 mg/l BrO3- ± 0.0006 mg/l BrO3- KBrO3 in H2O 6-8 IC NIST
133007 Bromate Standard Solution 0.1000 mg/l BrO3- ± 0.0040 mg/l BrO3- KBrO3 in H2O 6-8 IC NIST
133008 Cadmium Standard Solution 0.00500 mg/l Cd ± 0.00200 mg/l Cd Cd(NO₃)₂ in HNO3 3-4 ICP-OES NIST
132228 Cadmium Standard Solution 0.100 mg/l Cd ± 0.003 mg/l Cd Cd(NO₃)₂ in H2O 3-5 ICP-OES NIST
133009 Chloride Standard Solution 0.100 mg/l Cl- ± 0.006 mg/l Cl- NaCl in H₂O 5-7 IC NIST
133100 Chloride Standard Solution 1.00 mg/l Cl- ± 0.04 mg/l Cl- NaCl in H₂O 5-7 IC NIST
133011 Chloride Standard Solution 2.50 mg/l Cl- ± 0.08 mg/l Cl- NaCl in H₂O 5-7 Photometry NIST
132229 Chloride Standard Solution 10.0 mg/l Cl- ± 0.5 mg/l Cl- NaCl in H₂O 5-7 Photometry NIST
132230 Chloride Standard Solution 50 mg/l Cl- ± 3 mg/l Cl- NaCl in H₂O 5-7 Photometry NIST
132231 Chloride Standard Solution 250 mg/l Cl- ± 8 mg/l Cl- NaCl in H₂O 5-7 Photometry NIST
133012 Chromium Standard Solution 0.050 mg/l Cr(VI) ± 0.002 mg/l Cr(VI) K2CrO4 in H2O 6-8 ICP-OES NIST
133013 Chromium Standard Solution 1.00 mg/l Cr(VI) ± 0.03 mg/l Cr(VI) K2CrO4 in H2O 6-8 Photometry NIST
125028 COD Standard Solution 20.0 mg/l COD ± 0.7 mg/l COD Kaliumhydrogenphthalat in H2O 4-6 Photometry NIST
125029 COD Standard Solution 100 mg/l COD ± 3 mg/l COD Kaliumhydrogenphthalat in H2O 4-6 Photometry NIST
125030 COD Standard Solution 200 mg/l COD ± 4 mg/l COD Kaliumhydrogenphthalat in H2O 3-5 Photometry NIST
125031 COD Standard Solution 400 mg/l COD ± 5 mg/l COD Kaliumhydrogenphthalat in H2O 3-5 Photometry NIST
125032 COD Standard Solution 1,000 mg/l COD ± 11 mg/l COD Kaliumhydrogenphthalat in H2O 3-5 Photometry NIST
125033 COD Standard Solution 2,000 mg/l COD ± 32 mg/l COD Kaliumhydrogenphthalat in H2O 3-5 Photometry NIST
125034 COD Standard Solution 8,000 mg/l COD ± 68 mg/l COD Kaliumhydrogenphthalat in H2O 3-5 Photometry NIST
125035 COD Standard Solution 50,000 mg/l COD ± 894 mg/l COD Kaliumhydrogenphthalat in H2O 3-5 Photometry NIST
132234 Fluoride Standard Solution 0.200 mg/l F- ± 0.012 mg/l F- NaF in H2O 4-6 Photometry NIST
132233 Fluoride Standard Solution 0.50 mg/l F- ± 0.02 mg/l F- NaF in H2O 4-6 Photometry NIST
132235 Fluoride Standard Solution 1.00 mg/l F- ± 0.03 mg/l F- NaF in H2O 4-6 Photometry NIST
132236 Fluoride Standard Solution 1.50 mg/l F- ± 0.04 mg/l F- NaF in H2O 4-6 Photometry NIST
133014 Iron Standard Solution 0.0500 mg/l Fe(III) ± 0.0015 mg/l Fe(III) Fe(NO₃)₃ in HNO₃ 2 ICP-OES NIST
133018 Iron Standard Solution 0.1000 mg/l Fe(III) ± 0.0030 mg/l Fe(III) Fe(NO₃)₃ in HNO₃ 2 ICP-OES NIST
133019 Iron Standard Solution 0.300 mg/l Fe(III) ± 0.009 mg/l Fe(III) Fe(NO₃)₃ in HNO₃ 2 Photometry NIST
133020 Iron Standard Solution 1.00 mg/l Fe(III) ± 0.04 mg/l Fe(III) Fe(NO₃)₃ in HNO₃ 2 Photometry NIST
133003 Lead Standard Solution 0.0500 mg/l Pb ± 0.0040 mg/l Pb Pb(NO₃)₂ in HNO₃ 3,5-4,5 ICP-OES NIST
133004 Lead Standard Solution 0.100 mg/l Pb ± 0.005 mg/l Pb Pb(NO₃)₂ in HNO₃ 3,5-4,5 ICP-OES NIST
132237 Manganese Standard Solution 0.050 mg/l Mn ± 0.004 mg/l Mn Mn(NO3)2 in H2O 3-5 Photometry NIST
132238 Manganese Standard Solution 0.200 mg/l Mn ± 0.005 mg/l Mn Mn(NO3)2 in H2O 3-5 Photometry NIST
132239 Manganese Standard Solution 1.00 mg/l Mn ± 0.03 mg/l Mn Mn(NO3)2 in H2O 3-5 Photometry NIST
132240 Nitrate Standard Solution 1.00 mg/l NO3- ± 0.03 mg/l NO3- NaNO3 in H2O 5-7 IC NIST
132241 Nitrate Standard Solution 10.0 mg/l NO3- ± 0.3 mg/l NO3- NaNO3 in H2O 5-7 Photometry NIST
132242 Nitrate Standard Solution 50.0 mg/l NO3- ± 2.0 mg/l NO3- NaNO3 in H2O 5-7 Photometry NIST
125036 Nitrate Standard Solution 0.50 mg/l NO3-N ± 0.05 mg/l NO3-N NaNO3 in H2O 5-7 Photometry NIST
125037 Nitrate Standard Solution 2.50 mg/l NO3-N ± 0.06 mg/l NO3-N NaNO3 in H2O 5-7 Photometry NIST
125038 Nitrate Standard Solution 15.0 mg/l NO3-N ± 0.4 mg/l NO3-N NaNO3 in H2O 5-7 Photometry NIST
125039 Nitrate Standard Solution 40.0 mg/l NO3-N ± 1.0 mg/l NO3-N NaNO3 in H2O 5-7 Photometry NIST
125040 Nitrate Standard Solution 200 mg/l NO3-N ± 5 mg/l NO3-N NaNO3 in H2O 5-7 Photometry NIST
133021 Nitrite Standard Solution 0.0100 mg/l NO2- ± 0.0012 mg/l NO2- NaNO2 in NaOH 8,5-9,5 IC NIST
125041 Nitrite Standard Solution 0.200 mg/l NO2-N ± 0.009 mg/l NO2-N NaNO2 in H2O 5-7 Photometry NIST
125042 Nitrite Standard Solution 40.0 mg/l NO2-N ± 1.3 mg/l NO2-N NaNO2 in H2O 5-7 Photometry NIST
125043 Nitrogen (total) Standard Solution 2.50 mg/l N ± 0.06 mg/l N Glycin in H2O 5-7 Photometry NIST
125044 Nitrogen (total) Standard Solution 12.0 mg/l N ± 0.3 mg/l N Glycin in H2O 5-7 Photometry NIST
125045 Nitrogen (total) Standard Solution 100 mg/l N ± 3 mg/l N Glycin in H2O 5-7 Photometry NIST
125046 Phosphorus Standard Solution 0.400 mg/l PO4-P ± 0.016 mg/l PO4-P Turpinal SL in H2O 4-6 Photometry NIST
125047 Phosphorus Standard Solution 4.00 mg/l PO4-P ± 0.08 mg/l PO4-P Turpinal SL in H2O 3-5 Photometry NIST
125048 Phosphorus Standard Solution 15.0 mg/l PO4-P ± 0.4 mg/l PO4-P Turpinal SL in H2O 2-4 Photometry NIST
125049 Phosphorus Standard Solution 75.0 mg/l PO4-P ± 1.6 mg/l PO4-P Turpinal SL in H2O 2-4 Photometry NIST
132244 Silicate Standard Solution 0.1000 mg/l SiO2 ± 0.0040 mg/l SiO2 SiO2 in H2O 6-8 Photometry NIST
132243 Silicate Standard Solution 0.500 mg/l SiO2 ± 0.025 mg/l SiO2 SiO2 in H2O 6-8 Photometry NIST
132245 Silicate Standard Solution 1.000 mg/l SiO2 ± 0.030 mg/l SiO2 SiO2 in H2O 6-8 Photometry NIST
125050 Sulfate Standard Solution 40 mg/l SO4- ± 6 mg/l SO4- Na2SO4 in H2O 4-6 Photometry NIST
125051 Sulfate Standard Solution 125 mg/l SO4- ± 6 mg/l SO4- Na2SO4 in H2O 4-6 Photometry NIST
125052 Sulfate Standard Solution 400 mg/l SO4- ± 20 mg/l SO4- Na2SO4 in H2O 4-6 Photometry NIST
125053 Sulfate Standard Solution 800 mg/l SO4- ± 27 mg/l SO4- Na2SO4 in H2O 4-6 Photometry NIST
133022 Surfactants nonionic Standard Solution 1.00 mg/l Triton® X-100 ± 0.16 mg/l Triton® X-100 Triton X-100® in H2O 4-6 Photometry USP
133023 Surfactants nonionic Standard Solution 5.00 mg/l Triton® X-100 ± 0.30 mg/l Triton® X-100 Triton X-100® in H2O 4-6 Photometry USP
133024 Surfactants nonionic Standard Solution 10.00 mg/l Triton® X-100 ± 0.60 mg/l Triton® X-100 Triton X-100® in H2O 4-6 Photometry USP
132246 TOC Standard Solution 5.00 mg/l TOC ± 0.10 mg/l TOC Kaliumhydrogenphthalat in H2O 4-6 TOC-Analyzer NIST
132247 TOC Standard Solution 10.0 mg/l TOC ± 0.2 mg/l TOC Kaliumhydrogenphthalat in H2O 4-6 TOC-Analyzer NIST
132248 TOC Standard Solution 25.0 mg/l TOC ± 0.5 mg/l TOC Kaliumhydrogenphthalat in H2O 4-6 TOC-Analyzer NIST
132249 TOC Standard Solution 50.0 mg/l TOC ± 1.0 mg/l TOC Kaliumhydrogenphthalat in H2O 3-5 TOC-Analyzer NIST
132251 TOC Standard Solution 100 mg/l TOC ± 2 mg/l TOC Kaliumhydrogenphthalat in H2O 3-5 TOC-Analyzer NIST
132252 TOC Standard Solution 200 mg/l TOC ± 4 mg/l TOC Kaliumhydrogenphthalat in H2O 3-5 TOC-Analyzer NIST
132253 TOC Standard Solution 500 mg/l TOC ± 10 mg/l TOC Kaliumhydrogenphthalat in H2O 3-5 TOC-Analyzer NIST