GC Analysis of Fatty Acids in Walnuts and Peanuts
By: Katherine K. Stenerson, Reporter US Volume 31.3
Nuts are considered nutrient dense foods and their consumption has been associated with reduced risk of coronary heart disease. The health benefits of nuts are partially attributable to their high content of unsaturated fatty acids. For example, α-linolenic acid, or “ALA”, is an unsaturated fatty acid found in flax seeds and walnuts1. ALA is a precursor to the formation within the body of the essential fatty acids eicosapentaenoic acid (EPA) and docosahexaenoic acid (DHA)2.
Established methods from AOCS® and AOAC® can be used to determine the fatty acid composition of nuts. This work demonstrates the application of aspects of these methods in the determination of the fatty acid composition of walnuts and peanuts. We will present data showing analysis on GC capillary columns of three different selectivities: Omegawax®, SP™-2560, and SLB®-IL111.
Column Selectivity for Fatty Acid Methyl Esters
The moderately polar Omegawax is made with a polyethylene glycol (PEG) based phase. It elutes fatty acid methyl esters (FAMEs) by degree of unsaturation, with minimal overlap between different carbon chain lengths. It is used in applications requiring analysis of saturated, mono and polyunsaturated fatty acids (PUFAs). However, it cannot provide optimal resolution of cis and trans isomer groups. For analysis of cis/trans FAMEs, a more polar column is required. The SP-2560 is a highly polar cyanosilicone column. The selectivity of this phase enables it to resolve cis and trans isomers, along with providing positional geometric isomer separations. The SLB-IL111 is an ionic liquid column and has higher polarity than the SP-2560. In the analysis of cis/trans FAMEs, the SLB-IL111 has demonstrated elution patterns which are complimentary to the SP-2560. We used the Omegawax in the profiling of the fatty acids (including PUFAs) present in walnuts and peanuts. The SP-2560 and SLB-IL111 columns were then used to determine C18:1 cis/trans isomers.
Experimental
1 g samples of walnuts (shelled and chopped) and peanuts (dry roasted and unsalted) were prepared using acid digestion/alkali hydrolysis followed by methylation as described in AOCS Official Method Ce 1k-093. BHT was added as an antioxidant prior to extraction. All samples were concentrated to 1 mL prior to GC analysis. The GC columns used for the analysis were as follows:
  1. Omegawax®, 30 m x 0.25 mm I.D., 0.25 µm (Product No. 24136)
  2. SP™-2560, 100 m x 0.25 mm I.D., 0.20 µm (Product No. 24056)
  3. SLB®-IL111, 100 m x 0.25 mm I.D., 0.20 µm (Product No. 29647-U)
The GC analysis conditions were from AOCS Ce 1i-07 (Omegawax column), and AOCS Ce 1h-05 (SP-2560 column)4,5. Peak identifications were done by retention time matching to the Supelco 37-Component FAME Mix (Product No. CRM47885), run under the same GC conditions. This work was previously published6,7.
Results and Discussion
Analysis on the Omegawax Column
Chromatograms of walnut and peanut extracts analyzed on the Omegawax column are presented in Figures 1 and 2. This column would be used if determining saturated and cis-unsaturated fatty acids per AOCS method Ce-1i-07. The relative percentages of the fatty acids detected in the extracts were calculated as percent of total fatty acid methyl ester (FAME) area. Table 1 shows the percentages calculated compared to the expected fatty acid compositions for walnut and peanut oils.
FAMEs in Walnut Extract on the Omegawax
Figure 1. FAMEs in Walnut Extract on the Omegawax
GC Conditions
column:
Omegawax, 30 m x 0.25 mm I.D., 0.25 µm (Product No. 24136); oven: 170 °C, 1 °C/min to 225 °C; inj. temp.: 250 °C; detector: FID, 260 °C; carrier gas: helium, 1.2 mL/min constant flow; injection: 1 µL, 100:1 split; liner: 4 mm I.D., split/splitless type, wool packed single taper FocusLiner™ design; sample: extract of walnuts
FAMEs in Peanut Extract on the Omegawax
Figure 2. FAMEs in Peanut Extract on the Omegawax
Conditions
sample: extract of peanuts; All other conditions same as Figure 1
Table 1. Fatty Acid Compositions: Published Data for Nut Oils vs. Experimentally Determined from Whole Nuts
  Walnut Oil:
Published
Data8
Walnut Extract:
Experimentally
Determined
Peanut Oil:
Published
Data8
Peanut Extract:
Experimentally
Determined
C14:0 0–0.1% 0.1%
C16:0 7–8% 9.0% 8.3–14% 12.7%
C16:1 0.1–0.2% 0.1% 0–0.2% 0.1%
C18:0 1.8–2.2% 2.7% 1.9–4.4% 2.9%
C18:1 cis 17–19% 14.3% 36.4–67.1% 54.8%
C18:2 56–60% 59.4% 14.0–43.0% 23.9%
C18:3 13–14% 14.0% 0–0.1% 0.1%
C20:0 0.1% 0.1% 1.1–1.7% 1.1%
C20:1 0.2% 0.2% 0.7–1.7% 0.9%
C22:0 0.1% 0.1% 2.1–4.4% 1.9%
C22:1 0–0.3% 0.2%
C24:0 1.1–2.2% 1.1%
C24:1 0–0.3% 0.4%
The fatty acid profiles obtained were in good agreement with the published data8. In walnuts, as expected, linoleic (C18:2n6c) was the most abundant fatty acid, followed by oleic (C18:1n9c), ALA (C18:3n3) and palmitic (C16:0). Walnuts are considered a significant plant source of ALA. Both ALA and linoleic acid are the predominant essential fatty acids in humans2. For peanuts, the most abundant fatty acid was oleic, followed by linoleic and palmitic. The health benefits of peanuts and peanut oil are associated with their high oleic acid content. Oleic acid is believed to help lower cholesterol and reduce the risk of heart disease. Oleic acid, also found in olive oil, is a major contributor to the health benefits associated with the “Mediterranean Diet”9.
Analysis on SP-2560 and SLB-IL111 Columns
The FAME composition of vegetable oils is normally determined using AOCS Method Ce 1h-05, which designates the use of a highly polar cyanosilicone column5. The SP-2560 column is specifically mentioned in this method. The use of a highly polar column enables simultaneous determination of trans isomers of unsaturated fatty acids along with saturated and cis-unsaturated. Since our sample extracts were obtained from whole nuts, no trans fats were expected to be present. This was confirmed by analysis on both the SP-2560 and SLB-IL111 columns for walnuts (Figures 3 and 4) and peanuts (not shown). For maximum sensitivity, the undiluted sample extract was injected under the conditions indicated in the method. As indicated, no trans isomers of the C18:1n9 or C18:2n6 FAMEs were detected using either column.
FAMEs in Walnut Extract on the SP-2560
Figure 3. FAMEs in Walnut Extract on the SP-2560
Conditions
column:
SP-2560, 100 m x 0.25 mm I.D., 0.20 µm (Product No. 24056); oven: 180 °C isothermal; inj. temp.: 250 °C; detector: FID, 260 °C; carrier gas: helium, 1.0 mL/min; injection: 1 µL, 100:1 split; liner: 4 mm I.D., split/splitless type, wool packed single taper FocusLiner™ design; sample: extract of walnuts
FAMEs in Walnut Extract on the SLB-IL111
Figure 4. FAMEs in Walnut Extract on the SLB-IL111
Conditions
column: SLB-IL111, 100 m x 0.25 mm I.D., 0.20 µm (Product No. 29647-U); oven: 168 °C isothermal; All other conditions the same as Figure 3
Conclusion
The data presented here demonstrates the high PUFA content found in both walnuts and peanuts. The Omegawax provided the selectivity necessary to elute the FAMEs found in the nut extracts by chain length and degree of unsaturation. Confirmation of the absence of trans fatty acids was achieved through use of the highly polar SP-2560 and confirmed with the extremely polar SLB-IL111.
Legal Information
AOCS is aregistered trademark of The American Oil Chemists’ Society
AOAC is a registered trademark of AOAC INTERNATIONAL
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
Omegawax and SLB are registered trademarks of Sigma-Aldrich Co. LLC
SP is a trademark of Sigma-Aldrich Co. LLC
Materials
 References
  1. Ros, E. Health Benefits of Nut Consumption. Nutrients 2010, 2, 652–682.
  2. McKay, Diane L.; Sibley, Donna. Omega-3 Fatty Acids From Walnuts. Nutrition Dimension, April 2009, 4th edition.
  3. AOCS Official Method Ce 1k-09. Direct Methylation of Lipids in Foods for the Determination of Total Fat, Saturated, cis-Monounsaturated, cis-Polyunsaturated and trans Fatty Acids by Gas Chromatography. AOCS Official Methods and Recommended Practices, 2011–2012 Method Additions and Revisions.
  4. AOCS Official Method Ce 1i-07. Determination of Saturated, cis-Monounsaturated and cis-Polyunsaturated Fatty Acids in Marine and Other Oils Containing Long Chain Polyunsaturated Fatty Acids (PUFAs) by Capillary GLC. AOCS Official Methods and Recommended Practices.
  5. AOCS Official Method Ce 1h-05. Determination of cis-, trans-, Saturated, Monounsaturated and Polyunsaturated Fatty Acids in Vegetable or Nonrunimant Animal Oils and Fats by Capillary GLC. AOCS Official Methods and Recommended Practices.
  6. K.K. Stenerson, L.M. Sidisky and M.D. Buchanan. Comparison of the Supelco 37-Component FAME Mix on the SP-2560 and SLB-IL111. Supelco Reporter 29.3 (June 2011), pages 8–10.
  7. K.K. Stenerson, M.R. Halpenny, L.M. Sidisky and M.D. Buchanan. Ionic Liquid GC Column Option for the Analysis of Omega 3 and Omega 6 Fatty Acids. Supelco Reporter 31.1 (February 2013), pages 10–11.
  8. Physical and Chemical Characteristics of Oils, Fats and Waxes; David Firestone, Eds.; AOCS Press, 1999; pp 74, 107.
  9. “Researchers Say”. ScienceDaily,10 Jan.2005. Sciencedaily.com (4 Apr. 2012).