Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Ammonium in Curative Water
Prove InstrumentsPhotometric determination using the indophenol blue method
Sample Preparation
Titriplex® III solution
Dissolve 12 g Titriplex® III and 2.6 g sodium hydroxide in 100 ml of water for analysis.
Curative water often contents an high amount of alkaline earth metals. The analysis may be hindered by the precipitation of alkaline earth hydroxides. In such cases, after allowing a sufficient time for the color to develop, add dropwise Titriplex® III solution to dissolve the precipitation. Treat the blank reading also with the same amount of Titriplex® III. The analysis is not interfered by Ca - contents up to 1000 mg/l.
Analysis
Determine with the below mentioned test kits.
Calculation
Ammonium content in mg/l NH4 = analysis value in mg/l NH4
Materials