Ammonium in Disposal Drainage Water
Prove InstrumentsPhotometric determination using the indophenol blue method
Sample Preparation
Dilution 1:100 for influx and 1:10 for efflux.
Analysis
Determine with the below mentioned test kits.
Calculation
Ammonium content in mg/l NH4 = analysis value in mg/l NH4 x 100 or x 10 depending on the dilution.
Materials
Product 114544 is not compatible with Move 100.