Ammonium in Manure
Prove InstrumentsPhotometric determination using the indophenol blue method
Sample Preparation
Folded filter
Membrane filter with 0.45 μm pore diameter
Put 10 ml of the sample into a 1000-ml volumetric flask, fill up to volume with water for analysis and mix. After the solid parts have settled, take 10 ml of the supernatant solution and, mixed with a spatula - tipfull of charcoal activated for soil tests, put it into a 50-ml volumetric flask and fill up to volume with water for analysis. Before determination filter the charcoal through a folded filter. In case the solution remains cloudy, filter again through a membrane filter.
Analysis
Determine with the below mentioned test kits.
Calculation
Ammonium content in mg/kg NH4 = analysis value in mg/l NH4 x 2000
Materials
Product 114544 is not compatible with Move 100.