Nitrogen (Total) in Soils
Prove InstrumentsPhotometric determination subsequent to extraction with calcium chloride solution
Calcium chloride solution 0.025 mol/l
Dissolve 3.68 g calcium chloride dihydrate with 1 l of water for analysis.
Sample Preparation
In a glass bottle mix 50 g of naturally moist sample, free from coarse stones, with 100 ml of a calcium chloride solution 0.025 mol/l. Add a spatula-tipfull of charcoal activated for soil tests and shake the closed bottle in a shaking machine for 1 hour (alternative stir in a beaker). Let the suspension settle and filter it through a folded filter. For determination of the water-content dry a similar sample to constant weight in the drying kiln at 105 °C. A drying of the sample before determination is not advisable because of the fast change in the nitrogen-forms. The analysis should take place very quickly. Storing the sample in plastic bags at room temperature changes the analysis values after a short time already.
Analysis
Determine with the below mentioned test kits.
Calculation
Nitrogen (total) content in mg/kg N = analysis value in mg/l N x 2
Materials
Products 100613 and 114763 are not compatible with Move 100.