Spectroquant® Prove 600 Spectrophotometer
For Process Water Analysis
Spetroquant® Prove 600: Complex Analysis from a Powerful UV/VIS Spectrophotometer
Designed for process water analysis the Spectroquant® Prove 600 is perfect for complex kinetics and spectral measurements. The Prove 600 is compatible with 16 mm round cells as well as 10, 20, 50 and 100 mm cells for exceptional analysis sensitivity. The spectrophotometer is pre-programmed for over 180 Spectroquant® kits and methods including sensitive chloride, silicate and sulfate tests to help avoid damage to cooling and boiler water systems. The specially designed cell port has ambient light protection so there is no need to close the lid and the Prove 600 is resistant to most chemicals used in process water analysis. Multiple application notes are available for the Spectroquant® photometers. For example, a unique photometric bromate application for the analysis of drinking water.
Built In Analytical Quality Control and Documentation
All Spectroquant® cell and reagent tests are labeled with the data-rich Live ID system. The Live ID code transmits test method, lot number, expiry date, and calibration updates directly to the spectrophotometer. Results are automatically calculated. The built in AQA Prime software provides complete verification and documentation for all stages of quality control. A clear list of filterable results is displayed on the smart screen. Brief overviews to complete protocols and results can easily be exported in the format of your choice.
Ordering Information
Product No. Description
173018 Spectroquant® Prove 600
Not sure whether the Spectroquant® Prove 100 is the right spectrophotometer for your lab? Compare our various Spectroquant® Prove spectrophotometers, view the Prove features at a glance, and request more information.