Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Spectroquant® Prove Connect
Our Spectroquant® spectrophotometers offer easy analysis of drinking, waste and process water with over 200 pre-programmed Spectroquant® test kits.
You have multiple ways to easily move your data to other devices. Use the USB for the connection to your printer. Automate and simplify your result transfer with one of our new software products: Either transfer your results into a pre-existing LIMS system or connect to our new dashboard service. The two new connectivity solutions are:
  • Prove Connect to LIMS
  • Prove Connect to Dashboard
Spectroquant® Prove Connect to LIMS
Spectroquant® Prove Connect to LIMS
  • Allows easy link to LIMS (Laboratory Information Management System)
  • Offers flexible integration to other systems as ELN (Electronic Lab Notebook)
  • Simply connect up by using LAN
  • Supports data formats: CSV, XML and AnIML
Compatibility
Prove Connect supports the following instruments:
Spectroquant® Prove Connect to LIMS can be used with the following instruments:
  • Microsoft Windows 7, Windows 10, Windows Server 2012
  • Oracle Java Runtime version 8 or higher
  • 1 GB of free disk space
  • 4 GB of RAM
Other configurations may also work but are not supported by Merck.
Item number “Prove Connect to LIMS“ Y1100860001
Manual Spectroquant Prove Connect to LIMS
Configurations   System
Prove Connect to Dashboard
“Prove Connect to Dashboard” connects your Spectroquant® Prove instruments directly to a Dashboard service. You can access the Dashboard application simply by opening a dashboard in your usual browser. There you get a clear overview of your connected Prove instruments, registered users, and results. Trend analysis and report generation is also possible.
Clear advantages:
  • Back up Spectroquant® Prove results directly to our Dashboard Service
  • Gain access to your data: mobile and secure
  • Create and present your data in reports and trend analysis
  • Use our supporting sample preparation workflows and pictograms
Sample Preparation  Open Measurement
Compatibility
Prove Connect to Dashboard is available for the following instruments:
Each instrument is “ready” to be connected with these two buttons in the system menu of each Prove instrument:
All you need is the Prove Connect Dashboard application to connect your device to the Prove instrument:
Item number Y1100850001
Manual Spectroquant Prove Connect to Dashboard