Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Glucuronide Standards
Many drugs are rapidly and extensively metabolized to their glucuronide conjugates for excretion. Glucuronide reference standards are suitable for use in the manufacture of hydrolysis controls for GC/MS or LC/MS methods. They can also be used in the production of calibrators or controls in the direct analysis of glucuronides by LC-MS/MS. Direct detection of glucuronides by LC-MS/MS methods decreases sample-prep time by eliminating the hydrolysis step, which can improve accuracy of results.
Sigma-Aldrich's Cerilliant brand offers a wide selection of Certified Spiking Solutions® and Certified Reference Materials (CRMs) for native and stable-labeled glucuronides of numerous drug classes including:
● Opiates ● Anesthetics
● Steroids ● Hormones
● Benzodiazepines ● Ethanol
● Synthetic analgesics ● Phytochemicals
● Synthetic analgesics ● Cannabinoids
We provide a comprehensive Certificate of Analysis (COA) with each product that includes all analytical data on homogeneity, accuracy of concentration as compared to a calibration curve, and uncertainty and traceability information to support regulatory requirements.
Materials
Some products may not be globally available
Related Links