Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Buffer Solutions for Volumetric Karl Fischer Titration
In addition to complete dissolution of the sample, an optimum pH of 5-8 is of decisive importance for accurate water determination according to Karl Fischer. In most cases, the buffering of the reaction medium by the Karl Fischer reagent used will be sufficient. With some samples, however, such as strong bases or acids, the pH is shifted into the acid or alkaline range. In such cases, an additional buffer solution must be added to maintain the pH in the optimum range for the Karl Fischer reaction. We provide two ready-to-use buffer solutions for use in Karl Fischer titrations: our Aquastar® buffer solution for strong acids and an Aquastar® buffer solution for strong bases.
Materials
If you have trouble finding the products you're looking for, please contact our customer support for assistance.