Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Removal of Detergents from Biological Product Matrices
By: J. Patrick Myers, US Reporter Volume 30.3
Most modern biopharmaceutical research and manufacturing requires the use of one or more detergents. Detergents are amphipathic molecules having both hydrophilic and hydrophobic properties. These unique properties are used to alter the interactions between biological molecules in aqueous solutions.
Detergents are organized into broad classes based on the nature of the polar, hydrophilic part of the molecule. The classes include ionic (further broken down into negatively charged anionic and positively charged cationic), nonionic (uncharged), and zwitterionic (having both positively and negatively charged groups, but with a net charge of zero). Under the proper conditions, detergents form micelles in water.
A micelle is a three dimensional structure formed when the concentration of a detergent increases in water beyond the amount that can form a monolayer on the surface. Micelles are typically spheres in which the hydrophobic part of the detergent molecules associate with each other, while the hydrophilic parts of the molecules contact the water[1]. The number of detergent molecules per micelle (aggregation number) and the range of detergent concentration above which micelles form (called the critical micelle concentration, CMC) are properties specific to each particular detergent. The critical micelle temperature (CMT) is the lowest temperature at which micelles can form. Below the CMT, detergent molecules exist in a crystalline form in solution and are not effective. Above the CMT, detergents form micelles and become effective in disrupting lipid-lipid interactions.
The molecules that make up biological membranes are also amphipathic[2]. They are made up of polar “heads” attached to lipophilic, hydrophobic “tails”. Stable membranes are formed when the “tails” of the molecules associate with each other in a bilayer sheet-like arrangement, leaving the polar “heads” to contact the polar, aqueous milieu on both sides of the membrane. Additional molecules embedded in the lipid bi-layer, including proteins and cholesterol, are important to the flexibility and function of the membrane. The formation of detergent micelles is important because it allows the dispersion of water-insoluble, hydrophobic compounds into aqueous solutions. It also allows the disruption of biological membranes and extraction of proteins. The disintegration of membranes occurs by disrupting the lipid-lipid and lipid-protein interactions in the membrane.
In biological research and biopharmaceutical manufacturing, detergents are used for many purposes. They include stabilizing proteins and nucleic acids during electrophoresis; dissolving membrane proteins; prevention of nonspecific binding in affinity purification and immunoassay procedures; control of protein crystallization; and disruption of cell membranes to release membrane bound and intracellular proteins.
In biopharmaceutical production, the detergent must be removed from the product during downstream processing activities[3,4,5]. Methods for removing detergents include gel filtration, size exclusion, dialysis, and hydrophobic adsorption chromatography. Data presented in Tables 1 and 2 demonstrate that pre-packed, sterile, endotoxin-free Porozorb™ cartridges can also be used to effectively remove detergents and other nonpolar, hydrophobic materials from biological preparations.
Table 1 shows the capacity of various Porozorb cartridges for several common detergents used in biopharmaceutical applications. Table 2 demonstrates the time required to reduce a 0.1% concentration of one of those detergents, Triton® X-100, in a 10 liter solution to below detectable levels.
Table 1. Capacity of Porozorb Cartridges for Various Detergents
          Calculated Saturation Capacity
Porozorb
Cartridge
Cat. No.   Bed Volume
(mL)
ID (cm) Length (cm) Triton X-100
(g)/ cartridge
SDS (g)/
cartridge
CHAPS (g)/
cartridge
254 57500 250 8 6.55 44 125 78
1004 57502 1000 8 26.2 168.8 462 288
4004 57513-U 4000 12.71 42.04 504 1400 873.6
Table 2. Time Required to Reduce 0.1% Triton X-100 Detergent in 10-Liter Solution to Below Detection Level.
(Detergent detected by monitoring absorbence at 275nm)
  Flow Rate 3.6 liter/hour Flow Rate 10.3 liter/hour Flow Rate 22.8 liter/hour
Bed Volume
(mL)
Breakthrough Volume
(liter)
Recirculation Time
(hour)
Breakthrough Volume
(liter)
Recirculation Time
(hour)
Breakthrough Volume
(liter)
Recirculation Time
(hour)
250 3 11 <1 6 <1 3
1000 none 9 50 3.5 1 2
4000 none 7 none 2 >40 1.5
The AMBERLITE® XAD-4 resin (Figure 1) used in Porozorb™ cartridges is a proven technology that is highly effective in removing various detergents from cell culture media in biopharmaceutical applications, such as vaccine production.
Figure 1. Structure of Hydrophobic Macroreticular Resin Bead
Porozorb cartridges (Figure 2) are disposable, and available in sizes appropriate for analytical to process scale purification schemes.
Figure 2. Disposable Porozorb Cartridges (4 L and 1 L)
The AMBERLITE XAD-4 resin contained in the Porozorb™ cartridge is cleaned without the use of organics, and it is also tested for residues (CFR 21: 173.65)[6] and organics. Each cartridge is packaged separately, and is accompanied by a Certificate of Analysis for sterility and endotoxicity. Sterility testing follows USP XXIII guidelines. Endotoxicity is tested using a validated LAL kit, with any detectable level of endotoxin (0.03 EU/mL) indicating a positive test. Porozorb cartridges can also be packed with a variety of other resins and media to meet your specific application needs. In addition, we can provide bulk materials and/or smaller pre-packed devices for method development — contact Supelco custom services.
References
  1. http://www.piercenet.com/browse.cfm?fldID=5558F7E4-5056-8A76-4E55- 4F3977738B63
  2. http://wolfson.huji.ac.il/purification/PDF/detergents/ANATRACE_DetergentsUse.pdf
  3. Horigome, T. Sugano, H., A rapid method for removal of detergents from protein solution. Analytical Biochemistry, Volume 130, Issue 2, 15 April 1983, pages 393-396.
  4. http://www.rohmhaas.com/ionexchange/pharmaceuticals/detergent_removal.htm
  5. Trescec, A.,et al., Removal of Detergent and Solvent from Solvent-Detergent-treated Immunoglobulins, .Chromatogr. A, 852(1999) 87-91
  6. Electronic Code of Federal Regulations; 21 CFR 173.65, www.ecfr.gov
Related Information
For our complete offering of products and services for chromatographic purification, visit sigma-aldrich.com/purification
Trademarks
Porozorb is a trademark of Sigma-Aldrich Co. LLC
AMBERLITE and Triton are registered trademarks of The Dow Chemical Company or an affiliated company of Dow