Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Supel™-Select Polymeric SPE
HLB and Ion-Exchange phases for a wide range of applications and pH conditions
Supel-Select SPE phases are ideal for the solid phase extraction of a broad range of compounds from aqueous samples. While reversed-phase interactions dominate retention on the Supel-Select HLB, and the retention mechanisms of the Supel-Select SAX and SCX are predominately based on ion-exchange, the hydrophilic modifications of the styrene-based polymer backbone allow for retention and recovery of more polar compounds.
HLB Phase Chemistry:  Hydrophilic modified styrene polymer
SAX Phase Chemistry:  Quaternary amine functionalized hydrophilic modified styrene polymer
SCX Phase Chemistry:  Sulfonic acid functionalized hydrophilic modified styrene polymer
pH Compatibility:  0-14
Particle Size:  50-70 µm
MS Suitability:  Yes
Surface Area:  160-420 m2/g
Pore Volume: 0.8-1.2 mL/g
Pore Size: 80-200 Å
Feature Benefit
Hydrophilic modified styrene resin Extract and recover a broad range of analytes (polar to non-polar, acidic to basic) from aqueous samples
Method simplicity Save time, money, and frustration during method development
Greater capacity Allows for smaller bed weights = smaller elution volumes = time savings in sample processing
Resistant to overdrying More robust methodology and better reproducibility
Low UV and MS extractables Lower background for greater sensitivity
Stringent production & QC Greater lot-to-lot, tube-to-tube, and well-to-well consistency for improved accuracy and precision
High and Reproducible Recoveries
The hydrophilic, lipophilic balanced Supel-Select HLB SPE allows users to extract a broad range of compounds using a single sorbent and generic methodology. Analyte recovery was high across all the compounds tested, and results were highly reproducible across three production lots.
Minimal Extractables
Supel-Select HLB SPE phases are quality controlled for both LC-UV and LC-MS/MS extractables. As assays require greater sensitivity, analytical “cleanliness” of both SPE phases and associated hardware becomes more critical.
In this simple test, acetonitrile was passed through both Supel-Select HLB SPE tubes and the leading competitor’s tubes followed by LC-UV analysis.
Supel-Select HLB SPE imparted less than 25% of UV extractables than the leading competitor.