Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
NMR Spectroscopy Tubes
Aldrich NMR Tubes
Every Aldrich premium-quality NMR tube is inspected and gauged to meet published specifications.
  • Aldrich Statement of Conformance stamped on each box of tubes
  • Made of ASTM Type I borosilicate glass* for compatibility with ground joints, valves, and vacuum racks
  • Precision ground and highly polished for spinning stability and highest resolution
  • Thin-wall glass with round-bottoms
  • Sandblasted marking spot for labelling samples
    *Except disposable grade NMR tubes.
NMR Tube Selection Guide
Choose the NMR tube grade that most closely matches the frequency (in MHz) of the spectrometer.
Aldrich Grade MHz Applications Cross Ref. No.
(5mm tubes)
Wilmad Kontes
Disposable 30 Student labs, basic studies WG-Economy
Series 30 30 Student labs, basic studies 505-PS 897200
Series 200 60 to 270 Lower field studies 506-PP 897205
Series 300 60 to 360 Routine applications 507-PP 897220
Series 400 200 to 500 High-field, multiple-scan FT-NMR 528-PP 897235
Series 500 500+ Ultra-high resolution 535-PP 897240
Series 800 600 to 800 Highest resolution and performance 541-PP 897245
Grade L (in.) Wall uniformity
(in., TIR)
Camber (in.) Cat. No.
Add to Cart
3mm Microsample NMR tubes — 0.012in. wall thickness (Pkg=5 tubes)
Series 300 7 0.0020 0.0010 Z412902
  8 0.0020 0.0011 Z412910
Series 400 7 0.0010 0.0005 Z412929
  8 0.0010 0.0006 Z412937
Series 500 7 0.0005 0.00025 Z412945
  8 0.0005 0.00035 Z412953
5mm NMR tubes — 0.015in. wall thickness (Pkg=50 tubes)
Series 60 7 - - Z543888
  8 - - Z555908
5mm Thrift NMR tubes - 0.01475in. wall thickness (pkg=5 tubes)
Series 200 7 0.002 0.0005 Z555894
  8 0.002 0.0006 Z556092
Series 300 7 0.0015 0.001 Z556106
  8 0.0015 0.001 Z556114
Series 400 7 0.0005 0.00025 Z556122
  8 0.0005 0.00035 Z556130
Series 500 7 0.0005 0.00025 Z556149
  8 0.0005 0.00035 Z556157
5mm Precision NMR tubes - 0.01475in. wall thickness (pkg=5 tubes)
Series 80 7 0.003 0.002 Z412783
Series 200 7 0.0025 0.002 Z412791
Series 300 7 0.002 0.005 Z412805
  8 0.002 0.0006 Z412813
  9 0.002 0.0007 Z412821
Series 400 7 0.001 0.0005 Z412848
  8 0.001 0.006 Z412856
Series 500 7 0.0005 0.00025 Z412864
  8 0.0005 0.00035 Z412872
Series 800 7 0.0002 0.00015 Z412880
  8 0.0002 0.00015 Z412899
Series 900 7 0.0002 0.00015 Z550868
  8 0.0002 0.00015 Z548243
Wilmad NMR Tubes
Made of Pyrex borosilicate glass (unless otherwise noted) for compatibility with ground joints, valves, and vacuum racks. Precision ground and highly polished for spinning stability and highest resolution. Tubes have round bottoms and include a sandblasted marking spot for labeling samples. Tubes that are 9in. L are for making standards and for use with tip-off manifolds.
L(in.) Concentricity/camber (µm) Wilmad No. Cat. No. Add to Cart
3mm Microsample NMR tubes - 0.012in. wall thickness (Pkg=5 tubes)
300MHz tubes
7 51/25 307-PP-7 Z274321
8 51/25 307-PP-8 Z274348
9 51/25 307-PP-9 Z274356
360MHz tubes
7 25/25 327-PP-7 Z271861
8 25/25 327-PP-8 Z274364
9 25/25 327-PP-9 Z274372
400MHz tubes
7 25/13 328-PP-7 Z274380
8 25/13 328-PP-8 Z274399
9 25/13 328-PP-9 Z274402
500MHz tubes
7 13/6 335-PP-7 Z274410
8 13/6 335-PP-8 Z274429
9 13/6 335-PP-9 Z274437
5mm amber NMR tubes - 0.015in. wall thickness (Pkg=1 tube)
100MHz tube
7 76/51 505-PS-7 Z282251
360MHz tubes
7 51/25 507-PP-7 Z282278
7 25/25 527-PP-7 Z282286
5mm disposable NMR tubes - 0.015in. wall thickness, N51 glass (Pkg=100 tubes)
100MHz tubes
7 76/76 HIP-7 Z514144
8 76/76 HIP-8 Z514152
5mm economy NMR tubes - 0.015in. wall thickness. WG series made of N51 glass. (Pkg=5 tubes)
100MHz tubes
7 76/76 WG-5-7 Z276278
8 76/76 WG-5-8 Z276286
9 76/76 WG-5-9 Z276294
7 76/76 512-5-7 Z276308
8 76/76 512-5-8 Z276316
9 76/76 512-5-9 Z276324
7 51/51 506-IM-7 Z514128
8 51/51 506-IM-8 Z514136
5mm standard NMR tubes - 0.015in. wall thickness (Pkg=5 tubes)
100MHz tubes
7 76/51 505-PS-7 Z271985
8 76/51 505-PS-8 Z274445
9 76/51 505-PS-9 Z274453
7 51/51 506-PP-7 Z286028
8 51/51 506-PP-8 Z286036
9 51/51 506-PP-9 Z286044
200MHz tube
7 76/51 503-PS-7** Z410357
360MHz tubes
7 51/25 507-PP-7 Z271993
8 51/25 507-PP-8 Z274461
9 51/25 507-PP-9 Z272116
7 51/13 526-PP-7 Z274488
8 51/13 526-PP-8 Z274496
9 51/13 526-PP-9 Z274518
7 25/25 527-PP-7 Z274275
8 25/25 527-PP-8 Z274526
9 25/25 527-PP-9 Z274534
400MHz tubes
7 25/13 528-PP-7 Z272019
8 25/13 528-PP-8 Z274291
9 25/13 528-PP-9 Z274542
500 MHz tubes
7 13/6 535-PP-7 Z272027
7 13/6 545-PPT-7* Z406708
8 13/6 535-PP-8 Z274550
9 13/6 535-PP-9 Z274569
800 MHz tubes
7 3.8/3.8 541-PP-7 Z412007
8 3.8/3.8 541-PP-8 Z412015
900 MHz tubes
7 2.5/3.8 542-PP-7 Z412023
8 2.5/3.8 542-PP-8 Z412031
* Ultra thin wall (0.09in.)
** Medium wall (0.03in.)
10mm NMR tubes - 0.018in. wall thickness (Pkg=5 tubes)
60MHz tubes
7 NA 513-1PS-7 Z274771
8 NA 513-1PS-8 Z274798
9 NA 513-1PS-9 Z274801
100MHz tubes
7 254/51 Z27,468-2 Z272035
8 254/51 513-1PP-8 Z274690
9 254/51 513-1PP-9 Z274704
300MHz tubes
7 76/38 513-3PP-7 Z272035
8 76/38 513-3PP-8 Z274712
9 76/38 513-3PP-9 Z272043
360MHz tubes
7 51/25 513-5PP-7 Z274720
8 51/25 513-5PP-8 Z274739
9 51/25 513-5PP-9 Z274747
400MHz tubes
7 38/13 513-7PP-7 Z272051
8 38/13 513-7PP-8 Z274755
9 38/13 513-7PP-9 Z274763
*Tubes do not have a precision i.d. tolerance.  
Wilmad Screw Cap NMR Tubes
NMR tube has an open-top, screw-thread cap with a PTFE/silicone septum that permits addition of reactants or samples via syringe through the cap. Also good for sample storage. Caps and septa are replaceable. Excellent for handling air-sensitive samples and in cell perfusion studies. Tubes are 7in. L.
Product Name Concentricity/camber (µm) Wilmad No. Cat. No. Add to Cart
5mm NMR tubes  (Pkg=1 tube)
100MHz tube 76/51 505-TR-7 Z271918
100MHz tube 51/51 506-TR-7 Z271926
360MHz tube 51/25 507-TR-7 Z271934
400MHz tube 25/13 528-TR-7 Z271942
360MHz tube 51/25 507-TR-7 Z271934
400MHz tube 25/13 528-TR-7 Z271942
500MHz tube 13/60 535-TR-7 Z271950
8mm NMR tubes  (Pkg=1 tube)
400MHz tube 38/13 513A-7TR Z274828
10mm NMR tubes  (Pkg=1 tube)
300MHz tube 76/38 513-3TRA-7 Z271969
400MHz tube 38/13 513-7TRA-7 Z271977
Replacement caps and septa for screw-cap NMR tubes
NMR tube size (mm) Cat. No. Pkg Add to Cart
Black open-top caps
5 Z162531 200
10 Z106453 200
Black solid-top caps w/white rubber liner
5 Z188808-1PAK
Z188808-10PAK
100
10x100
10 Z106410 200
Black solid-top caps w/PTFE-faced rubber liner
10 Z106437 200
PTFE-faced silicone septa
5 Z162566 120
10 Z106518 100
Norell™ NMR Sample Tubes
MHz Length (in.) Concentricity +/-(mm) Camber +/- (mm) Cat. No. Add to Cart
3 mm Select Series™ - ASTM Type 1 Class A Glass
o.d. (mm) 2.99 (+/- 0.004)
i.d. (mm) 2.41 (+/- 0.006)
900 7 0.002 0.003 NORS39007-5EA
900 8 0.002 0.003 NORS39008-5EA
800 7 0.025 0.0038 NORS38007-5EA
800 8 0.025 0.0038 NORS38008-5EA
600 7 0.004 0.006 NORS36007-5EA
600 8 0.004 0.006 NORS36008-5EA
500 7 0.005 0.013 NORS35007-5EA
500 8 0.005 0.013 NORS35008-5EA
400 7 0.007 0.019 NORS34007-5EA
400 8 0.007 0.019 NORS34008-5EA
300 7 0.007 0.025 NORS33007-5EA
300 8 0.007 0.025 NORS33008-5EA
200 7 0.01 0.038 NORS32007-5EA
200 8 0.01 0.038 NORS32008-5EA
5 mm Select Series™ - ASTM Type 1 Class A Glass
o.d. (mm) 4.97 (+/- 0.004)
i.d. (mm) 4.20 (+/- 0.006)
900 7 0.002 0.003 NORS59007-5EA
900 8 0.002 0.003 NORS59008-5EA
800 7 0.0025 0.0038 NORS58007-5EA
800 8 0.0025 0.0038 NORS58008-5EA
600 7 0.004 0.006 NORS56007-5EA
600 8 0.004 0.006 NORS56008-5EA
500 7 0.005 0.013 NORS55007-5EA
500 8 0.005 0.013 NORS55008-5EA
400 7 0.007 0.019 NORS54007-5EA
400 8 0.007 0.019 NORS54008-5EA
300 7 0.007 0.025 NORS53007-5EA
300 8 0.007 0.025 NORS53008-5EA
200 7 0.009 0.035 NORS52007-5EA
200 8 0.009 0.035 NORS52008-5EA
5 mm Standard Series™ - ASTM Type 1 Class B, N51A Glass
o.d. (mm) 4.97 (+/- 0.013)
i.d. (mm) 4.20 (+/- 0.025)
600 7 0.004 0.006 NOR509UP7-5EA
600 8 0.004 0.006 NOR509UP8-5EA
500 7 0.005 0.013 NOR508UP7-5EA
500 8 0.005 0.013 NOR508UP8-5EA
400 7 0.007 0.019 NOR507HP7-5EA
400 8 0.007 0.019 NOR507HP8-5EA
300+ 7 0.007 0.025 NOR506P7-25EA
300+ 8 0.007 0.025 NOR506P8-25EA
300 7 0.01 0.038 NORXR557-25EA
300 8 0.01 0.038 NORXR558-25EA
200 7 0.01 0.04 NOR505P7-25EA
200 8 0.01 0.04 NOR505P8-25EA
100 7 0.02 0.07 NOR5027-50EA
100 8 0.02 0.07 NOR5028-50EA
60 7 nominal nominal NORST5007-100EA
60 8 nominal nominal NORST5008-100EA
10 mm Standard Series™ - ASTM Type 1 Class B, N51A Glass
o.d. (mm) 10.00 (+/- 0.013)
i.d. (mm) 8.76 (+/- 0.025)
400 7 0.005 0.007 NOR1008UP7-5EA
400 8 0.005 0.007 NOR1008UP8-5EA
300 7 0.02 0.013 NOR1005P7-5EA
300 8 0.02 0.013 NOR1005P8-5EA
200 7 nominal nominal NOR10017-100EA
200 8 nominal nominal NOR10018-100EA
Shigemi Advanced NMR microtube assembly
Outer Sample Tube O.D. (mm) Bottom L (mm) Shigemi No. Cat. No. Add to Cart
Matched with CDCl3        
5 8 CMS-005TB Z529087-1EA
5 12 CMS-005TJ Z529095-1EA
5 15 CMS-005TV Z529109-1EA
8 8 CMS-008TB Z529117-1EA
8 12 CMS-008TJ Z529125-1EA
8 15 CMS-008TV Z529133-1EA
Matched with CD3OD        
5 8 MMS-005TB Z529141-1EA
5 12 MMS-005TJ Z529168-1EA
5 15 MMS-005TV Z529176-1EA
8 8 MMS-008TB Z529184-1EA
8 12 MMS-008TJ Z529192-1EA
8 15 MMS-005TV Z529206-1EA
Matched with DMSO-d6        
5 8 DMS-005TB Z529214-1EA
5 12 DMS-005TJ Z529222-1EA
8 8 DMS-008TB Z529249-1EA
8 12 DMS-008TJ Z529257-1EA
8 15 DMS-008TV Z529265-1EA
5 15 DMS-005TV Z529230-1EA
Matched with D2O        
5 8 BMS-005TB Z529451-1EA
5 12 BMS-005TJ Z529273-1EA
5 15 BMS-005TV Z529281-1EA
8 8 BMS-008TB Z529303-1EA
8 12 BMS-008TJ Z529311-1EA
8 15 BMS-008TV Z529338-1EA
Matched with CDCl3
8 - CMS005B Z529346-1EA
12 - CMS005J Z529354-1EA
15 - CMS005V Z529362-1EA
- Microtube cap CMSCAP Z529370-1EA
Matched with DMSO-d6
8 - DMS005B Z529389-1EA
12 - DMS005J Z529397-1EA
15 - DMS005V Z529400-1EA
- Microtube cap DMSCAP Z529419-1EA
Matched with D2O
8 - BMS005B Z543349-1EA
12 - BMS005J Z529427-1EA
15 - BMS005V Z529435-1EA
- Microtube cap BMSCAP Z529443-1EA
Cross-Reference Guide
Norell™ Wilmad® Aldrich®   Norell™ Wilmad® Aldrich®
NORST5007 HIP-7     NORS36007 335-PP-7 Z412945
NORST5008 HIP-8     NORS36008 335-PP-8 Z412953
NORST5007 WG-5MM-ECON-7     NORXR557 505-PS-7 Z412783
NORST5008 WG-5MM-ECON-8     NORXR558 505-PS-8  
NOR5027 512-7 Z543888   NORS52007 506-PP-7 Z412791
NOR5028 512-8 Z555908   NORS52008 506-PP-8  
NOR5027 1206-7     NORS53007 507-PP-7 Z412805
NOR5028 1206-8     NORS53008 507-PP-8 Z412813
NOR505P7 1208-07 Z555894     507-PP-9 Z412821
NOR505P8 1208-08 Z556092   NOR10017 513-1PP-7 Z412961
NORXR557 1226-7 Z556106   NOR10018 513-1PP-8  
NORXR558 1226-8 Z556114   NOR1005P7 513-3PP-7  
NOR506P7 1228-7 Z556122   NOR1005P8 513-3PP-8  
NOR506P8 1228-8 Z556130   NOR1008UP7 513-7PP-7 Z412988
NOR507HP7 1235-7 Z556149   NOR1008UP8 513-7PP-8  
NOR507HP8 1235-8 Z556157   NORS54007 526-PP-7  
NOR508UP7 1241-7     NORS54008 526-PP-8  
NOR508UP8 1241-8     NORS55007 528-PP-7 Z412848
NOR509UP7 1242-7     NORS55008 528-PP-8 Z412856
NOR509UP8 1242-8     NORS56007 535-PP-7 Z412864
NORS33007 305-PS-7     NORS56008 535-PP-8 Z412872
NORS33008 305-PS-8     NORS58007 541-PP-7 Z412880
NORS34007 307-PP-7 Z412902   NORS58008 541-PP-8 Z412899
NORS34008 307-PP-8 Z412910   NORS59007 542-PP-7 Z550868
NORS35007 328-PP-7 Z412929   NORS59008 542-PP-8 Z548243
NORS35008 328-PP-8 Z412937