Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
High-purity Carboxylic Acids for Trace-level Determinations
By: Michael Jeitziner, AnalytiX Volume 9 Issue 3
Sigma-Aldrich® offers new carboxylic acids used for trace analysis techniques such as ion-chromatography, AAS and polarography
Michael Jeitziner, Market Segment Manager Analytical Reagents & Standards michael.jeitziner@sial.com
Introduction
Organic acids are weak acids that don’t dissociate completely in water. They are often used in trace analysis applications as buffer and complexing agents for improving the separation. Further on, ascorbic acid is a highly effective antioxidant and therefore used as a stabiliser in various analytical applications. The most important fields of application are in metallurgy, environmental analysis, food analysis, toxicology, and clinical analysis. Sigma-Aldrich has recently introduced three organic acids with extremely low levels of inorganic impurities.
Find more information about our high-purity reagents
back to top
Hydride Generation-Atomic Absorption Spectroscopy (HG-AAS)
Metalloids like antimony, arsenic and selenium are mainly analysed by AAS using the hydride generation technique, which is easily connectable to various detection systems. The detection limits using this technique are below 1.0 μg/L.
A mixture of potassium iodide and ascorbic acid is most often used for the prereduction of As(V) to As(III), whereas potassium is the reducing agent, and ascorbic acid the antioxidant for stabilising the potassium iodide solution.1
Several heavy metal ions, such as iron(III), chromium(VI) and copper(II), can suppress the arsenic or antimony hydride generation. These interferences can be eliminated by the addition of potassium iodide and ascorbic acid.2
back to top
Ion Chromatography (IC) applications
IC is a technique that is used to separate and quantify ppt-%-levels of common anions and cations in aqueous samples based on the charge properties of the ions.
Citric acid, oxalic acid and other carboxylic acids in dissociated form chelate metal ions. By using different organic acids, a competitive complex formation takes place.
Besides their complex forming properties, the carboxylic acids also act as buffer substances. Their strength depends on their pKA values (see Table 1).
Table 1. Acid dissociation constants at 25 °C in water
Table 1. Acid dissociation constants at 25 °C in water3
Depending on the separation problem, variation of the pH value, the use of a complexing agent and/or an increase in column temperature are powerful tools to broaden the scope of cation chromatography.
L-Ascorbic acid (vitamin C) is added to reduce Fe(III) to Fe(II)4-7 for the determination of iron using cation chromatography. Further on, it can also be used in polarography to eliminate the interfering effect of iron(III) ions8 or for the speciation analysis of Fe(III) and Fe(II) ions.
Citric acid and oxalic acid form strong complexes especially with di- and trivalent cations. Tables 2 and 3 illustrate the effect of citric and oxalic acid on different analytes.
Table 2. “Effect of eluent composition on IC column retention times”. The original work can be downloaded from http://products.metrohm.com
Table 2. “Effect of eluent composition on IC column retention times”. The original work can be downloaded from http://products.metrohm.com (search for 8.000.6015EN)
Table 3. Effect of different concentrations of citric acid and oxalic acid on the retention time of various cations using the Metrosep C 2 – 150 separation column
Table 3. Effect of different concentrations of citric acid and oxalic acid on the retention time of various cations using the Metrosep C 2 – 150 separation column
back to top
References
  1. M. Vilanó, R. Rubio, Journal of AOAC INTERNATIONAL, 84 (2001), 551 – 555.
  2. S. Terashima et al, Bunseki Kagaku, 33 (1984), 561 – 563.
  3. Shriver, D.F; Atkins, P.W. (1999). Inorganic Chemistry (3rd ed.). Oxford: Oxford University Press. ISBN 0198503318. Chapter 5: Acids and Bases.
  4. Metrohm – IC Application Note No. C-54 “Potassium, iron, magnesium and calcium in clay”.
  5. Metrohm – IC Application Note No. C-79 “Nickel, zinc, cobalt, iron(II) and manganese in lithium bromide using post-column reaction”.
  6. Metrohm – IC Application Note No. C-63 “Five cations in lithium bromide using post-column reaction”.
  7. Metrohm – IC Application Note No. C-62 “Five cations including iron in monoethylene glycol (MEG)”.
  8. Metrohm – Polarography Application Bulletin No. 242/1 “Tungsten at Ultra Trace Graphite Electrode by anodic stripping voltammetry”.