Pseudomonas Media and Tests
By: Jvo Siegrist, Product Manager Microbiology, ivo.siegrist@sial.com, Analytix Volume 2007 Article 5
Detection, identification, differentiation and cultivation of Pseudomonas species
Figure 1.HiFluoro Pseudomonas Agar under UV light
Pseudomonas are motile (one or more polar flagella), rod shaped and aerobe Gram-negative bacteria. They are found almost everywhere, in soil, water, plants and animals. In most cases it is not pathogenic and in fact can be beneficial. For example, P. putida is used as a bio-scrubber to aid in the biodegradation of diverse organic compounds in polluted air and waste water. However, P. aeruginosa is an infamous opportunistic human pathogen most commonly affecting immuno-compromised patients. Along with P. maltophilia, it accounts for the majority of human infections. Pathogenic Pseudomonas are found throughout the body, most commonly in the urinary tract, respiratory tract, blood and wounds [1].
Rugged and opportunistic, Pseudomonas use a wide range of nutritional sources, even very simple nutritional environments without any organic compounds. They can remain viable for long periods of time in many different habitats and under very adverse conditions. They are also widespread, being found in water, saline solutions, utensils and even in cosmetics, pharmaceuticals and disinfectants, and many natural and manufactured foods. Psychrotrophic (cold-tolerant) Pseudomonas species are a significant food spoilage problem in refrigerated meat, fish, shell fish and dairy products. Because Pseudomonas thrive in water systems, they can be the source of contamination in the food and beverage industry [2].
Pseudomonas are not generally fastidious microorganisms. They can grow on very simple media like Kind Agar, for example, which contains a protein hydrolysate, magnesium chloride, potassium sulphate and agar. Analytical microbiology leverages a microbe’s unique biochemistry to aid in its identification. For example, selective Pseudomonas media use cetrimide, nalidixic acid, cephaloridine, penicillin G, pimaricin, malachite green and other inhibitory agents. The proteolytic activity, lipolytic activity, fluorescent pigment formation, nitrate utilisation, glutamate utilisation, hemolytic reaction and other biochemical reactions are used in the media for the identification and differentiation of Pseudomonas species.
Pseudomonas gives negative Voges Proskauer, indole and methyl red tests, but a positive catalase test. While some species show a negative reaction in the oxidase test, most species, including P. fluorescens, give a positive result (see Figure 2). Another feature associated with Pseudomonas is the secretion of pyoverdin (fluorescein, a siderophore), a fluorescent yellow-green pigment under iron-limiting conditions [3]. Certain Pseudomonas species may also produce additional pigments, such as pyocyanin (blue pigment, a siderophore) by P. aeruginosa [4], quinolobactin (yellow, dark green in presence of iron, a siderophore) by P. fluorescens [5], a reddish pigment called pyorubrin and pyomelanin (brown pigment). On blood agar a hemolytic reaction can be observed.
Figure 2.Oxidase test
Scientific classification of Pseudomonas:
| Kingdom: Bacteria | Order: Pseudomonadales |
| Phylum: Proteobacteria | Family: Pseudomonadaceae |
| Class: Gamma Proteobacteria | Genus: Pseudomonas |
Pseudomonas utilizes sugars as an energy source by using the Entner-Doudoroff pathway with pyruvate as the end product (dissimilation). The reaction utilizes a different set of enzymes from those used in glycolysis and the pentose phosphate pathway. Fermentation catabolism is not observed in Pseudomonas, but some species, like P. aeruginosa, P. stutzeri and P. denitrificans, are able to use nitrate as an electron acceptor instead of oxygen. Growth can also occur under anaerobic conditions when the denitrification pathway is used. Sigma-Aldrich supplies a wide array of products for the detection, identification, differentiation, enumeration and cultivation of Pseudomonas, using its biochemical characteristics, including Gram staining kit, and many types of selective growth media (Table 1) and diagnostic tests (Table 2). Additional information on media and tests for Pseudomonas and a wide range of other microbes can be found on our web site: www.sigmaaldrich.com/microbiology.
Table 1 Media for Pseudomonas
| Cat. No. | Brand | Nonselective Broths |
| A0465 | Sigma | Alternative Thioglycollate Medium |
| 53286 | Fluka | Brain Heart Broth |
| B5051 | Sigma | Bushnell Haas Broth |
| D3435 | Sigma | Dey-Engley Neutralizing Broth |
| 63649 | Fluka | Membrane filter Rinse Fluid (USP) |
| 70149 | Fluka | Nutrient Broth No 3 |
| 03856 | Fluka | Nutrient Broth No. 4 |
| 70179 | Fluka | Peptone Water |
| 77187 | Fluka | Peptone Water, phosphate-buffered |
| 40893 | Fluka | Peptone Water, phosphate-buffered,Vegitone |
| 70157 | Fluka | Thioglycollate Broth (USP Alternative) |
| 41960 | Fluka | Vegitone Infusion Broth |
| Cat. No. | Brand | Selective Enrichment Broths & Biochemical Identification Broths |
| 00185 | Fluka | Acetamide Nutrient Broth |
| 17129 | Fluka | Asparagine Proline Broth |
| 78886 | Fluka | Cetrimide Broth |
| 63163 | Fluka | Malachite Green Broth |
| 39484 | Fluka | Methyl Red Voges Proskauer Broth |
| 14305 | Fluka | Motility Nitrate Medium |
| 72548 | Fluka | Nitrate Broth |
| Cat. No. | Brand | Nonselective Agars for Cultivation, Enumeration and Isolation |
| 70147 | Fluka | Milk Agar |
| 70148 | Fluka | Nutrient Agar |
| 44776 | Fluka | Nutrient Agar Plates (Diameter 55 mm) |
| 80957 | Fluka | Plate Count Skim Milk Agar |
| 17209 | Fluka | R-2A Agar |
| 17175 | Fluka | Skim Milk Agar, modified |
| 51414 | Fluka | Skim Milk Agar, modified |
| 70159 | Fluka | Tryptone Glucose Extract Agar |
| T2188 | Sigma | Tryptone Glucose Yeast Extract Agar |
| 01497 | Fluka | Yeast Extract Agar |
| Cat. No. | Brand | Nonselective Agars for Differentiation |
| 70133 | Fluka | Blood Agar (Base) |
| 21065 | Fluka | Calcium caseinate Agar |
| 55420 | Fluka | CLED Agar |
| 16636 | Fluka | HiCrome(™) UTI Agar, modified |
| 60788 | Fluka | King Agar A |
| 60786 | Fluka | King Agar B |
| 75315 | Fluka | OF Test Nutrient Agar |
| P1852 | Sigma | Pseudomonas Agar (for Fluorescein) |
| 91015 | Fluka | Tributyrin Agar |
| Cat. No. | Brand | Selective Agars for Detection and Isolation |
| 11012 | Fluka | Cetrimide Nalidixic acid Agar |
| 22470 | Fluka | Cetrimide Agar |
| 70887 | Fluka | Cetrimide Agar |
| P2102 | Sigma | Pseudomonas Agar Base |
| 14521 | Fluka | Cetrimide Agar Plates (Diameter 55 mm) |
| 17168 | Fluka | Milk Agar, modified according to Brown & Scott |
| 17208 | Fluka | Pseudomonas Isolation Agar |
| Cat. No. | Brand | Selective Agars with Differential System for Differentiation, Detection and Isolation |
| 50875 | Fluka | GSP Agar |
| 78996 | Fluka | HiFluoro (™) Pseudomonas Agar Base |
Table 2 Test for identification and differentiation of Pseudomonas
| Cat. No. | Brand | Diagnostic Tests for Pseudomonas |
| 88597 | Fluka | Catalase Test |
| 05686 | Fluka | DMACA Indole Disks |
| 49825 | Fluka | DMACA Reagent |
| 78719 | Fluka | Kovac‘s Reagent Strips |
| 60983 | Fluka | Kovac‘s Reagent for indoles |
| 67309 | Fluka | Kovac‘s Reagent for indoles |
| 08714 | Fluka | Methyl Red Solution |
| 70439 | Fluka | Oxidase Test |
| 40560 | Fluka | Oxidase Strips |
| 07345 | Fluka | Oxidase Reagent acc. Gaby-Hadley A |
| 07817 | Fluka | Oxidase Reagent acc. Gaby-Hadley B |
| 18502 | Fluka | Oxidase Reagent acc. Gordon-McLeod |
References
- Collins, F.M. Pasteurella, Yersinia, and Francisella. In Barron‘s Medical Microbiology, 4th ed., S. Barron, et al., eds., University of Texas Medical Branch at Galveston: Galveston, TX, 1996; Chapter 27.
- Mead, G.C. et al. Br. Poult. Sci. 1977, 18, 661-670.
- Meyer, J.M.; Geoffroy, V.A.; Baida, Appl. Environ. Microbiol. 2002, 68 (6), 2745-2753.
- Lau, G.W.; Hassett, D.J.; Ran, H.; Kong, F. Trends in molecular medicine 2004, 10 (12), 599-606.
- Matthijs, S.; Tehrani, K.A.; Laus, G.; Jackson, R.W.; Cooper, R.M.; Cornelis, P. Environ. Microbiol. 2007, 9 (2), 425-