Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Nancy-520 for DNA Detection and Quantitation
By: Monika Baeumle, BioFiles 2009, 4.1, 4.
Nancy-520 is a fluorescent stain for double stranded DNA (dsDNA) on agarose electrophoresis gels, with higher sensitivity than ethidium bromide and an easy, fast, and robust staining procedure. It is less mutagenic than Ethidium bromide and SYBR® Green I, according to Ames test. Nancy-520 has an excitation maximum at 520 nm and an emission maximum at 560 nm (see Figure 1). Nancy-520 is provided as a 5000× stock solution (500 μl), which is sufficient for 50 agarose gels. The limit of detection is 0.5 ng/band of dsDNA. In addition, Nancy-520 can be used to determine dsDNA concentrations in solution.
Figure 1.Normalized fluorescence excitation (red) and emission (blue) spectra of Nancy-520 in the presence of dsDNA, measured in a cuvette on a Varian Cary® Eclipse fluorescence spectrophotometer with 2 mL of TBE, pH 8.3. The excitation spectrum was measured at a fixed emission wavelength of 560 nm, and the emission spectrum was measured at a fixed excitation wavelength of 520 nm.
back to top
DNA Staining
Nancy-520 can be used in a standard post-electrophoresis staining application for DNA on agarose gels. After the electrophoresis run, the gel is stained for 1 hour in the dark. The fluorescence image of the gel can be directly detected after a short rinse (see Figure 2). Alternatively, Nancy-520 can also be used for pre-electrophoresis staining by simply adding the dye to the liquid agarose before pouring the gel. The gel can be imaged directly after the electrophoresis run without any further steps. Nancy-520 allows more detection possibilities than SYBR® Green. Detection is performed by illuminating the gel on a UV screen or using a Dark Reader®, and imaging the gel using a CCD-camera (e.g., KODAK® Gel-Logic-100) with a 535 nm or 590 nm band-pass filter, or a Polaroid® camera. Alternatively, a laser-scanner can be used (e.g., Fuji® FLA-3000) with a 473 nm excitation setting and 520 nm emission filter, or with a 532 nm excitation setting and 580 nm emission-filter. Other imaging systems are possible with similar excitation sources and emission filter settings.
Figure 2.DNA marker (Lambda DNA Hind III digest) at 2 different concentrations, was separated on a 1% agarose gel, stained with Nancy-520 post-electrophoresis and imaged under 2 different conditions. Left: λex UV-Screen (300 nm)/λem 590 nm bandpass filter/CCD camera. Right: Laser-scanner Fuji FLA-3000/λex 532 nm/λem 580 nm cut-off filter.
back to top
DNA Quantitation in Solution
Nancy-520 can be used to determine dsDNA concentrations in solution. This application can be performed in a glass-bottomed 96 well plate, using known concentrations of dsDNA as a standard. Nancy-520 has a linear detection range between 0-2 μg/ml of DNA (see Figure 3).
Figure 3.Quantitation of DNA (Lambda DNA Pst I Digest) in solution using Nancy-520,performed in a 96 well plate. Fluorescence was measured on a laser-scanner (FLA-3000,Fuji) with 532 nm excitation and 580 nm emission filter. Different concentrations of DNA in TE buffer, pH 7.5, and their corresponding fluorescence values are shown.