Fermentation: Enzymes, Substrates and SCFA
By: Chloe McClanahan, Biofiles Edition 6.2
The influence of probiotics on the metabolic activity of intestinal microbiota may be evaulated by detecting bacterial enzymatic activity in fecal samples, specifically β-glucosidase, β-glucuronidase, and urease activity. Experimental studies using probiotics have indicated a significant decrease in β-glucuronidase activity. A decrease in β-glucuronidase decreases the release of carcinogens from carcinogen-glucuronide conjugates within the colonic lumen.
back to top
Enzymes
Name Specific Activity Unit Definition Form Cat. No.
β-Glucosidase
from almonds
β-Glucosidase
from almonds
≥2 units/mg solid
10–30 units/mg solid
One unit will liberate 1.0 μmole of glucose from salicin per min at pH 5.0 at 37 °C. lyophilized powder
lyophilized powder
G0395
G4511
β-Glucuronidase
from Escherichia
coli
≥20,000 units/mg protein
1,000,000–5,000,000 units/g protein
5,000,000–20,000,000 units/g protein
20,000,000–60,000,000 units/g protein
20,000,000–60,000,000
units/g protein
≥10,000,000 units/g protein
One Sigma or modified Fishman unit will liberate 1.0 μg of phenolphthalein from phenolphthalein glucuronide per hr at 37 °C at pH 6.8 (30 min assay). lyophilized powder
lyophilized powder
lyophilized powder
aqueous glycerol solution
lyophilized powder
lyophilized powder
G8420
G7396
G7646
G8162
G7896
G8295
Urease from Canavalia ensiformis (Jack bean) 15,000–50,000 units/g solid
50,000–100,000 units/g solid
≥600,000 units/g solid
One micromolar unit will liberate 1.0 μmole of NH 3 from urea per min atpH 7.0 at 25 °C. It is equivalent to 1.0 I.U. or 0.054 Sumner unit (1.0 mg ammonia nitrogen in 5 minutes at pH 7.0 at 20 °C) powder
powder
powder
glycerol solution
U1500
U4002
U0251
U1875
Detection Substrates
Glucoside
Name Assay Cat. No.
3-Acetylumbelliferyl β-D-glucopyranoside ≥95% A3582
5-Bromo-4-chloro-3-indolyl β-D-glucopyranoside ≥97% B4527
6-Bromo-2-naphthyl β-D-glucopyranoside ≥99%, TLC B7877
6-Chloro-3-indolyl β-D-glucopyranoside ≥99.0%, HPLC 93546
Fluorescein di-(β-D-glucopyranoside) ≥90% F4521
Indoxyl β-D-glucoside ≥97% I3750
Indoxyl β-D-glucoside ≥97% I6893
2-Methoxy-4-(2-nitrovinyl)phenyl β-D-glucopyranoside ≥95% M2918
4-Methylumbelliferyl α- D -glucopyranoside M9766
4-Methylumbelliferyl β-D-glucopyranoside M3633
2-Nitrophenyl β-D-glucopyranoside N8016
4-Nitrophenyl α-D-glucopyranoside ≥99% N1377
4-Nitrophenyl β-D-glucopyranoside ≥98%, TLC N7006
Resorufin β-D-glucopyranoside ≥90% R4758
Glucuronide
Name Assay Cat. No.
5-Bromo-4-chloro-3-indolyl β-D-glucuronide cyclohexylammonium salt ≥98% B3783
5-Bromo-4-chloro-3-indolyl β-D-glucuronide cyclohexylammonium salt ≥98%, TLC B6650
5-Bromo-4-chloro-3-indolyl β-D-glucuronide cyclohexylammonium salt 10 mg substrate per tablet
B8049
5-Bromo-4-chloro-3-indolyl β-D-glucuronide cyclohexylammonium salt ≥98% B0522
5-Bromo-4-chloro-3-indolyl β-D-glucuronide sodium salt ≥98% B5285
5-Bromo-4-chloro-3-indolyl β-D-glucuronide sodium salt 10 mg substrate per tablet B8174
5-Bromo-6-chloro-3-indolyl β-D-glucuronide cyclohexylammonium salt ≥98% B4532
6-Chloro-3-indolyl-β-D-glucuronide cyclohexylammonium salt ≥97.0%, HPLC 24907
Indoxyl β-D-glucuronide cyclohexylammonium salt I7638
4-Methylumbelliferyl β-D-glucuronide hydrate ≥98%, TLC M9130
Naphthol AS-BI β-D-glucuronide N1875
4-Nitrophenyl β-D-glucuronide ≥98%, TLC N1627
Phenolphthalein β-D-glucuronide P0501
Phenolphthalein β-D-glucuronide sodium salt P0376
4-Trifl uoromethylumbelliferyl glucuronide potassium salt ≥98%, TLC T6410
Short-Chain Fatty Acids (SCFA)
Probiotics and prebiotics enhance SCFA end product synthesis. An increase in SCFA production lowers the gastrointestinal pH thus improving pathogen resistance and stimulating epithelial cell proliferation. The predominant SCFA, butyrate, is the preferred substrate for colonocytes which likely contributes to a normal colonocyte population and a lower cancer risk. SCFA production can be measured with gas chromatography analysis on fecal samples or intestinal modeling systems. However, due to normal colon absorption of 95% of the SCFA metabolites, stable isotope technology is a superior method of analysis.
Non-Labeled Short-Chain Fatty Acids
Name Cat. No.
Acetic acid, ReagentPlus ®, ≥99% A6283
Butyric acid, ≥99% B103500
Hexanoic acid, 99% H12137
Isobutyric acid, 99% I1754
Isovaleric acid, 99% 129542
Propionic acid, reagent grade, ≥99% P1386
Valeric acid, ≥99% 240370
Isotec® Short-Chain Fatty Acid Isotopes
Name Cat. No.
Sodium acetate-1- 13 C, S & P tested, 99 atom % 13 C 668656
Sodium acetate-1- 13 C, 99 atom % 13 C 279293
Sodium butyrate-1- 13 C, 99 atom % 13 C 292656
Sodium propionate-1- 13 C, 99 atom % 13 C 279455