Laminin
By: George Sitterley, BioFiles 2008, 3.8, 11.
BioFiles 2008, 3.8, 11.
Laminin has active domains for collagen binding, cell adhesion, heparin binding, and neurite outgrowth (PHD) fragment.1 Laminin chains are designated A (molecular weight 400kDa), B1 (molecular weight 210 kDa) and B2 (molecular weight 200 kDa). The cohesion between these chains is the result of many inter- and intrachain disulfide bonds. Together, they cause the molecule to look like a crucifix.
Laminin is a ubiquitous non-collagenous connective tissue glycoprotein that is a major constituent of basement membranes.
Laminins
Name Source Storage Temp Target Cells For Attachment Concentration For Use Cat. No.
Laminin, aqueous solution from Engelbreth- Holm-Swarm murine sarcoma basement membrane
−20°C
Epithelial cells, endothelial cells, muscle cells, tumor cells, hepatocytes, Schwannoma
1 ‑ 2 μg/cm2
L2020-1MG
Laminin, liquid from human placenta
−70°C
Epithelial cells, endothelial cells, muscle cells, tumor cells, hepatocytes, Schwannoma
1 ‑ 2 μg/cm2
L6274-.5MG
back to top Back to Top
Materials
References
  1. Kleinman, H. K., et al., Use of extracellular matrixcomponents for cell culture. Anal. Biochem., 166, 1-13 (1987).
back to top Back to Top
Related Links
Categories