Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
1 kb DNA Ladder
Catalog No. D0428
Product Description
This DNA 1 kb Ladder contains 11 fragments consisting of 500 bp repeats from 0.5 to 3 kb, 1 kb repeats from 3 to 6 kb, and 2 kb repeats from 6 to 10 kb.
DNA Ladder Fragment Sizes: kilobase pairs (kb)
10.0
8.0
6.0
5.0
4.0
3.0
2.5
2.0
1.5
1.0
0.5
The product is supplied in a solution of 10 mM Tris-HCl, pH 8.0, with 1.0 mM EDTA.
The DNA 1 kb Ladder is suitable for size determination of double-stranded DNA using DNA electrophoresis. This marker can be used with either agarose or polyacrylamide gels. The recommended agarose gel concentration is 0.75% for this marker.
Recommended load volume: 5 ml
Number of loads per vial: 100
Adjustments may be made for different well sizes and individual preferences.
Precautions and Disclaimer
This product is for R&D use only, not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.
Preparation Instructions
A sample of the marker should be diluted with gel loading buffer to an appropriate loading concentration.
Storage/Stability
Store the product at –20 °C.
Materials
KAA,KH,MAM 09/11-1
Related Links