Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Amicon® Ultra Centrifugal Filters
Concentrate protein, don’t lose it. Recover protein reliably when you concentrate with Amicon® Ultra filters. We’ve engineered Amicon® Ultra filters with low-binding Ultracel® regenerated cellulose membrane. Unlike membranes in centrifugal filters from other suppliers, Ultracel® membrane’s design and manufacturing translate to more precise molecular weight cut-offs, making Amicon® Ultra filters the best for protein concentration.
How to choose the correct Amicon® Ultra filter for concentrating your protein, DNA or other macromolecule of interest
  • Determine the starting volume of your sample
  • Determine the molecular weight (MW) of your macromolecule of interest, and choose an ultrafiltration membrane with a molecular weight cut-off (MWCO) that is 2-3 times smaller than the MW.
  • Select pack size
Order using the table below.
Video
Amicon® Ultra filter vs. competitor centrifugal concentrator. Though the competitor device may appear to filter faster, protein is being lost through the membrane.
Ordering Information
Product No. Description Maximum Initial Sample Volume (mL) Final Concentrate (Retentate) Volume (μL) MWCO Qty/Pk Cart No.
UFC5003 Amicon® Ultra-0.5 Centrifugal Filters 0.5 15–20 3000 8
UFC5003 Amicon® Ultra-0.5 Centrifugal Filters 0.5 15–20 3000 24
UFC5003 Amicon® Ultra-0.5 Centrifugal Filters 0.5 15–20 3000 96
UFC5003 Amicon® Ultra-0.5 Centrifugal Filters 0.5 15–20 3000 500
UFC5010 Amicon® Ultra-0.5 Centrifugal Filters 0.5 15–20 10000 8
UFC5010 Amicon® Ultra-0.5 Centrifugal Filters 0.5 15–20 10000 24
UFC5010 Amicon® Ultra-0.5 Centrifugal Filters 0.5 15–20 10000 96
UFC5010 Amicon® Ultra-0.5 Centrifugal Filters 0.5 15–20 10000 500
UFC5030 Amicon® Ultra-0.5 Centrifugal Filters 0.5 15–20 30000 8
UFC5030 Amicon® Ultra-0.5 Centrifugal Filters 0.5 15–20 30000 24
UFC5030 Amicon® Ultra-0.5 Centrifugal Filters 0.5 15–20 30000 96
UFC5030 Amicon® Ultra-0.5 Centrifugal Filters 0.5 15–20 30000 500
UFC5050 Amicon® Ultra-0.5 Centrifugal Filters 0.5 15–20 50000 8
UFC5050 Amicon® Ultra-0.5 Centrifugal Filters 0.5 15–20 50000 24
UFC5050 Amicon® Ultra-0.5 Centrifugal Filters 0.5 15–20 50000 96
UFC5050 Amicon® Ultra-0.5 Centrifugal Filters 0.5 15–20 50000 500
UFC5100 Amicon® Ultra-0.5 Centrifugal Filters 0.5 15–20 100000 8
UFC5100 Amicon® Ultra-0.5 Centrifugal Filters 0.5 15–20 100000 24
UFC5100 Amicon® Ultra-0.5 Centrifugal Filters 0.5 15–20 100000 96
UFC5100 Amicon® Ultra-0.5 Centrifugal Filters 0.5 15–20 100000 500
UFC200324 Amicon® Ultra-2 Centrifugal Filters 2 15–20 3000 24
UFC201024 Amicon® Ultra-2 Centrifugal Filters 2 15–20 10000 24
UFC203024 Amicon® Ultra-2 Centrifugal Filters 2 15–20 30000 24
UFC205024 Amicon® Ultra-2 Centrifugal Filters 2 15–20 50000 24
UFC210024 Amicon® Ultra-2 Centrifugal Filters 2 15–20 100000 24
UFC8003 Amicon® Ultra-4 Centrifugal Filters 4 30–70 3000 8
UFC8003 Amicon® Ultra-4 Centrifugal Filters 4 30–70 3000 24
UFC8003 Amicon® Ultra-4 Centrifugal Filters 4 30–70 3000 96
UFC8010 Amicon® Ultra-4 Centrifugal Filters* 4 30–70 10000 8
UFC8010 Amicon® Ultra-4 Centrifugal Filters* 4 30–70 10000 24
UFC8010 Amicon® Ultra-4 Centrifugal Filters* 4 30–70 10000 96
UFC8030 Amicon® Ultra-4 Centrifugal Filters 4 30–70 30000 8
UFC8030 Amicon® Ultra-4 Centrifugal Filters 4 30–70 30000 24
UFC8030 Amicon® Ultra-4 Centrifugal Filters 4 30–70 30000 96
UFC8050 Amicon® Ultra-4 Centrifugal Filters 4 30–70 50000 8
UFC8050 Amicon® Ultra-4 Centrifugal Filters 4 30–70 50000 24
UFC8050 Amicon® Ultra-4 Centrifugal Filters 4 30–70 50000 96
UFC8100 Amicon® Ultra-4 Centrifugal Filters 4 30–70 100000 8
UFC8100 Amicon® Ultra-4 Centrifugal Filters 4 30–70 100000 24
UFC8100 Amicon® Ultra-4 Centrifugal Filters 4 30–70 100000 96
UFC9003 Amicon® Ultra-4 Centrifugal Filters 15 150–300 3000 8
UFC9003 Amicon® Ultra-4 Centrifugal Filters 15 150–300 3000 24
UFC9003 Amicon® Ultra-4 Centrifugal Filters 15 150–300 3000 96
UFC9010 Amicon® Ultra-4 Centrifugal Filters* 15 150–300 10000 8
UFC9010 Amicon® Ultra-4 Centrifugal Filters* 15 150–300 10000 24
UFC9010 Amicon® Ultra-4 Centrifugal Filters* 15 150–300 10000 96
UFC9030 Amicon® Ultra-15 Centrifugal Filters 15 150–300 30000 8
UFC9030 Amicon® Ultra-15 Centrifugal Filters 15 150–300 30000 24
UFC9030 Amicon® Ultra-15 Centrifugal Filters 15 150–300 30000 96
UFC9050 Amicon® Ultra-15 Centrifugal Filters 15 150–300 50000 8
UFC9050 Amicon® Ultra-15 Centrifugal Filters 15 150–300 50000 24
UFC9050 Amicon® Ultra-15 Centrifugal Filters 15 150–300 50000 96
UFC9100 Amicon® Ultra-15 Centrifugal Filters 15 150–300 100000 8
UFC9100 Amicon® Ultra-15 Centrifugal Filters 15 150–300 100000 24
UFC9100 Amicon® Ultra-15 Centrifugal Filters 15 150–300 100000 96
*Certified for clinical applications.