Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Live Cell Imaging and Microscopy with the CellASIC® ONIX2 System
Precision control of your cell culture environment. The CellASIC® ONIX2 microfluidic system incorporates extraordinary improvements to traditional dynamic live cell analysis and experimentation, using microfluidics technology and computer automation.
  • Manipulate the culture environment (gas, temperature, media components, flow) under controlled conditions
  • Monitor the cells as they react in real time
  • System works with your existing microscope and capture/analysis software
Components of the CellASIC® ONIX2 Microfluidic System
Controller System: Small footprint, integrated microincubator controller maintains fluid movement, reagent additions, temperature, and gas conditions.
Microfluidic Plates: Application-specific plates (bacteria, yeast, mammalian cells, and more) for automated, perfusion culture, designed to optimize cell health during dynamic live cell imaging
Manifold: Low-profile, high quality, high optical clarity culture chambers and manifold, mount easily on an inverted microscope—without the need for a bulky environmental chamber.
Software: Intuitive software enables quick and easy set-up of detailed protocols for truly automated hands-free cell culture.
Key Advantages of CellASIC® ONIX2 Microfluidic Plates
  • Rapid perturbations of environmental parameters due to the tiny microfluidic volumes involved
  • Perfusion microbarriers allow continuous reagent transport without shearing stress
  • High resolution imaging through optical glass bottom and chamber structures that keep cells in a single focal plane
  • Ability to run multiple independent experiments simultaneously
  • Continuous access of nutrients and ongoing removal of waste during live cell imaging, promoting optimal cell health
Product No. CellASIC® ONIX2 Microfluidic System and Manifolds Qty/Pk
Request Information
CellASIC® ONIX2 Microfluidic System
  • ONIX2 Filter Multiconnector
  • ONIX2 Software USB Drive
  • ONIX2 Replacement Filter Pack
  • ONIX2 Accessory Fittings
  • ONIX2 Self Check Plate
  • ONIX2 Cleaning Plate
1
Request Information CellASIC® ONIX2 Manifold XT (Temperature Controlled) 1
Request Information CellASIC® ONIX2 Manifold Basic (No Temperature Control) 1
Product No. CellASIC® Microfluidic Plates
Qty/Pk Add to Cart
Y04C-02-5PK CellASIC® ONIX plate for haploid yeast cells 5
Y04D-02-5PK CellASIC® ONIX plate for diploid yeast cells 5
B04A-03-5PK CellASIC® ONIX plate for bacteria cells 5
M04S-03-5PK CellASIC® ONIX switching plate for mammalian cells 5
M04G-02-5PK CellASIC ONIX gradient plate for mammalian cells (4 chamber) 5
M04L-03-5PK CellASIC ONIX open-top plate for mammalian cells (4 chamber)   5