Centricon® Plus-70 Devices
Fast concentration of 15 mL – 70 mL samples
Centricon® Plus-70 DevicesCentricon® Plus-70 devices are capable of quickly concentrating samples 50X-200X, depending on the type and starting volume of the sample. Because of their low hold-up volume and low-binding membrane/housing, recovery typically exceeds 90%.
Centricon® Plus-70 Centrifugal Filter Units
Catalog Number NMWL Chemistry Pack Size Add to Cart
UFC700308 3 KDa Regenerated Cellulose (RC) 8
UFC701008 10 KDa Regenerated Cellulose (RC) 8
UFC703008 30 KDa Regenerated Cellulose (RC) 8
UFC710008 100 KDa Regenerated Cellulose (RC) 8