Durapore® PVDF Membrane Filters
Durapore® PVDF Membrane FiltersThe best-known properties of Durapore® Polyvinylidene Fluoride (PVDF) Membranes are their extremely low protein binding and minimal interaction with sample components. Binding less protein per unit surface area compared to nylon, PTFE, or nitrocellulose membranes, Durapore® filters are commonly used to clarify and sterilize protein-containing solutions.
Durapore® Membrane Filters are available in multiple pore sizes. Both hydrophobic and hydrophilic filters are available to match application requirements.
Key applications of Durapore® filters:
  • Sterilizing filtration
  • Filtration of protein-containing solutions
  • Mycoplasma reduction
  • Filtration of gases
  • Venting
Durapore® Membrane Disc Specifications
Membrane Material Polyvinylidene Fluoride (PVDF)
Pore Sizes (μm) 0.1 - 5
Thickness (μm) 80-140
Water Flow (mL/min/cm2) @27.5 in. Hg (hydrophilic) 4 - 208
Air Flow (L/min/cm2) @ 10 psi (hydrophobic) 0.9 - 4.9 (typical values)
Wettability Hydrophilic and hydrophobic options
Temperature Limitations 85 °C max
Refractive Index 1.42
Protein Binding Capacity (IgG)  < 10 μg/cm2
Chemical Compatibility Recommended for aqueous solvents
Sterilizability Autoclave, EtO, gamma irradiation
Surface Option White
Key Properties Low protein-binding (hydrophilic), solvent-resistant (hydrophobic)
Key Applications Sterilizing filtration, clarifying filtration of protein-containing solutions, mycoplasma reduction, gas filtration and venting (hydrophobic), solvent filtration (hydrophobic)
Also used in Millex® syringe filters and Sterile Millex® syringe filters
Hydrophilic Durapore® Membrane Filters
Product No. Pore Size Filter Diameter (⌀) Dimensions Pack Size Add to Cart
DVPP01300 0.65 µm 13 mm   100
DVPP02500 0.65 µm 25 mm   100
DVPP04700 0.65 µm 47 mm   100
DVPP09050 0.65 µm 90 mm   50
DVPP14250 0.65 µm 142 mm   50
GVWP00010 0.22 µm Not Applicable 1 ft. x 10 ft. roll 1
GVWP01300 0.22 µm 13 mm   100
GVWP02500 0.22 µm 25 mm   100
GVWP04700 0.22 µm 47 mm   100
GVWP09050 0.22 µm 90 mm   50
GVWP14250 0.22 µm 142 mm   50
HVLP00010 0.45 µm Not Applicable   1
HVLP01300 0.45 µm 13 mm   100
HVLP02500 0.45 µm 25 mm   100
HVLP04700 0.45 µm 47 mm   100
HVLP09050 0.45 µm 90 mm   50
HVLP14250 0.45 µm 142 mm   50
SVLP01300 5.0 µm 13 mm   100
SVLP02500 5.0 µm 25 mm   100
SVLP04700 5.0 µm 47 mm   100
SVLP09050 5.0 µm 90 mm   50
SVWG04700 5.0 µm 47 mm   100
VVLP01300 0.1 µm 13 mm   100
VVLP02500 0.1 µm 25 mm   100
VVLP04700 0.1 µm 47 mm   100
VVLP09050 0.1 µm 90 mm   50
VVLP14250 0.1 µm 142 mm   50
Hydrophobic Durapore® Membrane Filters
Product No. Pore Size Filter Diameter (⌀) Pack Size Add to Cart
GVHP00010 0.22 µm Not Applicable 1
GVHP01300 0.22 µm 13 mm 100
GVHP02500 0.22 µm 25 mm 100
GVHP04700 0.22 µm 47 mm 100
GVHP09050 0.22 µm 90 mm 50
GVHP14250 0.22 µm 142 mm 50
HVHP01300 0.45 µm 13 mm 100
HVHP02500 0.45 µm 25 mm 100
HVHP04700 0.45 µm 47 mm 100
HVHP09050 0.45 µm 90 mm 50
HVHP14250 0.45 µm 142 mm 50
VVHP04700 0.1 µm 47 mm 100
Accessory
Product No. Description Pack Size Add to Cart
XX6200006P Filter forceps, blunt end, stainless steel 3
Related Links