Epidermal growth factor receptor (EGFR) signaling
Background
Epidermal growth factor receptor (EGFR) is the first discovered and prototypical member of receptor tyrosine kinase family (RTK) of receptors. It is activated by various ligands in extracellular milieu and transmits the cellular response to mediate various cellular activities, including cell proliferation, cell survival, growth and development. EGFR is expressed in many organs and aberrant expression is implicated in various cancers. This review highlights various signaling components of EGFR pathway and their signal transduction pathways.
Epidermal Growth Factor Receptors (EGFR) and ligands
The ErbB family comprises of four receptors, which include EGFR (ErbB-1/HER1), ErbB-2 (Neu, HER2), ErbB-3 (HER3) and ErbB-4 (HER4). This receptor tyrosine kinase family (RTK) of proteins has an extracellular ligand-binding domain, hydrophobic transmembrane domain and a cytoplasmic tyrosine kinase domain. ErbB receptors are activated by growth factors of EGF-family characterized by three disulphide-bonds that confer binding specificity. Additional structural motifs include immunoglobulin-like domains, heparin-binding sites and glycosylation sites1,2. EGFR ligands and their specific receptors are given in Table 1.
Table 1. EGFR receptors & ligands
Receptor
Ligands
EGFR EGF, Transforming growth factor-α (TGF-α), Amphiregulin (AR), Betacellulin (BTC), Heparin-binding epidermal growth factor (HB-EGF) and Epiregulin (EPR)
ErbB2 None
ErbB3 NRG-1 and NRG-2
ErbB4 Betacellulin (BTC), Heparin-binding EGF-like growth factor (HB-EGF), Epiregulin (EPR), Tomoregulin, NRG-1, NRG2, NRG3 and NRG4
Receptor activation
Receptor activation is triggered by the cascade of events on cell membrane.
  • Ligand binding: Each ligand binds to the extracellular domain of cognate ErbB receptors
  • Receptor dimerization: Ligand binding induces the formation of receptor homo or hetero dimers3.
  • Activation of kinase domain: Autophosphorylation of key tyrosine residues within carboxy terminal tail activates  the receptor and acts as docking sites for proteins with Src homology 2 (SH2) and phosphotyrosine binding domain (PTB) to trigger cellular signaling3,4
EGFR Signaling
Image 1. EGFR Signaling
EGFR Signaling
Ras/Raf signaling cascade
Receptors: Dimerization of any two ErbB receptors
Key functions: Cell survival and cell proliferation5
After receptor activation, the complex formed by Grb2 and Sos binds directly or through association of adapter protein Shc, to specific tyrosine residues on the receptor6,7. This leads to conformational change in Sos, which can recruit and activate Ras-GDP. Ras-GDP activates Raf-1, which further activates extracellular regulated kinases 1 and 2 (ERK1 and ERK2) mediated through mitogen-activated protein kinases (MAPK)8,9. Activated kinases eventually move into the nucleus to phosphorylate specific transcription factors like Elk1 and C-myc to induce cell proliferation.
Phosphatidylinositol 3-kinase/Akt signaling cascade
Receptors: Dimerization of ErbB2 with either ErbB4 or ErbB3
Key functions: Cell growth, apoptosis resistance, cell invasion and migration10
Phosphatidylinositol comprises of p85 and p110 sub units that dock to the ErbB receptor to generate secondary messenger phosphatidylinositol 3,4,5-triphosphate, that further activates serine/threonine kinase AKT. Upon activation, AKT phosphorylates mTOR and subsequently S6K which mediates protein synthesis10.
Signal transducers and activators of transcription (STAT) pathway
Receptors: ErbB
Key functions: Tumor progression, oncogenesis and angiogenesis11
STAT proteins docks to the phosphotyrosine residues of ErbB receptors via Src homology 2 domains and up on dimerization, translocate into the nucleus to promote the expression of specific target genes like Myc, Nos2, p21 and cytokines12.
Phospholipase Cγ signaling
Receptors: ErbB1
Key functions: Regulation of ion channels, cell migration, calcium-mediated signaling13
Phospholipase Cγ interacts with ErbB1 and hydrolyses phosphatidylinositol 4,5-diphosphate (PIP2) to generate inositol 1,3,5-triphosphate (IP3) and 1,2 diacylglycerol (DAG). IP3 increases intracellular calcium levels and DAG mediates activation of protein kinase C (PKC)13. Activated PKC in turn activates MAPK and c-Jun NH2-terminal kinase14.
Nck/PAK signaling cascade
Receptors: ErbB1
Key functions: Cell survival and cell migration15
Nck is an adaptor protein containing SH2 domain that binds to EGF receptor and triggers downstream signaling. Nck activates PAK1 (p21/CDC42/Rac1-Activated Kinase-1) binding through SH3 domain. Activated PAK1 in turn activates JNKs (c-Jun Kinases) mediated by MEKK1 (MAP/ERK Kinase Kinase-1) and MKK4/7 (MAP Kinase Kinase-4/7), respectively. JNK translocates to the nucleus and phosphorylates transcription factors like c-Fos and c-Jun16.
Cbl mediated endocytosis
Receptors: ErbB1
Key functions: Endocytosis
Following ligand binding, Cbl is a substrate that binds to the EGF receptors through SH2 domains or through GRB2 adaptor protein and triggers lysosomal degradation of receptors17.
EGFR translocation into nucleus
EGF Receptors have an ability to escape lysosomal degradation and translocate into the nucleus to mediate biological functions. In the nucleus, these receptors promote transcription of cell survival genes like Cyclin D1 gene and also act as cofactors for STAT and E2F1 transcription factors18.  Nuclear localization of EGFR is implicated in disease severity by conferring resistance to growth inhibitory effects of anti-cancer mAbs19.
Our most popular, high-quality EGF proteins for your research use
Product # SRP3196 SRP3329 SRP3027 E9644 E5036 E4127 E5160 E1257 SRP6253
Species Mouse Mouse Human Human Human Mouse Mouse Mouse Human
Expressed in/ extracted from E. coli E. coli E. coli E. coli E. coli murine sub maxillary gland E. coli murine sub maxillary gland HEK 293
Endotoxin-tested
Carrier-free
Purity ≥98% ≥98% ≥98% ≥97% ≥97%   ≥90%   ≥97%
ED50/EC50 ≤0.1 ng/ml ≤0.1 ng/ml ≤0.1 ng/ml 0.08-0.8 ng/mL 0.08-0.8 ng/mL 0.1-10 ng/mL 0.05-1 ng/mL 0.02-2.0 ng/mL 0.1-10 ng/mL
EGFRs in Development & Disease
EGF receptors and their ligands are important for development of various organs; EGFR knockout models show embryonic lethality and defective tissues/organs. However, mutations in ErbB ligands do not show any lethal phenotype, because the complimentary pathways of other ligands compensate the abrogated signals of mutated ErbB ligand.20-25 EGFR are abnormally activated or expressed in many diseases. Specifically, their role in cancer progression is well investigated. Some of genetic alterations and their phenotypic effects are summarized in Table 2.
Table 2. Genetic alterations of EGFR receptors in human disorders
Genetic Alteration Implication
EGFR mutation Non-small cell lung cancer26
Cetuximab resistance in colorectal cancer27
Non-small-cell lung cancer28
Anaplastic and follicular thyroid cancer29
Lung adenocarcinomas30
Inflammatory skin and bowel disease36
Overexpression of ErbB2 Gastric and breast cancer31,32
Overexpression of ErbB2 and ErbB3 Prostate cancer33
Dysregulated EGFR signaling Polycystic kidney disease, glomerulonephritis, diabetic nephropathy34
Overexpression of EGFR Generation of new oligodendrocytes potential treatment of white matter injury in pre-mature children35
Conclusion
EGFR signaling has pleiotropic functions in development and regulates various physiological functions. The aberrant expression of EGF receptors has been increasingly implicated in various disorders. More research is yet to clearly uncover the role of EGFR signaling in development of various organs including, brain, heart, skin, kidneys, mammary glands and lungs. Efforts to identify novel drugs with high efficacy, sustained drug activity and less cross reactivity to combat drug-resistant cancer conditions continue to pique lot of research interest.
Explore complete offering of high quality EGF recombinant proteins and receptors for your research use here.
Study EGF signaling with consistency in your stem cell cultures. Visit our Stem Cell Learning Center to learn more.
 References
  1. Arteaga, C. L., and Engelman, J. A. (2014) ERBB receptors: from oncogene discovery to basic science to mechanism-based cancer therapeutics. Cancer Cell 25, 282–303.
  2. Kovacs, E., Zorn, J. A., Huang, Y., Barros, T., and Kuriyan, J. (2015) A structural perspective on the regulation of the epidermal growth factor receptor. Annu. Rev. Biochem. 84, 739–764.
  3. Capuani, F., Conte, A., Argenzio, E., Marchetti, L., Priami, C., Polo, S., Di Fiore, P. P., Sigismund, S., and Ciliberto, A. (2015) Quantitative analysis reveals how EGFR activation and downregulation are coupled in normal but not in cancer cells. Nat. Commun. 6, 7999.
  4. Zhang, X., Gureasko, J., Shen, K., Cole, P. A., and Kuriyan, J. (2006) An allosteric mechanism for activation of the kinase domain of epidermal growth factor receptor. Cell 125, 1137–1149.
  5. Gaestel, M. (2006) MAPKAP kinases - MKs - two’s company, three’s a crowd. Nat. Rev. Mol. Cell Biol. 7, 120–130.
  6. Lowenstein, E. J., Daly, R. J., Batzer, A. G., Li, W., Margolis, B., Lammers, R., Ullrich, A., Skolnik, E. Y., Bar-Sagi, D., and Schlessinger, J. (1992) The SH2 and SH3 domain-containing protein GRB2 links receptor tyrosine kinases to ras signaling. Cell 70, 431–442.
  7. Batzer, A. G., Rotin, D., Ureña, J. M., Skolnik, E. Y., and Schlessinger, J. (1994) Hierarchy of binding sites for Grb2 and Shc on the epidermal growth factor receptor. Mol. Cell. Biol. 14, 5192–5201.
  8. Hallberg, B., Rayter, S. I., and Downward, J. (1994) Interaction of Ras and Raf in intact mammalian cells upon extracellular stimulation. J. Biol. Chem. 269, 3913–3916.
  9. Liebmann, C. (2001) Regulation of MAP kinase activity by peptide receptor signalling pathway: paradigms of multiplicity. Cell. Signal. 13, 777–785.
  10. Vivanco, I., and Sawyers, C. L. (2002) The phosphatidylinositol 3-Kinase AKT pathway in human cancer. Nat. Rev. Cancer 2, 489–501.
  11. Niu, G., Wright, K. L., Huang, M., Song, L., Haura, E., Turkson, J., Zhang, S., Wang, T., Sinibaldi, D., Coppola, D., Heller, R., Ellis, L. M., Karras, J., Bromberg, J., Pardoll, D., Jove, R., and Yu, H. (2002) Constitutive Stat3 activity up-regulates VEGF expression and tumor angiogenesis. Oncogene 21, 2000–2008.
  12. Bromberg, J. (2002) Stat proteins and oncogenesis. J. Clin. Invest. 109, 1139–1142.
  13. Patterson, R. L., van Rossum, D. B., Nikolaidis, N., Gill, D. L., and Snyder, S. H. (2005) Phospholipase C-gamma: diverse roles in receptor-mediated calcium signaling. Trends Biochem. Sci. 30, 688–697.
  14. Schönwasser, D. C., Marais, R. M., Marshall, C. J., and Parker, P. J. (1998) Activation of the mitogen-activated protein kinase/extracellular signal-regulated kinase pathway by conventional, novel, and atypical protein kinase C isotypes. Mol. Cell. Biol. 18, 790–798.
  15. Ye, D. Z., and Field, J. (2012) PAK signaling in cancer. Cell. Logist. 2, 105–116.
  16. Tomar, A., and Schlaepfer, D. D. (2010) A PAK-activated linker for EGFR and FAK. Dev. Cell 18, 170–172.
  17. Yu, P., Fan, Y., Qu, X., Zhang, J., Song, N., Liu, J., and Liu, Y. (2016) Cbl-b regulates the sensitivity of cetuximab through ubiquitin-proteasome system in human gastric cancer cells. J. BUON Off. J. Balk. Union Oncol. 21, 867–873.
  18. Lo, H.-W., Hsu, S.-C., Ali-Seyed, M., Gunduz, M., Xia, W., Wei, Y., Bartholomeusz, G., Shih, J.-Y., and Hung, M.-C. (2005) Nuclear interaction of EGFR and STAT3 in the activation of the iNOS/NO pathway. Cancer Cell 7, 575–589.
  19. Lo, H.-W., Xia, W., Wei, Y., Ali-Seyed, M., Huang, S.-F., and Hung, M.-C. (2005) Novel prognostic value of nuclear epidermal growth factor receptor in breast cancer. Cancer Res. 65, 338–348.
  20. Miettinen, P. J., Berger, J. E., Meneses, J., Phung, Y., Pedersen, R. A., Werb, Z., and Derynck, R. (1995) Epithelial immaturity and multiorgan failure in mice lacking epidermal growth factor receptor. Nature 376, 337–341.
  21. Sibilia, M., and Wagner, E. F. (1995) Strain-dependent epithelial defects in mice lacking the EGF receptor. Science 269, 234–238.
  22. Park, S. K., Miller, R., Krane, I., and Vartanian, T. (2001) The erbB2 gene is required for the development of terminally differentiated spinal cord oligodendrocytes. J. Cell Biol. 154, 1245–1258.
  23. Leu, M., Bellmunt, E., Schwander, M., Fariñas, I., Brenner, H. R., and Müller, U. (2003) Erbb2 regulates neuromuscular synapse formation and is essential for muscle spindle development. Dev. Camb. Engl. 130, 2291–2301.
  24. Erickson, S. L., O’Shea, K. S., Ghaboosi, N., Loverro, L., Frantz, G., Bauer, M., Lu, L. H., and Moore, M. W. (1997) ErbB3 is required for normal cerebellar and cardiac development: a comparison with ErbB2-and heregulin-deficient mice. Dev. Camb. Engl. 124, 4999–5011.
  25. Golub, M. S., Germann, S. L., and Lloyd, K. C. K. (2004) Behavioral characteristics of a nervous system-specific erbB4 knock-out mouse. Behav. Brain Res. 153, 159–170.
  26. Maheswaran, S., Sequist, L. V., Nagrath, S., Ulkus, L., Brannigan, B., Collura, C. V., Inserra, E., Diederichs, S., Iafrate, A. J., Bell, D. W., Digumarthy, S., Muzikansky, A., Irimia, D., Settleman, J., Tompkins, R. G., Lynch, T. J., Toner, M., and Haber, D. A. (2008) Detection of mutations in EGFR in circulating lung-cancer cells. N. Engl. J. Med. 359, 366–377.
  27. Montagut, C., Dalmases, A., Bellosillo, B., Crespo, M., Pairet, S., Iglesias, M., Salido, M., Gallen, M., Marsters, S., Tsai, S. P., Minoche, A., Seshagiri, S., Somasekar, S., Serrano, S., Himmelbauer, H., Bellmunt, J., Rovira, A., Settleman, J., Bosch, F., and Albanell, J. (2012) Identification of a mutation in the extracellular domain of the Epidermal Growth Factor Receptor conferring cetuximab resistance in colorectal cancer. Nat. Med. 18, 221–223.
  28. Rosell, R., Moran, T., Queralt, C., Porta, R., Cardenal, F., Camps, C., Majem, M., Lopez-Vivanco, G., Isla, D., Provencio, M., Insa, A., Massuti, B., Gonzalez-Larriba, J. L., Paz-Ares, L., Bover, I., Garcia-Campelo, R., Moreno, M. A., Catot, S., Rolfo, C., Reguart, N., Palmero, R., Sánchez, J. M., Bastus, R., Mayo, C., Bertran-Alamillo, J., Molina, M. A., Sanchez, J. J., Taron, M., and Spanish Lung Cancer Group. (2009) Screening for epidermal growth factor receptor mutations in lung cancer. N. Engl. J. Med. 361, 958–967.
  29. Liu, Z., Hou, P., Ji, M., Guan, H., Studeman, K., Jensen, K., Vasko, V., El-Naggar, A. K., and Xing, M. (2008) Highly prevalent genetic alterations in receptor tyrosine kinases and phosphatidylinositol 3-kinase/akt and mitogen-activated protein kinase pathways in anaplastic and follicular thyroid cancers. J. Clin. Endocrinol. Metab. 93, 3106–3116.
  30. Gao, S. P., Mark, K. G., Leslie, K., Pao, W., Motoi, N., Gerald, W. L., Travis, W. D., Bornmann, W., Veach, D., Clarkson, B., and Bromberg, J. F. (2007) Mutations in the EGFR kinase domain mediate STAT3 activation via IL-6 production in human lung adenocarcinomas. J. Clin. Invest. 117, 3846–3856.
  31. Fukushige, S., Murotsu, T., and Matsubara, K. (1986) Chromosomal assignment of human genes for gastrin, thyrotropin (TSH)-beta subunit and C-erbB-2 by chromosome sorting combined with velocity sedimentation and Southern hybridization. Biochem. Biophys. Res. Commun. 134, 477–483.
  32. Slamon, D. J., Godolphin, W., Jones, L. A., Holt, J. A., Wong, S. G., Keith, D. E., Levin, W. J., Stuart, S. G., Udove, J., and Ullrich, A. (1989) Studies of the HER-2/neu proto-oncogene in human breast and ovarian cancer. Science 244, 707–712.
  33. Qiu, Y., Ravi, L., and Kung, H. J. (1998) Requirement of ErbB2 for signalling by interleukin-6 in prostate carcinoma cells. Nature 393, 83–85.
  34. Harskamp, L. R., Gansevoort, R. T., van Goor, H., and Meijer, E. (2016) The epidermal growth factor receptor pathway in chronic kidney diseases. Nat. Rev. Nephrol. 12, 496–506.
  35. Scafidi, J., Hammond, T. R., Scafidi, S., Ritter, J., Jablonska, B., Roncal, M., Szigeti-Buck, K., Coman, D., Huang, Y., McCarter, R. J., Hyder, F., Horvath, T. L., and Gallo, V. (2014) Intranasal epidermal growth factor treatment rescues neonatal brain injury. Nature 506, 230–234.
  36. Campbell, P., Morton, P. E., Takeichi, T., Salam, A., Roberts, N., Proudfoot, L. E., Mellerio, J. E., Aminu, K., Wellington, C., Patil, S. N., Akiyama, M., Liu, L., McMillan, J. R., Aristodemou, S., Ishida-Yamamoto, A., Abdul-Wahab, A., Petrof, G., Fong, K., Harnchoowong, S., Stone, K. L., Harper, J. I., McLean, W. H. I., Simpson, M. A., Parsons, M., and McGrath, J. A. (2014) Epithelial inflammation resulting from an inherited loss-of-function mutation in EGFR. J. Invest. Dermatol. 134, 2570–2578.