Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Ethidium Bromide
Product Nos. E1510, E1385, E7637, and E8751
Synonyms: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide; 2,7-Diamino-10-ethyl-9-phenylphenanthridinium bromide; Homidium Bromide; Dromilac; EtBr
Product Description
Appearance: Red to purple powder
Molecular formula: C21H20N3Br
Molecular mass: 394.3
Melting point: 255-266°C with decomposition1,2
Spectral data:
UV/Vis Absorbance:
Maxima at 210 nm, EmM = 0.20-0.5; 285 nm, EmM = 5.0-10.0; 316 nm, EmM = 50.0; 343 nm, EmM = 40.0 (water)3
Published absorption spectrum (methanol) with λmax at 296 nm and
525 nm1
λmax = 475 nm, EmM = 5.76 (0.66 M glycine buffer)4
Solvent effects on absorbance:
λmax = 480 nm (water); 515 (glycerol); 520 nm (methanol): 520 nm (acetone); 532 nm (ethanol); 535 (DMSO) and 540 nm (pyridine).5 The absorbance maximum relative to water is red-shifted to longer wavelengths upon binding to nucleic acids.6,7
Fluorescence:
Excitation/emission wavelengths reported for the EtBr-nucleic acid complex:
Ex at 526 nm, Em at 605 nm (aqueous)8
Ex at 360 nm, Em at 590 nm (in PBS)9
Ex at 525 nm, Em at 600 nm (10 mM TBE, pH 8.0)10
Ex (absorption) at 510 nm, Em at 590 nm.11
Ex (absorption) at 482 nm (blue-green), Em at 616 nm (red-orange).12 The fluorescence yield of EtBr increases as solvent polarity decreases.13
Ethidium bromide is a well-known and widely used fluorescent dye in biotechnology research. Early usage was as a veterinary trypanocide.14 It is a mutagenic compound which intercalates double-stranded DNA and RNA.1,10 The fluorescence of EtBr increases 21-fold upon binding to double-stranded RNA, 25-fold on binding double-stranded DNA (although histones block binding of EtBr to DNA). Ethidium bromide has been used in a number of fluorimetric assays for nucleic acids.15,16,17 It has been shown to bind to single-stranded DNA (although not as strongly)2 and triple-stranded DNA.18 Because of the binding to DNA, EtBr is a powerful inhibitor of DNA polymerase.4
E7637, Molecular Biology grade powder, is suitable for use in gel electrophoresis and DNA isolation procedures. E1510, Aqueous Solution (10 mg per ml), is suitable for use in gel electrophoresis and DNA isolation procedures. E1385 Molecular Biology grade Aqueous Solution (500 mg per ml), is suitable for use in gel electrophoresis.
Related Products
Methylene blue (M4159) was reported as an alternative stain.19 Thiazole orange homodimer (monomer = Product No. 390062) was noted as nonmutagenic, but useful for detecting DNA in agarose gels.20
For use in cell studies, dihydroethidium (Product No. D7008), also known as hydroethidine, is a possible alternative. It is the reduced form of the dye and is converted (dehydrogenated) inside the cell to the ethidium cation, which then intercalates into DNA.21,22
At present, the recommended ultimate disposal method is by incineration as discussed in the SDS.23 Processing solutions through Rohm and Haas Amberlite XAD-16 resin was shown to be effective to the limit of detection, with none of the completely decontaminated solution found to be mutagenic.24,25 Sigma offers XAD-16 as a bulk product, as well as several products that adsorb the dye for easier disposal. See Rezorian A161 cartridges (57611 or 57610) and the Extractor™ for EtBr decontamination (Z361569). A 5 ml Rezorian A161 cartridge can be used to treat more than 16 liters of solution (0.5 mg/liter of EtBr) before dye breakthrough. The luer lock cartridge can be used with 4 mm I.D. tubing, syringes or low-pressure chromatographic systems. The Extractor™ for EtBr decontamination can process up to 10 liters of solution (0.5 mg/liter of EtBr). Other methods of treatment for disposal of aqueous EtBr solutions have been suggested.26,27,28 Sigma has not verified and does not endorse these procedures.
Preparation Instructions
At room temperature, EtBr dissolves in water at 10 mg/ml to give a red solution.2 It should be soluble up to 20 mg/ml in water or up to 2 mg/ml in ethanol.1 It is soluble 1 part in 750 parts chloroform.3
Stock solutions of EtBr in water or PBS are stable for at least two years at room temperature if protected from light.11
Storage/Stability
The solid powder has shown minimal change after two years stored at room temperature, protected from light.
Procedure
Use in Electrophoresis Staining6
  1. The dye is usually incorporated into the gel and the electrophoresis buffer at 0.5 mg/ml. Note: Electrophoresis mobility of linear double-stranded DNA is reduced by approximately 15% in the presence of the dye.
  2. To stain after gel has been run, immerse gel in electrophoresis buffer or water containing EtBr (0.5 mg/ml) for 30-45 minutes at room temperature.
  3. Destaining is optional. Detection of very small amounts (<10 ng) of DNA is made easier if the background fluorescence caused by unbound EtBr is reduced by soaking the stained gel in water or 1 mM MgSO4 for 20 minutes at room temperature.
Materials
 References
  1. Green, F.J., Sigma Aldrich Handbook of Stains, Dyes and Indicators, p. 318.
  2. Sigma quality control; Sigma molecular biology laboratories.
  3. Merck Index, 12th ed., #4767 (1996).
  4. W.H. Elliott, Biochem. J., 86, 562-567 (1963).
  5. Olmsted, J., III. and Kerans, D.R., Biochem., 16, 3647 (1977).
  6. Sambrook, J. et al., Molecular Cloning: A Laboratory Manual, 2nd ed., (Cold Spring Harbor Laboratory, 1989), p 6.15, 6.44 etc.
  7. Le Pecq, J.-B., in Methods of Biochemical Analysis, 20, 43-86.
  8. Bechtol, K.B. et al., American Biotechnology Lab, p. 8, December 1994.
  9. Anal. Biochem., 65, 225 (1975).
  10. Severini, A. and Morgan, A.R., Anal. Biochem., 193, 83-89 (1991).
  11. Crary, B. and Boyrsenko, M., Biotechniques, 4, 98-101 (1986).
  12. Givan, A., Flow Cytometry: First Principles (Wiley-Liss, NY) p. 104 and p. 64,
  13. Methods in Enzymology, 32, 239 (1974).
  14. Watkins, T.I., J. Chem. Soc., 3059-3064, (1952).
  15. Moore, S.P. and Sutherland, B.M., Anal. Biochem., 144, 15-19 (1985).
  16. El-Hamalawi, A.-R. A. et al., Anal. Biochem., 67, 384 (1975).
  17. Karsten, U. and Wollenberger, A., Anal. Biochem., 77, 464 (1977).
  18. Scaria, P.V. and Shafer, R.H., J. Biol. Chem., 266, 5417-5423 (1991).
  19. Technique: A Journal of Methods in Cell and Molec. Biol., 1, 183-187 (1989).
  20. Wilke, W.W. et al., Mod. Pathol., 7 (3), 385-387 (1994).
  21. Saiki, I. et al., J. Natl. Cancer Inst., 77 (6),1235-1240 (1986).
  22. Budd, S.L et al., FEBS Lett., 415, 21-24 (1997).
  23. Sigma-Aldrich Safety Data Sheet (SDS).
  24. Joshua, H., BioTechniques, 4 (3), 207-208 (1986).
  25.  Lunn, G. and Sansone, E.B., Biotech. Histochem., 66, 307-315 (1991).
  26. Lunn, G. and Sansone, E.B., Anal. Biochem., 162, 453 (1987).
  27. Communications from M.A. Armour, University of Alberta, Edmonton, Canada – prior to presentation at First Asian Symposium on Academic Activity for Water Treatment, Tokyo (1992).
  28. Zocher, R. et al., Biol. Chem. Hoppe Seyler, 369 (10), 1191-1194 (1988).
ckv 04/29/99
Related Links