Evaluation of Virus Production in Vero Cells Grown in EX-CELL™ Vero Serum-Free Medium
Rebecca S. Schuchhardt, Kevin Kolell, Sarah L. Gilliland
Abstract
EX-CELL Vero Serum-Free Medium for Vero Cells is an animalcomponent free, low-protein medium designed and optimized to support high density cultures of African Green Monkey Kidney (Vero) cells. The Vero cell line can be easily adapted to EX-CELL Vero using a direct adaptation method. EX-CELL Vero supports Vero cell density up to 2.5 x 105 cells/cm2 with doubling time as short as 29 hours. Vero cells in EX-CELL Vero were infected with Herpes Simplex Virus-Type 2 (HSV-2) and produced on average 106.0 TCID50/mL at a multiplicity of infection (MOI) of 0.01. It was concluded that EX-CELL Vero supports rapid Vero cell growth and produces HSV-2 titers comparable to serum-supplemented cultures.
Introduction
The Vero cell line is used in applications such as development of viral vaccines and the production of recombinant adenoviral vectors. Traditionally, Vero cells are grown in serum-supplemented basal medium such as Dulbecco's Modified Eagle's Medium (DMEM). EX-CELL Vero is an animal-component free, lowprotein, serum-free medium specially formulated to support largescale, high-density Vero culture and virus production. The medium contains very low levels of recombinant protein (approximately 0.5 mg/L), facilitating downstream processing of expressed products and eliminating regulatory concerns associated with serum and animal proteins. Additionally, the liquid formulation of EX-CELL Vero is formulated without L-glutamine, which avoids problems associated with L-glutamine degradation, therefore improving product shelf life. Our experiments show that EX-CELL Vero supports high-density, serum-free Vero cell growth and the production of virus yields comparable to serumsupplemented cultures.
Materials
Cells
  • African Green Monkey Kidney cell line, American Type Culture Collection, Catalog No. CCL-81
Virus
  • Herpes Simplex Virus 2 (HSV-2), American Type Culture Collection, Catalog No. VR-540
Serum-Free Media
  • EX-CELL Vero, SAFC Biosciences, Catalog No. 14585
Other Media and Supplements
  • Dulbecco's Modified Eagle's Medium/High Modified (DMEM/High), SAFC Biosciences, Catalog No. 51444
  • Fetal Bovine Serum (FBS) Gamma Irradiated, SAFC Biosciences, Catalog No. 12107
  • L-Glutamine Solution 200mM, SAFC Biosciences, Catalog No. 59202
  • Trypsin-EDTA Solution 1X, 0.25% trypsin, 0.1% EDTA, trypsin gamma irradiated by SER-TAINTM Process, SAFC Biosciences, Catalog No. 59429
  • Dulbecco's Phosphate Buffered Saline (DPBS Modified), SAFC Biosciences, Catalog No. 59321
  • Trypsin inhibitor from Glycine max (soybean) (STI), Sigma-Aldrich Co., Catalog No. T6522
Methods
EX-CELL Vero was prepared by adding L-glutamine at a final concentration of 4 mM at time of use. Unless otherwise noted, all EX-CELL Vero was supplemented before use. 5% FBS was added to DMEM/High at time of use (referred to as DMEM-5%). STI was prepared as a concentrated (10 mg/mL) solution in DPBS and filter sterilized (0.2 μm). Working stock solutions of STI were diluted to 1 mg/mL with sterile DPBS as needed. All media were stored at 2 to 8 C, protected from light. Other supplements were stored at their recommended temperatures.
Basic Culture Techniques
Cells were routinely subcultured every four days (96 hours ± 6 hours) at a seeding density of 2 x 104 cells/cm2 in EX-CELL Vero and DMEM-5% in 75 cm2 vent cap T-flasks. The total volume of media was 20 mL per flask. Cultures were maintained at 37 ± 1 C in a humidified incubator with 5% CO2. All experiments were performed in duplicate flasks and cell counts were determined using a Cedex counter. All cultures were maintained using aseptic technique and without the use of antibiotics or fungicides.
Trypsinization
Vero cultures in EX-CELL Vero and DMEM-5%:
Spent medium was aspirated and the monolayer rinsed with 5 mL DPBS. The DPBS was aspirated and 1 mL trypsin was added to the flasks. The flasks were incubated at 37 C for 3 - 5 minutes until the cells dissociated. 4.5 mL of STI was then added to the flasks to neutralize the trypsin, the cells were gently resuspended in 4.5 mL of the respective medium and cultures were placed in a 15 mL conical tube. Cultures were centrifuged at 228 g for five minutes to pellet the cells. The supernatant was removed, discarded and the pellet resuspended in 10 mL of the respective medium. A 1 mL aliquot was taken for counting on the Cedex and cells were subsequently diluted in the appropriate quantity of prepared medium and incubated as above.
Notes:
  1. Vero cells in both media appeared sensitive to trypsin. It was found that using a lower volume of STI did not fully inactivate the trypsin and caused the cells to exhibit an unusual morphology.
  2. Over-trypsinizing the cells may occur quickly. It is important to inactivate the trypsin within 15 minutes (less time in smaller flasks). Over-trypsinized cells do not recover.
Direct Adaptation
Vero cultures were started from frozen cells in DMEM-5% in 25 cm2 T-flasks and were expanded and maintained in 75 cm2 T-flasks. Vero cells were subcultured directly into EX-CELL Vero at a seeding density of 2 x 104 cells/cm2. The cells reached 100% confluency within three days and displayed normal doubling times.
Herpes Simplex Virus-2 Production
To determine the capability of EX-CELL Vero to support the production of virus, Vero cells were infected with Herpes Simplex Virus-2 (HSV-2). Vero cells were seeded in EX-CELL Vero and DMEM-5% in 25 cm2 T-flasks at 2 x 104 cells/cm2 in a total volume of 5 mL medium per flask. Flasks were incubated at 37 C with 5% CO2. Once the flasks were confluent (Figure 1 and Figure 2), one flask of each condition was sacrificed and the total cell count was determined. The cells were then infected with the appropriate amount of virus to yield a MOI of 0.01. Flasks were incubated as above and during the infection the cells were monitored and displayed cytopathetic effect (CPE). The cells became rounded and many, although not all, of the cells detached from the bottom of the flask. The flasks were harvested when greater than 75% CPE was observed, which was Day 4 (Figure 3 and Figure 4). Prior to titration, each flask was subjected to three rounds of thawing (37 C) and freezing (-70 C) to lyse the cells. Any remaining attached cells were sloughed off the flask by rapping the flasks against the palm of the hand and by pipetting the lysate forcefully against the bottom of the flask. The cell lysates were transferred to 15 mL conical tubes and centrifuged at ~2000 g for 10 minutes to pellet the cell debris. The supernatants were transferred to new tubes and titrated.
Figure 1: Point of Infection of Vero cells in EX-CELL Vero (Day 4).
Figure 2: Point of infection of Vero cells in DMEM-5% (Day 4).
Figure 3: Day 4 of Vero cells infected with HSV-2 in DMEM-5%.
This was the point of harvest (Day 4 post-infection).
Figure 4: Day 4 of Vero cells infected with HSV-2 in EX-CELL Vero.
This was the point of harvest (Day 4 post-infection).
TCID50/mL Titrations
HSV-2 production in EX-CELL Vero was determined by TCID50. Vero cells growing in DMEM-5% were harvested by trypsinization, counted and diluted to a final concentration of 2 x 105 cells/mL in DMEM supplemented with 5% FBS (DMEM-5%). Using a 12-channel pipettor, 100 μL of cells were dispensed into 96-well microtiter plates (2 x 104 cells/well) and allowed to attach at 37 C, 5% CO2 for two days. Duplicate serial dilutions of each lysate were made in DMEM-2% in sterile snap-cap disposable tubes (1:10 dilutions, 10-4 to 10-8 plated). The spent medium on the 96-well plates was decanted into a sterile container under the laminar flow hood. The dilutions were plated in duplicate on each plate in wells 1 - 10 (wells 11 and 12 in all rows served as negative controls). The rows were dispensed with the highest (10-8) dilution in column 10 and the lowest dilution (10-4) in column 6. The second dilution of each set was dispensed in columns 5 to 1. The plates were incubated at 37 C, 5% CO2 for three days and then observed on an inverted microscope for CPE. Wells were considered positive even if only a small area of the well showed CPE. The titer was determined using the Reed and Muench statistical method1.
Conclusions
Vero cells were adapted from DMEM-5% to EX-CELL Vero in attached cultures. During adaptation a seeding density of 2 x 104 cells/cm2 was used to obtain cultures that reached confluency within four days. Vero cells took 3 - 4 passages to adapt to EX-CELL Vero.
EX-CELL Vero supported HSV-2 production in Vero cells. The cells exhibited classic CPE in culture and produced HSV-2 titers in the range of 106.0 TCID50/mL. As shown in Figure 5, this range is comparable with those seen in serumsupplemented cultures.
Figure 5: Vero cells were infected with HSV-2 96 hours post-seeding at a MOI of 0.01. Flasks
were harvested 96 hours post-infection. Results represent TCID50/mL at the harvest point.
 Reference
  1. Reed. L.J. and Muench, H.A., A Simple Method of Estimating Fifty Per Cent Endpoints, Am J Hyg. 1938;27:493-7.
EX-CELL™ and SER-TAIN™ are trademarks of SAFC Biosciences, Inc.
Related Links