Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Fluoropore™ and Omnipore™ PTFE Membrane Filters
Fluoropore™ and Omnipore™ PTFE Membrane FiltersPolytetrafluoroethylene (PTFE) membrane filters, with their broad chemical compatibility, are versatile filters for general clarification. Choose Fluoropore™ Hydrophobic PTFE Membrane Filters for filtering organic-based liquids or gases, and choose and Omnipore™ Hydrophilic PTFE Membrane Filters for filtering aqueous solutions.
Radiation monitoring: Fluoropore™ membranes are particularly suited to radioactive alpha particle monitoring, because they trap these particles on their surface, resulting in efficient detection with high resolution. Furthermore, they are backed with a high-contrast HDPE, making them easier to handle and install in radiation monitors.
Fluoropore™ and Omnipore™ Membrane Disc Specifications
  Fluoropore™ Membrane* Omnipore™ Membrane
Membrane Material Hydrophobic Polytetrafluoroethylene
(PTFE)
Hydrophilic Polytetrafluoroethylene
(PTFE)
Pore Sizes (μm) 0.2 - 3 0.1 - 10
Thickness (μm) 50 -150 30 - 85
Porosity (%) 85  
Liquid Flow (mL/min/cm2) @27.5 in. Hg 24 - 286 (methanol) 2 – 1250 (water)
Air Flow specs (L/min/cm2) @ 10 psi 5 - 20 n/a
Wettability Hydrophobic Hydrophilic
Temperature Limitations 130 °C max n/a
Chemical Compatibility Compatible with both aqueous and organic solvents Compatible with both aqueous and
organic solvents
Sterilizability Autoclave, EtO Autoclave, EtO
Surface Option White White
Key Properties Solvent-resistant, fast-flowing,
low pressure drop, low extractables,
low binding
Compatible with both aqueous
and organic solvents, broad chemical compatibility
Key Applications Clarifying acids, bases and solvents,
air monitoring, filtering and venting gases,
UV spectroscopy, radiation monitoring
Clarifying acids, bases and aqueous solutions
*All Fluoropore™ Membrane Filter Discs (except for Prod. No. FHUP04700) are bonded to a HDPE (high density polyethylene) support.
Fluoropore™ (PTFE) Membrane Filters
Product No. Description Pore Size Filter Diameter (⌀) Pack Size Add to Cart
FALP01300 Fluoropore™ Membrane Filter, PTFE, hydrophobic, 1.0 µm, 13 mm, white, plain 1.0 µm 13 mm 100
FALP02500 Fluoropore™ Membrane Filter, PTFE, hydrophobic, 1.0 µm, 25 mm, white, plain 1.0 µm 25 mm 100
FALP03700 Fluoropore™ Membrane Filter, PTFE, hydrophobic, 1.0 µm, 37 mm, white, plain w/pads 1.0 µm 37 mm 100
FALP04700 Fluoropore™ Membrane Filter, PTFE, hydrophobic, 1.0 µm, 47 mm, white, plain 1.0 µm 47 mm 100
FALP14250 Fluoropore™ Membrane Filter, PTFE, hydrophobic, 1.0 µm, 142 mm, white, plain 1.0 µm 142 mm 50
FGLP01300 Fluoropore™ Membrane Filter, PTFE, hydrophobic, 0.22 µm, 13 mm, white, plain 0.22 µm 13 mm 100
FGLP02500 Fluoropore™ Membrane Filter, PTFE, hydrophobic, 0.22 µm, 25 mm, white, plain 0.22 µm 25 mm 100
FGLP04700 Fluoropore™ Membrane Filter, PTFE, hydrophobic, 0.22 µm, 47 mm, white, plain 0.22 µm 47 mm 100
FGLP09050 Fluoropore™ Membrane Filter, PTFE, hydrophobic, 0.22 µm, 90 mm, white, plain 0.22 µm 90 mm 50
FGLP14250 Fluoropore™ Membrane Filter, PTFE, hydrophobic, 0.22 µm, 142 mm, white, plain 0.22 µm 142 mm 50
FHLP01300 Fluoropore™ Membrane Filter, PTFE, hydrophobic, 0.5 µm, 13 mm, white, plain 0.45 µm 13 mm 100
FHLP02500 Fluoropore™ Membrane Filter, PTFE, hydrophobic, 0.5 µm, 25 mm, white, plain 0.45 µm 25 mm 100
FHLP03700 Fluoropore™ Membrane Filter, PTFE, hydrophobic, 0.5 µm, 37 mm, white, plain 0.45 µm 37 mm 100
FHLP04700 Fluoropore™ Membrane Filter, PTFE, hydrophobic, 0.45 µm, 47 mm, white, plain 0.45 µm 47 mm 100
FHLP09050 Fluoropore™ Membrane Filter, PTFE, hydrophobic, 0.45 µm, 90 mm, white, plain 0.45 µm 90 mm 50
FHLP14250 Fluoropore™ Membrane Filter, PTFE, hydrophobic, 0.5 µm, 142 mm, white, plain 0.45 µm 142 mm 50
FSLW02500 Fluoropore™ Membrane Filter, PTFE, hydrophobic, 3.0 µm, 25 mm, white, plain 3.0 µm 25 mm 100
FSLW04700 Fluoropore™ Membrane Filter, PTFE, hydrophobic, 3.0 µm, 47 mm, white, plain 3.0 µm 47 mm 100
FSLW09025 Fluoropore™ Membrane Filter, PTFE, hydrophobic, 3.0 µm, 90 mm, white, plain 3.0 µm 90 mm 25
FSLW14200 Fluoropore™ Membrane Filter, PTFE, hydrophobic, 3.0 µm, 142 mm, white, plain 3.0 µm 142 mm 10
Unlaminated Fluoropore™ (PTFE) Membrane Filter
Product No. Description Pore Size Filter Diameter (⌀) Pack Size Add to Cart
FHUP04700 Fluoropore™ Membrane Filter, PTFE, hydrophobic, unlaminated, 0.5 µm, 47 mm, white, plain 0.45 µm 47 mm 100
Omnipore™ (hydrophilic PTFE) Membrane Filters
Product No. Description Pore Size Filter Diameter (⌀) Pack Size Add to Cart
JAWP01300 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 1.0 µm, 13 mm, white, plain 1.0 µm 13 mm 100
JAWP02500 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 1.0 µm, 25 mm, white, plain 1.0 µm 25 mm 100
JAWP04700 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 1.0 µm, 47 mm, white, plain 1.0 µm 47 mm 100
JAWP09025 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 1.0 µm, 90 mm, white, plain 1.0 µm 90 mm 25
JAWP14225 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 1.0 µm, 142 mm, white, plain 1.0 µm 142 mm 25
JCWP01300 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 10 µm, 13 mm, white, plain 10.0 µm 13 mm 100
JCWP02500 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 10 µm, 25 mm, white, plain 10.0 µm 25 mm 100
JCWP04700 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 10 µm, 47 mm, white, plain 10.0 µm 47 mm 100
JCWP09025 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 10 µm, 90 mm, white, plain 10.0 µm 90 mm 25
JCWP14225 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 10 µm, 142 mm, white, plain 10.0 µm 142 mm 25
JGWP01300 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 0.2 µm, 13 mm, white, plain 0.2 µm 13 mm 100
JGWP02500 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 0.2 µm, 25 mm, white, plain 0.2 µm 25 mm 100
JGWP04700 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 0.2 µm, 47 mm, white, plain 0.2 µm 47 mm 100
JGWP09025 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 0.2 µm, 90 mm, white, plain 0.2 µm 90 mm 25
JGWP14225 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 0.2 µm, 142 mm, white, plain 0.2 µm 142 mm 25
JHWP01300 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 0.45 µm, 13 mm, white, plain 0.45 µm 13 mm 100
JHWP02500 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 0.45 µm, 25 mm, white, plain 0.45 µm 25 mm 100
JHWP04700 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 0.45 µm, 47 mm, white, plain 0.45 µm 47 mm 100
JHWP09025 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 0.45 µm, 90 mm, white, plain 0.45 µm 90 mm 25
JHWP14225 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 0.45 µm, 142 mm, white, plain 0.45 µm 142 mm 25
JMWP01300 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 5.0 µm, 13 mm, white, plain 5.0 µm 13 mm 100
JMWP02500 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 5.0 µm, 25 mm, white, plain 5.0 µm 25 mm 100
JMWP04700 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 5.0 µm, 47 mm, white, plain 5.0 µm 47 mm 100
JMWP09025 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 5.0 µm, 90 mm, white, plain 5.0 µm 90 mm 25
JMWP14225 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 5.0 µm, 142 mm, white, plain 5.0 µm 142 mm 25
JVWP01300 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 0.1 µm, 13 mm, white, plain 0.1 µm 13 mm 100
JVWP02500 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 0.1 µm, 25 mm, white, plain 0.1 µm 25 mm 100
JVWP04700 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 0.1 µm, 47 mm, white, plain 0.1 µm 47 mm 100
JVWP09025 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 0.1 µm, 90 mm, white, plain 0.1 µm 90 mm 25
JVWP14225 Omnipore™ Membrane Filter, PTFE, Hydrophilic, 0.1 µm, 142 mm, white, plain 0.1 µm 142 mm 25