Formamide Molecular Biology Reagent
Product No. F9037
CAS RN 75-12-7
EC 200-842-0
Synonyms: Amide C1, Formic Amide, methanamide, carbamaldehyde
Product Description
Formamide is a reagent that is an ionizing solvent in aqueous buffers. It is widely utilized in biochemistry and molecular biology, particularly in nucleic acids research. Procedures have been reported for the use of formamide in DNA sequencing and in polyacrylamide sequencing gels, which helps to eliminate secondary structure in nucleic acids and thus compressions in the gel data.
Other protocols incorporate formamide for RNA denaturation, Northern blot hybridization and stripping, ribonuclease protection assays, RNA storage, and Southern hybridization.2 The use of formamide for the quantitation of mRNAs in hepatocytes solubilized in guanidium thiocyanate has been reported.3  Protocols have been described for the use of formamide in DNA isolation from cultured bacteriophage ë and cultured mammalian cells.2
Formamide may be included at levels up to 50% in a streptavidin-induced electrophoretic mobility shift procedure for the isolation of single-stranded DNA from PCR products.4
Formamide is also utilized in such large scale applications as the manufacture of formic esters, the production of hydrocyannic acid by catalytic dehydration, and as a softener for paper.1 A study of the use of formamide to recover estradiol degradation products in a transdermal drug delivery formulation has been published.5 A protocol that incorporates formamide into the analysis of â-blocker compounds by nonaqueous capillary electrophoresis coupled to electrospray ionization mass spectrometry has been described.6
Precautions and Disclaimer
This product is for R&D use only, not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices. 
Preparation Instructions
This product is miscible in water (formamide:water, 50% v/v), yielding a clear, colorless solution.
Storage/Stability
Storage Temperature 2-8 °C
This product is deionized and packaged under argon in amber glass bottles. Once the bottle is opened and the formamide is exposed to oxygen, this product will begin to oxidize to formic acid. Therefore, once opened, the bottle will need to be purged with nitrogen and stored frozen to prevent oxidation.
Product Profile
Molecular Formula: CH3NO
Molecular Weight: 45.04
Melting Point: 2.55 °C1
Boiling Point: 210.5 °C (760 torr; partial decomposition occurs into carbon monoxide and ammonia at atmospheric pressure starting at 180 °C)1
Molarity (neat liquid): 25.09 (based on density of 1.13 g/ml)
Specificity
This product is designated as Molecular Biology grade and is suitable for use in nucleic acid hybridizations.
Materials
 References
  1. The Merck Index, 12th ed., Entry# 4264.
  2. Molecular Cloning: A Laboratory Manual, 3rd ed., Sambrook, J. and Russell, D.W., CSHL Press (Cold Spring Harbor, NY: 2001), pp. 2.59-2.60, 6.13-6.15, 6.56, 6.58-6.60, 7.8, 7.33, 7.44, 7.45, 7.67, 12.81-12.82, 12.109-12.110.
  3. Kaabache, T., et al., Direct solution hybridization of guanidine thiocyanate-solubilized cells for quantitation of mRNAs in hepatocytes. Anal. Biochem., 232(2), 225-230 (1995).
  4. Pagratis, N. C., Rapid preparation of single stranded DNA from PCR products by streptavidin induced electrophoretic mobility shift. Nucleic Acids Res., 24(18), 3645-3646 (1996).
  5. Li, J., Elimination of polymer interference in chromatographic analysis of estradiol degradation products in a transdermal drug delivery formulation by proper selection of extraction solvents. J. Pharm. Sci., 91(8), 1873-1879 (2002).
  6. Geiser, L., et al., Potential of formamide and N-methylformamide in nonaqueous capillary electrophoresis coupled to electrospray ionization mass spectrometry. Application to the analysis of beta-blockers. J. Chromatogr. A, 979(1-2), 389-398 (2002).
Related Links