Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
DNA Preparation Options for Gene Synthesis
When ordering your gene synthesis from Sigma-Aldrich, you can choose to receive more than the standard miniprep amount (2-5 µg). Check the chart below to see typical yields from each prep scale. Additionally, you can choose your preparation as endotoxin-free. For pricing and turn-around time information, see our Gene Synthesis Pricing Page.
Prep Scale Culture Volume Typical Yield*
Mini 4mL 2-5 µg
Midi 100 mL 100 µg
Maxi 250 mL 500 µg
Mega 500 mL 1.5-2.5 mg
Giga 2.5 L 7-10 mg
*Actual yield will vary with the nature of the plasmid, bacterial hosts, and other factors.