Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Glass and Quartz Fiber Filters
Glass and Quartz Fiber FiltersGlass fiber and quartz fiber filters are depth filters that are frequently specified in contamination monitoring methods. Filters made of these materials are available with a variety of flow rates and throughput/dirt-holding capacities. Consequently, some of these filters are also ideal for prefiltration upstream of membrane filters, to prevent downstream clogging.
Our selection of depth filters includes:
  • Type APFF and AP40 glass fiber filters recommended for United States EPA Method 1311 for Toxicity Characteristic Leaching Procedure (TCLP)
  • Type AP40 glass fiber filter recommended for Total Suspended Solids (TSS) Method 2540D
  • Quartz fiber filters (Type AQFA) recommended for United States EPA PM10 ambient air monitoring method.
Glass and Quartz Fiber Disc Specifications
Membrane Material Borosilicate Glass or Quartz Microfibers
Pore Sizes (μm) 0.2 – 8.0
Thickness (μm) 230 - 1200
Water Flow Rate (mL/min/cm2) 1.2 - 6.0
Air Flow (L/min/cm2) @ 10 psi 10.6 - 139
Wettability Hydrophilic
Temperature Limitations 500 to 950 °C max
Chemical Compatibility Recommended for aqueous solvents
Sterilizability Autoclave, EtO and gamma irradiation
Surface Option White
Key Properties Performs prefiltration of particles as small as 0.6 μm, high temperature compatibility, non-shedding options, made of pure and binder-less materials
Key Applications Toxic Characteristic Leaching Procedure (TCLP), Total Suspended Solids (TSS), heavy metals analysis, prefiltration, air monitoring
Also found in Millex® Syringe Filters
Glass Fiber Filters with Binder Resin
Catalog Number Filter Code Filter Diameter (⌀) Pack Size Add to Cart
AP1502500 AP15 25 mm 100
AP1504200 AP15 42 mm 100
AP1504700 AP15 47 mm 100
AP1507500 AP15 75 mm 100
AP1509000 AP15 90 mm 100
AP1512450 AP15 124 mm 50
AP1514250 AP15 142 mm 50
AP2001300 AP20 13 mm 100
AP2002500 AP20 25 mm 100
AP2004200 AP20 42 mm 100
AP2004700 AP20 47 mm 100
AP2005500 AP20 55 mm 100
AP2007500 AP20 75 mm 100
AP2009000 AP20 90 mm 100
AP2012450 AP20 124 mm 50
AP2014250 AP20 142 mm 50
AP2501000 AP25 10 mm 100
AP2501300 AP25 13 mm 100
AP2502200 AP25 22 mm 100
AP2502500 AP25 25 mm 100
AP2504200 AP25 42 mm 100
AP2504700 AP25 47 mm 100
AP2509000 AP25 90 mm 100
AP2512450 AP25 124 mm 50
AP2514250 AP25 142 mm 50
Glass Fiber Filters without Binder Resin
Catalog Number Filter Code Filter Diameter (⌀) Pack Size Add to Cart
AP4001000 AP40 10 mm 100
AP4002405 AP40 24 mm 500
AP4002500 AP40 25 mm 100
AP4003705 AP40 37 mm 500
AP4004700 AP40 47 mm 100
AP4004705 AP40 47 mm 500
AP4007000 AP40 70 mm 100
AP4009000 AP40 90 mm 100
AP4014250 AP40 142 mm 50
APFA04700 APFA 47 mm 100
APFB02500 APFB 25 mm 100
APFB04700 APFB 47 mm 100
APFC02500 APFC 25 mm 100
APFC04700 APFC 47 mm 100
APFC09050 APFC 90 mm 50
APFD02500 APFD 25 mm 100
APFD04700 APFD 47 mm 100
APFD09050 APFD 90 mm 50
APFF02500 APFF 25 mm 100
APFF04700 APFF 47 mm 100
APFF09050 APFF 90 mm 50
APFF14250 APFF 142 mm 50
Quartz Fiber Filters
Catalog Number Filter Code Filter Diameter (⌀) Pack Size Add to Cart
AQFA03700 AQFA 37 mm 100
AQFA04700 AQFA 47 mm 100
AQFA09050 AQFA 90 mm 50
AQFA8X105 AQFA Not Applicable 50
Catalog Number Description Pack Size Add to Cart
XX6200006P Filter forceps, blunt end, stainless steel 3