Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Fluorescent Detection on SDS-PAGE Gels
Glycobiology Analysis Manual, 2nd Edition
Identify glycoproteins with superior selectivity. Allows fluorescent detection of glycoproteins directly in PAGE gels without blotting or use of antibodies.
Features and Benefits
  • Superior selectivity—enables more accurate detection of glycoproteins
  • Excellent sensitivity—allows you to detect 150 ng of glycoprotein  (5 to 30 ng carbohydrate)
  • Compatibility with both PAGE gels and PVDF membranes—allows flexibility in your applications
Materials
Related Links
Categories