Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Glycogen from Mytilus edulis (Blue mussel)
Product No. G1767
CAS RN 9005-79-2
Product Description
Sigma’s Molecular Biology Grade Glycogen is a prepared solution of glycogen from mussel in sterile redistilled water at a concentration of approx. 20 mg per ml. This preparation is purified to remove all detectable traces of nucleic acids, proteases, RNAses and DNAses. Therefore, this glycogen is a suitable carrier or co-precipitant in RNA and DNA purification.1,2 Picogram amounts of RNA or DNA can be precipitated from a volume of 0.5 ml by including 20 mg of glycogen (1 ml of solution).
Glycogen is generally preferred over tRNA, yeast RNA or sonicated DNA as a carrier, because it is less likely to interfere with downstream applications. Note that oligonucleotides as short as 20 base pairs can be recovered using linear polyacrylamide (LPA) in a DNA precipitation. Oligonucleotides as short as 8 base pairs can be recovered using glycogen.3
Precautions and Disclaimer
This product is for R&D use only, not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices. 
Storage/Stability
This solution may be stored at –20 °C for up to one year.
Product Profile
DNase, RNase and nickase: None detected.
Materials
 References
  1. Tracy, S., Prep. Biochem., 11, 251-268 (1981)
  2. Helms, C., et al., DNA, 4, 39-49 (1985)
  3. Hengen, P.N., Trends Biochem. Sci. (TiBS), 21, 224-225 (1996)
Related Links