Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
IsoQuick™ Kits for Mouse Monoclonal Isotyping
IsoQuick for Mouse Monoclonal Isotyping is fast, flexible and makes it easy to classify mouse monoclonal antibodies (MAbs)
IsoQuick is based on immunoassay technology in the format of a lateral flow device (LFD). This technology is simple, robust and proven. Isoquick is available in three separate LFD strips for maximum test flexibility and cost effectiveness. IsoQuick develops results in five minutes or less with clear, easy-to-read results and control lines. The simple interpretation guide and documentation pad make IsoQuick results easy to interpret and document in lab notebooks or project files.
Strips are ready to use directly from the tube. No extra tubes, buffers or controls are required. They can be used in cell supernatant with no dilution step. When testing ascites or purified antibody, a simple dilution step is required.
IsoQuick strips are packaged in kits with each of the three strips or they are available separately in multiples of 5 and 25 strips.
  • Flexibility—Offered in a 3-strip configuration so you only test what you need
  • Speed—Results are displayed in 5 minutes
  • Convenient and Simple—Use directly out of the tube. No dilutions required with cell supernatant or purified MAbs. Simple dilution for use with ascites
  • Accurate—Specific for each isotype/subtype and has sensitivity equal to or better than other isotype LDFs on the market
  • Cost Effective—Available individually or in kits so you're only using the tests that you need
IsoQuick for Mouse Monoclonal Isotyping is fast, flexible and makes it easy to classify mouse monoclonal antibodies (MAbs)
IsoQuick FAQs
What is isotyping and subtyping?
Isotyping is a classification of monoclonal antibodies based on the characteristics of the Immunoglobulin (Ig) in the sample. There are two formats of isotypes – heavy chain and light chain. The heavy chain includes all the Ig types and subtypes, while light chain includes the formats kappa and lambda.
Immunoglobulin has several characteristics and certain types are more specific than others. IgG (regardless of subtype) is more specific than either IgA or IgM.
What types of antibodies does this work with?
The IsoQuick strips are designed to be used in the heavy chain and light chain classification of monoclonal antibodies (MAb) made using a mouse host.
What matrices does it work with?
The IsoQuick strips will work to classify mouse monoclonal antibodies only.
What is the sensitivity of the strips?
The sensitivity for each IsoQuick strip is 0.1 µg/mL (ten nanograms per microliter)
What types of samples can I use it in?
The IsoQuick strips can be used in ascites, cell culture supernatant and purified monoclonal antibodies.
Do I need to dilute my samples?
If you are using IsoQuick strips in ascities, then you need to dilute your sample to a concentration of 1:10,000 to 1:100,000.
If you are using IsoQuick strips in cell culture supernatant no dilution is required. The strips can be used directly in your sample with a recommended sample size of 150 mL to 500 mL.
If you are using IsoQuick strips in purified MAbs, we recommend you dilute the sample to 0.05-2 mg/mL.
Why is it on three separate LFD strips?
By dividing the IsoQuick test onto three separate LFD strips, it is providing the user with the greatest flexibility both in their use of the strips and in their purchasing of strips. Because it is more common to classify MAbs at different times in the development or production process, and to classify for different isotypes during the process, the IsoQuick isotyping strips give the user the freedom to use just what is needed, when it is needed.
  • The IgG strip includes tests for heavy chain isotypes and subtypes IgG1, IgG2a, IgG2b and IgG3.
  • The IgA/M strip includes tests for heavy chain isotypes IgA and IgM.
  • The K/L strip includes tests for light chain isotypes kappa and lambda.
What other Isotypes and Subtypes are there and why do we not provide a test for them?
There are several other isotypes and subtypes that may be developed including IgD, IgE and IgG2c. These are rarely used in the development of monoclonal antibodies, and low demand for these tests resulted in their being eliminated from the IsoQuick design specification.
What are the phases in the MAb development or production process where isotyping may be used?
The process of developing a monoclonal antibody can be separated into three (3) basic phases. Phase 1 is screening. Phase 2 is Purification and Testing. Phase 3 is validating and producing.
Isotyping will be used in Phase 1 screening to help the scientist determine whether the development of the monoclonal antibody is on target with the isotype required by the specification. This will usually entail isotype testing for heavy chain types, with IgG subtypes being the mostly widely sought isotype classifications.
In Phase 2, testing will be more specific for the isotype or subtype that was specified in the development plan and that was identified in Phase 1. A change or inconsistency in isotype classification in phase 2 may mean restarting the development process or retracing steps to a known point earlier in the development cycle.
In Phase 3, testing will be done to confirm the heavy chain classification of the MAb, and to determine the light chain type as well. Light chain typing may be performed in either Phase 1 or Phase 2, but it is most often conducted as a confirmation step in Phase 3.
In the case of a new MAb that is developed both the heavy chain and light chain characteristics will be identified as part of the finished product specifications.
Is isotyping required of all MAbs that are developed or produced? Does the USDA require Isotype classification?
The USDA and other regulatory bodies may require isotype classification for certain monoclonal antibodies developed for human diagnostics or human treatments. It is not clear if other agencies require isotype classification for monoclonal antibodies not directly applied to human health care.
Who uses isotyping tests?
Any company engaged in the development, purification or production of monoclonal antibodies, including in-house labs and outsourced development or production facilities will use Isotyping tools. There are thousands of labs engaged in the development, purification and production of monoclonal antibodies. A large percentage, but not all labs developing monoclonal antibodies are using a mouse as a host.
What are the benefits of IsoQuick Mouse Monoclonal Isotyping kit versus other isotyping tests?
The IsoQuick Kit for Mouse Monoclonal Isotyping offers several advantages over other products in the market.
  • Flexibility – Dividing the core tests onto three LFD strips gives the user the greatest flexibility in purchasing and using.
  • Convenience – the IsoQuick strips are provided in simple desiccated tubes in packages of five (5) and twenty-five (25) tests.
  • Speed – IsoQuick tests develop positive test lines in five minutes.
  • Ease of Use – IsoQuick strips can be used "neat" in cell supernatant and purified MAbs. Dilution is not required, and testing can be done directly in the sample vessel. Some dilution may be required for testing in ascities, however, the same ease of use applies once the test sample is prepared – insert the IsoQuick strip and wait five minutes.
  • Accuracy – Each IsoQuick strip meets or exceeds the sensitivity specifications of other LFD isotyping products on the market including Roche strip. The ELISA plate tests may or may not be more sensitive.
  • Shelf-life – The IsoQuick strips will provide a minimum of twelve (12) months useful shelf-life. Users need not worry about over-purchasing and having tests go bad on the shelf.
How are the IsoQuick strips packaged and what is our pricing structure?
IsoQuick Mouse Monoclonal Isotyping strips will be sold in packages of five (5) strips and twenty-five (25) strips. You may purchase one or more of each of the strips in either canister size. A kit including one five-strip canister of each of the three IsoQuick strips and one twenty-five-strip canister of each of the three IsoQuick strips are also available.
Please call for specific pricing information, including volume discounts.
Why doesn't IsoQuick strips have the interpretation guide printed right on each strip?
Due to the need for specialized wide LFD production equipment and specialized printing equipment, it is not able to print the identity of each result line directly on the strip. Two interpretation tools are included in the kits that are convenient, easy to use and that will not blur or contaminate the test line. Each IsoQuick canister will have a life-size interpretation guide on the side. Additionally, each kit ordered will include a laminated interpretation and user guide for all three strips.
Products I9410 and I9535 have been discontinued; however, we still have inventory in stock.
Materials
IsoQuick is a trademark of EnviroLogix Inc.