Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
MF-Millipore™ MCE Membrane Filters
MF-Millipore™ MCE Membrane FiltersProperties of MF-Millipore™ Membrane Filter Discs make them one of the most widely and frequently used filters for analytical research and environmental monitoring.
  • Mixed cellulose esters (MCE) membranes are biologically inert (meaning that the membrane has no detectable effect on cells or biological systems into which it comes into contact).
  • MF-Millipore™ discs can be sterilized using ethylene oxide treatment, gamma irradiation, or by autoclaving
  • Discs are available in multiple pore sizes, surface options, colors, and +/- surfactant. Surfactant-free MF-Millipore™ discs have lower amounts of extractable water compared to standard discs.
Key applications of MF-Millipore™ filters:
  • Air monitoring
  • General clarification
  • Particle analysis
  • Bacteriological analysis
  • Sterilizing filtration
MF-Millipore™ Membrane Disc Specifications
Membrane Material Mixed Cellulose Esters (MCE)
Pore Sizes (μm) 0.025 - 8
Thickness (μm) 105 -150
Water Flow (mL/min/cm2) @27.5 in. Hg 20.1 - 625
Air Flow (L/min/cm2) @ 10 psi 0.12 - 68.9 (typical values)
Porosity (%) 70 - 84
Wettability Hydrophilic
Temperature Limitations 55 °C max
Refractive Index 1.50 -1.52
Protein Binding Capacity (IgG) 299 - 319 μg/cm2
Chemical Compatibility Recommended for aqueous solvents
Sterilizability Autoclave, EtO
Surface Option Black, white, gridded
Key Properties Versatile fast-flowing membrane, supports cell growth, high protein-binding
Key Applications Sterilizing filtration, air monitoring, general clarification, bacteriological analysis, drop dialysis, particle analysis, colony hybridization, nucleic acid and protein blotting, Elispot
Also used in Millex® syringe filters  
Standard MF-Millipore™ Membranes, white, plain
Catalog Number Pore Size Filter Diameter (⌀) Pack Size Add to Cart
AAWP01300 0.8 µm 13 mm 100
AAWP02500 0.8 µm 25 mm 100
AAWP03700 0.8 µm 37 mm 100
AAWP037P0 0.8 µm 37 mm 100
AAWP04700 0.8 µm 47 mm 100
AAWP0470M 0.8 µm 47 mm 50 pr
AAWP09000 0.8 µm 90 mm 100
AAWP14250 0.8 µm 142 mm 50
DAWP01300 0.65 µm 13 mm 100
DAWP02500 0.65 µm 25 mm 100
DAWP04700 0.65 µm 47 mm 100
DAWP09025 0.65 µm 90 mm 25
DAWP14250 0.65 µm 142 mm 50
GSWP01300 0.22 µm 13 mm 100
GSWP02500 0.22 µm 25 mm 100
GSWP03700 0.22 µm 37 mm 100
GSWP04700 0.22 µm 47 mm 100
GSWP09000 0.22 µm 90 mm 100
GSWP14250 0.22 µm 142 mm 50
HAWP01300 0.45 µm 13 mm 100
HAWP02400 0.45 µm 24 mm 100
HAWP02500 0.45 µm 25 mm 100
HAWP03700 0.45 µm 37 mm 100
HAWP04700 0.45 µm 47 mm 100
HAWP04700F1 0.45 µm 47 mm 100
HAWP0470M 0.45 µm 47 mm 50 pr
HAWP05000 0.45 µm 50 mm 100
HAWP09000 0.45 µm 90 mm 100
HAWP14250 0.45 µm 142 mm 50
PHWP02500 0.3 µm 25 mm 100
PHWP04700 0.3 µm 47 mm 100
PHWP09025 0.3 µm 90 mm 25
RAWP01300 1.2 µm 13 mm 100
RAWP02500 1.2 µm 25 mm 100
RAWP03700 1.2 µm 37 mm 100
RAWP04700 1.2 µm 47 mm 100
RAWP09025 1.2 µm 90 mm 25
RAWP14250 1.2 µm 142 mm 50
SCWP01300 8.0 µm 13 mm 100
SCWP02500 8.0 µm 25 mm 100
SCWP04700 8.0 µm 47 mm 100
SCWP09025 8.0 µm 90 mm 25
SCWP14250 8.0 µm 142 mm 50
SMWP01300 5.0 µm 13 mm 100
SMWP0190R 5.0 µm Not Applicable 100
SMWP02500 5.0 µm 25 mm 100
SMWP04700 5.0 µm 47 mm 100
SMWP09025 5.0 µm 90 mm 25
SMWP14250 5.0 µm 142 mm 50
SSWP01300 3.0 µm 13 mm 100
SSWP02500 3.0 µm 25 mm 100
SSWP04700 3.0 µm 47 mm 100
SSWP09025 3.0 µm 90 mm 25
SSWP14250 3.0 µm 142 mm 50
VCWP01300 0.1 µm 13 mm 100
VCWP02500 0.1 µm 25 mm 100
VCWP04700 0.1 µm 47 mm 100
VCWP09025 0.1 µm 90 mm 25
VCWP14250 0.1 µm 142 mm 50
VMWP01300 0.05 µm 13 mm 100
VMWP02500 0.05 µm 25 mm 100
VMWP04700 0.05 µm 47 mm 100
VMWP09025 0.05 µm 90 mm 25
VSWP01300 0.025 µm 13 mm 100
VSWP02500 0.025 µm 25 mm 100
VSWP04700 0.025 µm 47 mm 100
VSWP09025 0.025 µm 90 mm 25
VSWP14250 0.025 µm 142 mm 50
Standard MF-Millipore™ Membranes, white, gridded
Catalog Number Pore Size Filter Diameter (⌀) Pack Size Add to Cart
AAWG01300 0.8 µm 13 mm 100
AAWG0250C 0.8 µm 25 mm 100
AAWG03700 0.8 µm 37 mm 100
AAWG04700 0.8 µm 47 mm 100
HAWG01300 0.45 µm 13 mm 100
HAWG02500 0.45 µm 25 mm 100
HAWG03700 0.45 µm 37 mm 100
HAWG04700 0.45 µm 47 mm 100
RAWG02500 1.2 µm 25 mm 100
RAWG0250C 1.2 µm 25 mm 100
RAWG04700 1.2 µm 47 mm 100
Standard MF-Millipore™ Membranes, black, plain
Catalog Number Pore Size Filter Diameter (⌀) Pack Size Add to Cart
AABP02500 0.8 µm 25 mm 100
AABP04700 0.8 µm 47 mm 100
HABP02500 0.45 µm 25 mm 100
HABP04700 0.45 µm 47 mm 100
Standard MF-Millipore™ Membranes, black, gridded
Catalog Number Pore Size Filter Diameter (⌀) Pack Size Add to Cart
AABG01300 0.8 µm 13 mm 100
AABG02500 0.8 µm 25 mm 100
AABG04700 0.8 µm 47 mm 100
HABG01300 0.45 µm 13 mm 100
HABG02500 0.45 µm 25 mm 100
HABG04700 0.45 µm 47 mm 100
MF-Millipore™ Membranes without Triton surfactant, white, plain
Catalog Number Pore Size Filter Diameter (⌀) Pack Size Add to Cart
GSTF01300 0.22 µm 13 mm 100
GSTF02500 0.22 µm 25 mm 100
GSTF04700 0.22 µm 47 mm 100
GSTF14250 0.22 µm 142 mm 50
HATF01300 0.45 µm 13 mm 100
HATF02500 0.45 µm 25 mm 100
HATF04700 0.45 µm 47 mm 100
HATF08250 0.45 µm 82 mm 50
HATF08550 0.45 µm 85 mm 50
HATF14250 0.45 µm 142 mm 50
Accessory
Catalog Number Pore Size Pack Size Add to Cart
XX6200006P Filter forceps, blunt end, stainless steel 3