Mitex® Membrane Filters
Key Applications of Mitex® Filters
  • Air monitoring
  • Clarification of acids, bases, solvents, and cryogenic fluids
  • Hydraulic fluid analysis
Mitex® Membrane Disc Specifications
Membrane Material Hydrophobic Polytetrafluoroethylene (PTFE)
Thickness (μm) 125-200 
Water Flow (mL/min/cm2) @ 27.5 in. Hg 47 to greater than 125
Air Flow (L/min/cm2) @ 10 psi 117 - 167
Wettability Hydrophobic
Temperature Limitations 260 °C max
Chemical Compatibility Compatible with both aqueous and organic solvents
Sterilizability Autoclave, EtO
Surface Option White, plain or gridded
Key Properties Pure PTFE (unbacked), easy handling, solvent-resistant, fast-flowing, low extractables
Key Applications Clarifying acids, bases, solvents and cryogenic fluids, hydraulic fluid analysis
Materials
Product No. Description Pore Size Filter Diameter (⌀) Pack Size Add to Cart
LCWG02500 Mitex® Membrane Filter, PTFE, hydrophobic, 10 µm, 25 mm, white, gridded 10.0 µm 25 mm 100
LCWG04700 Mitex® Membrane Filter, PTFE, hydrophobic, 10 µm, 47 mm, white, gridded 10.0 µm 47 mm 100
LCWP01300 Mitex® Membrane Filter, PTFE, hydrophobic, 10 µm, 13 mm, white, plain 10.0 µm 13 mm 100
LCWP02500 Mitex® Membrane Filter, PTFE, hydrophobic, 10 µm, 25 mm, white, plain 10.0 µm 25 mm 100
P0950 Mitex® Membrane Filter, PTFE, hydrophobic, 10 µm, 47 mm, white, plain 10.0 µm 47 mm 100
LCWP09025 Mitex® Membrane Filter, PTFE, hydrophobic, 10 µm, 90 mm, white, plain 10.0 µm 90 mm 25
LCWP14250 Mitex® Membrane Filter, PTFE, hydrophobic, 10 µm, 142 mm, white, plain 10.0 µm 142 mm 50
LSWG02500 Mitex® Membrane Filter, PTFE, hydrophobic, 5.0 µm, 25 mm, white, gridded 5.0 µm 25 mm 100
LSWG04700 Mitex® Membrane Filter, PTFE, hydrophobic, 5.0 µm, 47 mm, white, gridded 5.0 µm 47 mm 100
LSWP01300 Mitex® Membrane Filter, PTFE, hydrophobic, 5.0 µm, 13 mm, white, plain 5.0 µm 13 mm 100
LSWP02500 Mitex® Membrane Filter, PTFE, hydrophobic, 5.0 µm, 25 mm, white, plain 5.0 µm 25 mm 100
LSWP03700 Mitex® Membrane Filter Monitor Refills with thin pads, PTFE membrane, 37 mm, 5.0 µm 5.0 µm 37 mm 100
LSWP04700 Mitex® Membrane Filter, PTFE, hydrophobic, 5.0 µm, 47 mm, white, plain 5.0 µm 47 mm 100
LSWP09025 Mitex® Membrane Filter, PTFE, hydrophobic, 5.0 µm, 90 mm, white, plain 5.0 µm 90 mm 25
LSWP14250 Mitex® Membrane Filter, PTFE, hydrophobic, 5.0 µm, 142 mm, white, plain 5.0 µm 142 mm 50