Mitigation of Deuterium Scrambling in Stable-Labeled Internal Standards during LC-MS MS Analysis
By: Authors: Joshua Cooper1, Billy Molloy2, Huahua Jian1, Lisa Calton2, Derrell Johnson1, Isil Dilek1, Uma Sreenivasan1, Don Cooper2, 1Cerilliant Corporation, 811 Paloma Drive, Suite A, Round Rock, TX 78665 2 Waters Corporation, Atlas Park, Simonsway, Manchester, M22 5 PP UKCONCLUSIONS
Introduction
Improvements in LC-MS/MS sensitivity and multiple reaction monitoring (MRM) capabilities are resulting in further adoption of LC-MS/MS technology in clinical settings. One specific clinical challenge with LC-MS/MS is the potential for matrix effects that cause interferences or impact ionization efficiency. As samples vary from patient to patient it can be challenging to anticipate and detect matrix effects.
Stable isotope-labeled internal standards are frequently used to compensate for matrix effects and to increase the accuracy of quantitation. A labeled internal standard that co-elutes with the drug being monitored can offset patient specific matrix effects (co-eluting concomitant medication, etc.) that may occur at the retention time of the analyte of interest.
Complications in the use of deuterium-labeled internal standards can arise from hydrogen-deuterium scrambling in solution or in the ion source at the selected transitions. In this study, we examined deuterium-labeled hormones and other compounds of clinical significance by LC-MS/MS at select transitions. We investigated reproducibility of the scrambling ratio and influences on scrambling from different LC-MS systems (tandem quadrupole vs. quadrupole time-of-flight), concentration, solution behavior, and deuterium placement in the internal standard.
Infusion Experiments of Clinically Significant Compounds Using QTof MS
  • Initial experiments were run to determine transitions and whether scrambling can occur at a selected transition.
  • Instrument: Waters Xevo G2 QTof in ESI+ mode.
  • Progesterone, Testosterone, Pregabalin, and Gabapentin (native and labeled) were infused at a concentration of 10 μg/mL in 50:50 ACN:0.1% formic acid in water at a flow rate of 20 μL/min.
  • 25-Hydroxyvitamin D2 and D3 (native and labeled) were infused at a concentration of 10 μg/mL in 60:40 MeOH:0.1% formic acid in water at a flow rate of 20 μL/min.
  • Infusion and MS parameters were optimized for signal and collision energy to give a good fragmentation pattern for each pre-cursor ion.
Compound Label Major Transitions Scrambling %Dn-1/Dn
Collision Energy
Testosterone D3 292.37->256.16 33 20
    292.37->274.18 5 20
    292.37->109.05 0 20
Progesterone D9 324.25->306.24 20 19
    324.25->288.23 77 19
    324.25->113.07 0 19
    324.25->100.07 0 19
Pregabalin D6 166.13->148.09 0 25
    166.13->130.09 0 25
    166.13->103.08 12 25
    166.13->89.07 40 25
Gabapentin D10 182.31->164.11 0 18
    182.31->147.09 0 18
25-Hydroxyvitamin D3
D6 407.37->389.23 2 12
    407.37->371.22 17 12
25-Hydroxyvitamin D2
D6 419.60->383.40 6 10
    419.60->337.33 0 10
25-Hydroxyvitamin D2
D3 416.40->380.38 12 10
    10416.40->340.35 5 10
Underlined numbers denote scrambling above 5.0%.
With initial monitoring & evaluation, transitions can be selected to minimize or eliminate deuterium scrambling in the internal standard.
Mitigation of Scrambling by Selection of an Appropriate Transition
Mitigation of Scrambling
Deuterium Scrambling in Clinically Significant Hormones from Multiple Reaction Monitoring (MRM) Experiments at High and Low Concentrations in Solution
  • Experiments were conducted on a Waters XevoTQ MS tandem quadrupole instrument using ESI+ mode. A Gradient system was used to elute the analytes from an ACQUITY BEH C18 2.1 x 50 mm column using water and methanol each containing 0.1% formic acid.
Hormones (Native & Labeled) Solution LowConcentration
(ng/mL)
High Concentration
(ng/mL)
Testosterone 1:1 Methanol:Water
0.5 20
Progesterone,Cortisol 1:1 Methanol:Water
10 500
Estradiol 1:1 Methanol:Water 1 500
25-Hydroxyvitamin D2/D3
60:40 Methanol:Water
30 500
  • Solutions were injected and MS parameters were adjusted to give optimal signal and fragmentation for each analyte.
Hormones Label Major
Transitions
Scrambling %
Dn-1/Dn
Scrambling % Dn+1/Dn
Scrambling % Dn-2/Dn
Collision Energy
Testosterone   289.25-> 96.9 0.1 0.0 NA 30
    289.25->108.9 0.1 0.0 NA 30
    289.25->253.1 0.0 NA 0.2 20
    289.25->271.1 0.0 NA 0.0 20
Testosterone D2 291.25-> 98.9
0.5 0.0 NA 30
    291.25->110.9 0.6 0.1 NA 30
    291.25->253.1 7.8 NA 0.3 20
    291.25->271.1 1.4 NA 0.0 20
Testosterone D3 292.25-> 96.9 0.8 1.3 NA 30
    292.25->108.9 0.6 1.6 NA 30
    292.25->256.1 24.4 NA 1.8 20
    292.25->274.1 3.7 NA 0.1 20
Testosterone D5 294.25-> 99.9 1.2 0.4 NA 30
    294.25->112.9 2.6 2.2 NA 30
    294.25->258.1 21.2 NA 2.7 20
    294.25->276.1 9.7 NA 0.3 20
Underlined numbers denote scrambling above 5.0%.
Hormones Label Major Transitions
Scrambling %
Dn-1/Dn
Scrambling %
Dn+1/Dn
Scrambling %
Dn-2/Dn
Collision
Energy
Progesterone   315.25-> 96.9
0.1 0.0 NA 30
    315.25->108.9 0.1 0.0 NA 30
    315.25->279.1 0.0 NA 0.3 20
    315.25->297.1 0.0 NA 0.0 20
Progesterone D9 324.25-> 99.9
1.3 0.6 NA 30
    324.25->112.9 1.9 2.4 NA 30
    324.25->288.1 40.4 NA 12.4 20
    324.25->306.1 16.1 NA 0.9 20
Estradiol   *255.15->133.0 0.1 0.0 NA 25
    *255.15->159.0 0 0.2 NA 25
Estradiol D5 *260.15->135.0 1.6 6.0 NA 25
    *260.15->161.0 1.5 8.8 NA 25
Cortisol   363.25->121.0 0.0 NA NA 25
    363.25->309.1 0.4 NA 0.1 25
    363.25->327.1 0.5 NA NA 25
    363.25->345.1 0.5 NA NA 25
Cortisol D2 365.25->123.0 48.3 NA NA 25
    365.25->311.1 8.4 NA 0.7 25
    365.25->329.1 6.6 NA NA 25
    365.25->347.1 7.5 NA NA 25
Cortisol D4 367.25-> 121.0
0.4 0.4 NA 25
    367.25->313.1 32.1 NA 10.0 25
    367.25->331.1 18.7 NA NA 25
    367.25->349.1 6.4 NA NA 25
* = Water loss used as precursor ion in Positive mode. Underlined numbers denote scrambling above 5.0%.
Optimal MRM Selection for Deuterium-labeled Internal Standards can be achieved in all cases except Cortisol-D2.
Hormones Label Major Transitions
Scrambling %
Dn-1/Dn
Scrambling %
Dn+1/Dn
Scrambling %
Dn-2/Dn
Collision
Energy
25-Hydroxyvitamin D2
  413.35->337.25 0.0 NA NA 15
    413.35->355.25 0.1 NA NA 15
    413.35->377.25 0.0 NA NA 15
    413.35->395.25 0.0 NA NA 15
25-Hydroxyvitamin D2
D3 416.35->340.25 10.1 NA NA 15
    416.35->358.25 0.4 NA NA 15
    416.35->380.25 15.0 NA NA 15
    416.35->399.25 1.7 NA NA 15
25-Hydroxyvitamin D2
D6 419.35->337.25 0.4 NA NA 15
    419.35->355.25 0.2 NA NA 15
    419.35->383.25 7.6 NA NA 15
    419.35->401.25 1.6 NA NA 15
25-Hydroxyvitamin D3
  401.35->159.0 0.2 NA NA 30
    401.35->107.0 0.1 NA NA 30
    401.35->365.25 0.1 NA NA 10
    401.35->383.25 0.0 NA NA 10
25-Hydroxyvitamin D3
D3 404.35->162.0 42.6 NA NA 30
    404.35->110.0 52.4 NA NA 30
    404.35->368.25 11.6 NA NA 10
    404.35->386.25 1.0 NA NA 10
25-Hydroxyvitamin D3
D6 407.35->159.0 0.7 NA NA 30
    407.35->107.0 1.3 NA NA 30
    407.35->371.25 20.2 NA NA 10
    407.35->389.25 4.3 NA NA 10
Underlined numbers denote scrambling above 5.0%.
Scrambling was not concentration dependant. The scrambling ratios were reproducible and consistent from high to low concentration.
Deuterium placement in the internal standard can influence level of scrambling as demonstrated by the change in scrambling percent values for different deuterium analogs of cortisol and 25-hydroxyvitamin D2/D3.
A Representative Example of Deuterium Scrambling on Tandom Quad MS: Progesterone-D9 (324->287)
Progesterone and the D9 analog were analyzed by multiple reaction monitoring (MRM) for water loss and fragment product ions using a Waters Xevo TQ MS in ESI+ mode.
Progesterone native
Progesterone native: Transition 315.25->279.1
No scrambling observed at transition 315.25->278.1
  • Peak at the expected m/z 315 -> 279 (double water loss) transition.
  • No peak observed in the corresponding scrambled transition.
Progesterone-D9
Progesterone-D9: Transition 324.25->288.1
Scrambling observed at transition 324.25->287.1
  • Scrambling observed at the m/z 324->288 (double water loss) transition for Dn-1.
Deuterium scrambling in Progesterone-D9 can be mitigated by monitoring alternative MRM transitions to the water loss – for example, 324->113.
Solution Behavior of Deuterium-Labeled Hormones by 1H NMR
  • Some deuterium-labeled internal standards contain deuterium at chemically exchangeable positions (example: Progesterone-D9).
  • An NMR experiment was conducted to understand whether the loss of deuterium was due to exchange in solution or scrambling within the LC-MS/MS.
  • Testosterone-D3(16,16,17-D3), Estradiol-D5(2,4,16,16,17-D5), and Progesterone-D9 (2,2,4,6,6,17α,21,21,21-D9) were placed in methanol-D4with 0.1% formic acid to accelerate any potential exchange and were then assessed by 1H NMR. Only Progesterone-D9 was expected to show exchange.
  • After 14 days at room temperature, only Progesterone-D9 showed deuterium exchange under the harsh acidic conditions.
  • No exchange was observed in Progesterone-D9after 10 days at room temperature in the absence of 0.1% formic acid.
  • Exchange in solution would not be expected to occur under normal diluent or LC-MS/MS analysis conditions.
Deuterium-Labeled Hormones
Progesterone-D9 in the presence of 0.1% formic acid/methanol-D4 over 2 weeks
Conclusions
  • Scrambling was observed on both tandem quad and QTof MS at select transitions for the deuterium-labeled internal standards studied. Ion source and diluent selection represent potential sources for scrambling/exchange.
  • Scrambling can be mitigated by selection of an appropriate transition with the exception of Cortisol-D2.
  • Awareness of potential scrambling is important for proper internal standard design and selection. Scrambling can be mitigatedoreliminated by altering instrument conditions and transition selection. This approach is important in clinical method development to ensure accurate quantitation and reproducible results for critical decision-making in patient care.
  • Deuterium-labeled internal standards are a viable option for LC-MS/MS analysis with selection of the appropriate transition. They also offer a more cost-effective alternative to carbon-13 or nitrogen-15-labeled analogs with benefits such as ready availability and lower cost per test.
Related Links