Nylon Membrane and Nylon Net Filters
Nylon and Nylon Net FiltersNylon is a hydrophilic yet solvent-resistant membrane, and therefore widely used for filtering
aqueous solutions as well as organic solvents. These membranes are strong and flexible.
Nylon net filters are unique because of their large, yet uniform pores. As a result, they are often
used to isolate single cells or even whole organisms (such as the worm C.elegans).
Nylon Membrane and Nylon Net Filter Disc Specifications
  Nylon Membrane Nylon Net
Filter Material Nylon Nylon
Pore Sizes (μm) 0.2 - 1.2 10 -180
Thickness (μm) 150 - 190 45 -135
Water Flow Rate (mL/min/cm2)@ 27.5 in. Hg 8.0 - 21.2 (typical value) n/a
Porosity(%) n/a 4 to 53
Wettability Hydrophilic Hydrophilic
Chemical Compatibility Compatible with both aqueous and organic solvents Compatible with both aqueous and organic solvents
Sterilizability Autoclave, EtO and gamma irradiation Autoclave, EtO and gamma irradiation
Surface Option White White
Key Properties good solvent resistance Large pore size options, uniform pore structure
Key Applications Particle removal and clarification of solvents, particle analysis Cell collection, large particulate filtration, particle analysis, flow cytometry sample prep, toxicology and drug screening on C. elegans and zebrafish
  Also found in Millex® syringe filters  
Nylon Membrane Filters
Catalog Number Pore Size Filter Diameter (⌀) Pack Size Add to Cart
GNWP02500 0.2 µm 25 mm 100
GNWP04700 0.2 µm 47 mm 100
HNWP02500 0.45 µm 25 mm 100
HNWP04700 0.45 µm 47 mm 100
Nylon Net Filters
Catalog Number Pore Size Filter Diameter (⌀) Pack Size Add to Cart
NY1100010 11.0 µm Not Applicable 1
NY1104700 11.0 µm 47 mm 100
NY1109000 11.0 µm 90 mm 50
NY1H00010 100.0 µm Not Applicable 1
NY1H02500 100.0 µm 25 mm 100
NY1H04700 100.0 µm 47 mm 100
NY1H09000 100.0 µm 90 mm 50
NY2000010 20.0 µm Not Applicable 1
NY2002500 20.0 µm 25 mm 100
NY2004700 20.0 µm 47 mm 100
NY2009000 20.0 µm 90 mm 50
NY2H02500 120.0 µm 25 mm 100
NY2H04700 120.0 µm 47 mm 100
NY3002500 10.0 µm 25 mm 100
NY3004700 10.0 µm 47 mm 100
NY3009000 10.0 µm 90 mm 50
NY4100010 41.0 µm Not Applicable 1
NY4102500 41.0 µm 25 mm 100
NY4104700 41.0 µm 47 mm 100
NY4109000 41.0 µm 90 mm 50
NY4H02500 140.0 µm 25 mm 100
NY4H04700 140.0 µm 47 mm 100
NY4H09000 140.0 µm 90 mm 50
NY6000010 60.0 µm Not Applicable 1
NY6002500 60.0 µm 25 mm 100
NY6004700 60.0 µm 47 mm 100
NY6009000 60.0 µm 90 mm 50
NY6H04700 160.0 µm 47 mm 100
NY6H09000 160.0 µm 90 mm 50
NY8002500 80.0 µm 25 mm 100
NY8004700 80.0 µm 47 mm 100
NY8009000 80.0 µm 90 mm 50
NY8H02500 180.0 µm 25 mm 100
NY8H04700 180.0 µm 47 mm 100
NY8H09000 180.0 µm 90 mm 50
Accessory
Catalog Number Description Pack Size Add to Cart
XX6200006P Filter forceps, blunt end, stainless steel 3