Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Phenol solution (equilibrated with 10mM Tris HCl, pH 8.0, with 1mM EDTA) for molecular biology
Product No. P4557
CAS RN 108-95-2
Synonyms: Equilibrated Phenol
Product Description
The product is a clear liquid capped with a layer of Tris buffer.
  • pH of phenolic phase: 6.7 ± 0.2
  • pH of phenolic phase: 7.9 ± 0.2 (after the addition of Equilibration Buffer)
 This preparation may be use in the extraction of nucleic acids.
Components
The product is supplied with Equilibration Buffer (Product No. B5658).
Precautions and Disclaimer
This product is for R&D use only, not for drug, household, or other uses. Please consult the Safety Data Sheet for information regarding hazards and safe handling practices. 
Preparation Instructions
For many applications, this product can be used without the addition of the Equilibration Buffer.
For applications requiring a higher pH, such as the isolation of large intact genomic DNA, addition of the included Equilibration Buffer is recommended. Add the entire contents of the small bottle of Equilibration Buffer to the large bottle of Phenol solution. Mix gently and allow the phases to separate before use, 2–4 hours. This will adjust the pH of the Phenol solution from pH 6.7 ± 0.2 to 7.9 ± 0.2.
Storage/Stability
Equilibrated Phenol is packaged under an inert gas and is for usable if the phenolic phase is colorless. Do not use if the phenolic phase has turned to a yellow or brown color. After the addition of Equilibration Buffer, the product may be stored for 6 months at 2–8 °C. For extended storage of the product after opening, either with or without the addition of Equilibration Buffer, –20 °C with protection from light is recommended.
Materials
 Reference
  1. Sambrook, J., and Russell, D.W., in Molecular Cloning: A Laboratory Manual, Third edition, Cold Spring Harbor Laboratory Press, (Cold Spring Harbor, New York: 2001), pp. 6.4-6.11
Related Links