Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Glutamate/GABA Synthesis and Metabolism
Glutamine is a common precursor for the biosynthesis of both glutamate and GABA. Glutamine can be transported in and out of neurons and astrocytes utilizing different glutamine carriers. Three such carriers have been cloned and characterized, referred to as ASCT2, GlnT and SN1. They are differentially expressed in brain cells; ASCT2 and SN1 being astrocytic and GlnT being neuronal. They play different roles in glutamine influx and efflux and hence control the availability of glutamine in glutamatergic and GABAergic neurons.
The neurotransmitter glutamate can be synthesized from glutamine by the action of phosphate-activated glutaminase. It appears, however, that glutamate derived from glutamine via this route is produced intramitochondrially and may subsequently undergo a transamination catalyzed by the mitochondrial isoform of aspartate aminotransferase. The α-ketoglutarate thus formed is translocated out of the mitochondria by the dicarboxylate carrier and transaminated in the cytoplasm by the cytoplasmic isoform of aspartate aminotransferase. Alternatively, glutamate may be formed from α-ketoglutarate and alanine catalyzed by alanine aminotransferase. This cytoplasmic glutamate is transported into vesicles by vesicular glutamate transporters. Three vesicular glutamate transporters have been cloned and they exhibit differential expression in glutamatergic neurons in various brain regions. This has important implications with regard to characterization of subpopulations of glutamatergic neurons. Glutamate metabolism, which to a large extent takes place in astroglial cells, is catalyzed either by glutamine synthetase or glutamate dehydrogenase.
The inhibitors for the enzymes involved in glutamate biosynthesis are not absolutely specific. This is particularly serious for aminooxyacetic acid, which at high concentrations will inhibit all pyridoxal phosphate-dependent enzymes. Another problem with amino-oxyacetic acid is that it potently inhibits both glutamate decarboxylase and GABA-transaminase (see Table below). Even methionine sulfoximine, which is proven to be an extremely useful tool to study the functional importance of glutamine synthetase, is not strictly specific for this enzyme, but also inhibits, for example, α glutamylcysteine synthetase, a key enzyme in the biosynthesis of glutathione. Therefore, these inhibitors must be used with caution.
The neurotransmitter GABA is formed from glutamate by the action of glutamate decarboxylase. It appears that glutamine serves as the precursor for glutamate, making phosphate-activated glutaminase, an important enzyme for GABA synthesis as well. Recent studies of these processes, using [13C]-labeled substrates and [13C] NMR spectroscopy to follow the metabolic fate of individual C-atoms, have suggested that this biosynthetic route is somewhat more complex than previously thought. It appears that glutamate formed from glutamine may be metabolized in the tricarboxylic acid (TCA) cycle prior to its conversion to GABA, which may allow new alternative regulatory mechanisms. Moreover, it appears that this pathway involving TCA cycle activity is differentially involved in the biosynthesis of GABA destined for the cytoplasmic and vesicular pools, respectively. GABA is metabolized by the action of GABA-transaminase, which is a ubiquitous enzyme being present in GABAergic neurons as well as other types of neurons and astrocytes. Inhibitors of this enzyme generally exhibit anticonvulsant actions.
Inhibitors of glutamate decarboxylase generally also inhibit GABA-transaminase due to the fact that both enzymes require pyridoxal phosphate for activity. However, in the case of the carbonyl-trapping agent aminooxyacetic acid its Ki value for inhibition of GABA-transaminase is 10-fold lower than that for glutamate decarboxylase. The two catalytic site directed suicide inhibitors of GABA-transaminase, γ-vinyl GABA and GABAculine, are excellent inhibitors of this enzyme in terms of specificity and potency.
The Table below contains accepted modulators and additional information. For a list of additional products, see the "Similar Products" section below.
Glutamate
Compound Enzyme Co-Factors/Substrate Inhibitors
L-Glutamine (G3126)
│                         │––––––––––––––→
Glutaminase (G5894)
Phosphate (S0751, S0876)
L-Glutamine (G3126)
L-Glutamate (G1626, G1251)
Ammonia (221228)
6-Diazo-5-oxo-L-nor-leucine
L-Glutamine (G3126)
L-Glutamate (M) (G1626, G1251)
│                         │–––––––––––––––→
Aspartate aminotransferase (m) (G2751)
Pyridoxal phosphate (P9255)
Oxaloacetate (O4126)
Aspartate (A9256)
Aminooxyacetic acid (C13408)
α-Ketoglutarate (m) (K1750)
│                         │–––––––––––––––→
Dicarboxylate carrier
Malate (02300)
Phenylsuccinate (P35200)
α-Ketoglutarate (c) (K1750)
│                         │–––––––––––––––→
Aspartate aminotransferase (c) (G2751)
Pyridoxal phosphate (P9255)
Oxaloacetate (O4126)
Aspartate (A9256)
Aminooxyacetic acid (C13408)
L-Glutamate (c-ves) (G1626, G1251)
│                         │–––––––––––––––→
Vesicular glutamate carrier
(A) α-Ketoglutarate (K1750) + NH4+ (221228)
│                         │–––––––––––––––→
L-Glutamate dehydrogenase (G2626)
NAD+ (N7004)
L-Glutamate (G1626, G1251)
(B) L-Glutamine (G3126)
│                         │–––––––––––––––→
Glutamine synthetase (G1270)
ATP (A2383)
NH4+ (221228)
L-Glutamate (G1626, G1251)
Methionine sulfoximine (M5379)
GABA
Compound Enzyme Co-Factors/Substrate Inhibitors
L-Glutamate (G1626, G1251)
│                         │–––––––––––––––→
L-Glutamate decarboxylase
Pyridoxal phosphate (P9255)
L-Glutamate (G1626, G1251)
Aminooxyacetic acid (C13408)
3-Mercaptopropionic acid (M5801)
γ-Acetylenic GABA (A230)
GABA (A2129)
│                         │–––––––––––––––→
GABA-transaminase
Pyridoxal phosphate (P9255)
α-Ketoglutarate (K1750)
Aminooxyacetic acid (C13408)
3-Mercaptopropionic acid (M5801)
γ-Vinyl GABA (V8261)
GABAculine (A3539)
Succinate semialdehyde
│                         │–––––––––––––––→
Succinate semialdehyde dehydrogenase
NADH (N8129)
Succinate semialdehyde
Succinate (S7501)
Appendix
Enzyme Tissue expression Function Disease
Glutaminase (G5894)
Kidney, brain and other tissues Generation of Ammonia and Glutamate Not known
Aspartate aminotransferase (G2751)
Liver, brain and other tissues Malate-aspartate shuttle Not known
L-Glutamate dehydrogenase (G2626)
Brain, liver, muscle and other tissues Amonia-fixation and -generation Olivoponto-cerebellar atrophy
Glutamine synthetase (G1270)
Brain, liver and other tissues Amonia-fixation Not known
L-Glutamate decarboxylase Brain, pancreas and intestine GABA synthesis Epilepsy, schizophrenia
GABA-transaminase Brain, liver and other tissues GABA metabolism Epilepsy
Succinate semialdehyde dehydrogenase Brain, liver and other tissues GABA metabolism (GABA-shunt) Not known
Abbreviations
c: Cytoplasmic
c-ves: Vesicles in cytoplasm
m: Mitochondrial
Similar Products
References